Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15975742-15976281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2B
Entrez Gene ID
136
Gene Name
adenosine A2b receptor
Gene Aliases
ADORA2
UniGene
Hs.167046
Gene Chromosome Location
Chr.17: 15927781-15975896 on Build GRCh38
Transcripts
NM_000676.2 XM_011523659.2 XM_017024197.1

SNP Detail

SNP ID Chromosome Location
rs560298271 Chr.17: 15975810-15975810 on Build GRCh38
rs28562527 Chr.17: 15976081-15976081 on Build GRCh38
rs150467006 Chr.17: 15975859-15975859 on Build GRCh38
rs4989137 Chr.17: 15976084-15976084 on Build GRCh38
rs781546979 Chr.17: 15976092-15976092 on Build GRCh38
rs927234717 Chr.17: 15975936-15975936 on Build GRCh38
rs770592198 Chr.17: 15976068-15976071 on Build GRCh38
rs867530602 Chr.17: 15976076-15976076 on Build GRCh38
rs529222520 Chr.17: 15976145-15976145 on Build GRCh38
rs938631817 Chr.17: 15975958-15975958 on Build GRCh38
rs367849107 Chr.17: 15976114-15976114 on Build GRCh38
rs940155183 Chr.17: 15975837-15975837 on Build GRCh38
rs565273342 Chr.17: 15975997-15975997 on Build GRCh38
rs775951238 Chr.17: 15976117-15976147 on Build GRCh38
rs537935140 Chr.17: 15976196-15976196 on Build GRCh38
rs952566933 Chr.17: 15975907-15975907 on Build GRCh38
rs373700533 Chr.17: 15976106-15976106 on Build GRCh38
rs58700066 Chr.17: 15976141-15976142 on Build GRCh38
rs536021996 Chr.17: 15976145-15976147 on Build GRCh38
rs746086111 Chr.17: 15976120-15976121 on Build GRCh38
rs903907299 Chr.17: 15976247-15976247 on Build GRCh38
rs574155512 Chr.17: 15975953-15975953 on Build GRCh38
rs562757937 Chr.17: 15975985-15975985 on Build GRCh38
rs1860644 Chr.17: 15976020-15976020 on Build GRCh38
rs7503100 Chr.17: 15976108-15976108 on Build GRCh38
rs113256621 Chr.17: 15976146-15976146 on Build GRCh38
rs4989136 Chr.17: 15976080-15976080 on Build GRCh38
rs745746625 Chr.17: 15975963-15975988 on Build GRCh38
rs549223710 Chr.17: 15975794-15975794 on Build GRCh38
rs534290139 Chr.17: 15975851-15975851 on Build GRCh38
rs961465032 Chr.17: 15975877-15975877 on Build GRCh38
rs569074237 Chr.17: 15976177-15976177 on Build GRCh38
rs912970539 Chr.17: 15976254-15976254 on Build GRCh38
rs955778081 Chr.17: 15975799-15975799 on Build GRCh38
rs1860643 Chr.17: 15975989-15975989 on Build GRCh38
rs56901517 Chr.17: 15976142-15976143 on Build GRCh38
rs200330192 Chr.17: 15976137-15976147 on Build GRCh38
rs537951424 Chr.17: 15975852-15975852 on Build GRCh38
rs565438204 Chr.17: 15975807-15975807 on Build GRCh38
rs9903298 Chr.17: 15976122-15976122 on Build GRCh38
rs57899385 Chr.17: 15976097-15976097 on Build GRCh38
rs748326146 Chr.17: 15976096-15976096 on Build GRCh38
rs904035267 Chr.17: 15975819-15975819 on Build GRCh38
rs577610422 Chr.17: 15975867-15975867 on Build GRCh38
rs568940358 Chr.17: 15975806-15975806 on Build GRCh38
rs1057222543 Chr.17: 15976225-15976225 on Build GRCh38
rs2482987 Chr.17: 15976086-15976086 on Build GRCh38
rs527719801 Chr.17: 15976003-15976003 on Build GRCh38
rs200730273 Chr.17: 15976068-15976068 on Build GRCh38
rs868709255 Chr.17: 15976100-15976100 on Build GRCh38
rs1001094031 Chr.17: 15975826-15975826 on Build GRCh38
rs59288887 Chr.17: 15976060-15976060 on Build GRCh38
rs972529309 Chr.17: 15975890-15975890 on Build GRCh38
rs553688641 Chr.17: 15976248-15976248 on Build GRCh38
rs112799568 Chr.17: 15976147-15976147 on Build GRCh38
rs9709247 Chr.17: 15976110-15976110 on Build GRCh38
rs919734512 Chr.17: 15975904-15975904 on Build GRCh38
rs866576325 Chr.17: 15976104-15976104 on Build GRCh38
rs891659531 Chr.17: 15976018-15976018 on Build GRCh38
rs772136682 Chr.17: 15976124-15976125 on Build GRCh38
rs201613912 Chr.17: 15976139-15976147 on Build GRCh38
rs373907623 Chr.17: 15975876-15975876 on Build GRCh38
rs4989142 Chr.17: 15976098-15976098 on Build GRCh38
rs1011336657 Chr.17: 15976019-15976019 on Build GRCh38
rs2324083 Chr.17: 15976142-15976142 on Build GRCh38
rs57070911 Chr.17: 15976098-15976099 on Build GRCh38
rs188058763 Chr.17: 15975978-15975978 on Build GRCh38
rs975794862 Chr.17: 15975797-15975797 on Build GRCh38
rs576085660 Chr.17: 15975988-15975988 on Build GRCh38
rs1028995336 Chr.17: 15975845-15975845 on Build GRCh38
rs868077093 Chr.17: 15976155-15976155 on Build GRCh38
rs58979803 Chr.17: 15976097-15976098 on Build GRCh38
rs9303133 Chr.17: 15976102-15976102 on Build GRCh38
rs867265536 Chr.17: 15976094-15976094 on Build GRCh38
rs891577110 Chr.17: 15975798-15975798 on Build GRCh38
rs1860645 Chr.17: 15976030-15976030 on Build GRCh38
rs554199859 Chr.17: 15975928-15975928 on Build GRCh38
rs1056874326 Chr.17: 15975774-15975774 on Build GRCh38
rs1043806934 Chr.17: 15976171-15976174 on Build GRCh38
rs755033858 Chr.17: 15976088-15976088 on Build GRCh38

View Full Product Details