Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16348524-16349026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPV
Entrez Gene ID
201161
Gene Name
centromere protein V
Gene Aliases
3110013H01Rik, CENP-V, PRR6, p30
UniGene
Hs.433422
Gene Chromosome Location
Chr.17: 16342534-16353498 on Build GRCh38
Transcripts
NM_181716.2 XM_017025349.1 XM_017025350.1 XM_017025352.1 XM_017025353.1 XM_017025354.1 XM_017025355.1 XM_017025356.1

Gene Symbol
PIGL
Entrez Gene ID
9487
Gene Name
phosphatidylinositol glycan anchor biosynthesis class L
Gene Aliases
CHIME
UniGene
Hs.499793
Gene Chromosome Location
Chr.17: 16217195-16351800 on Build GRCh38
Transcripts
NM_181716.2 XM_017025349.1 XM_017025350.1 XM_017025352.1 XM_017025353.1 XM_017025354.1 XM_017025355.1 XM_017025356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609878 c.536A>G p.N179S Chr.17: 16348659-16348659 on Build GRCh38
7180973 c.528G>A p.K176K Chr.17: 16348667-16348667 on Build GRCh38
6626708 c.558T>G p.A186A Chr.17: 16348637-16348637 on Build GRCh38
181765 c.563G>A p.R188H Chr.17: 16348632-16348632 on Build GRCh38
5006118 c.528_529insA p.Q178fs*3 Chr.17: 16348666-16348667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924351877 Chr.17: 16348840-16348840 on Build GRCh38
rs755046971 Chr.17: 16348770-16348770 on Build GRCh38
rs766431138 Chr.17: 16348931-16348931 on Build GRCh38
rs757551199 Chr.17: 16348568-16348568 on Build GRCh38
rs998106484 Chr.17: 16348942-16348942 on Build GRCh38
rs747316123 Chr.17: 16348599-16348599 on Build GRCh38
rs1052657504 Chr.17: 16348865-16348865 on Build GRCh38
rs750986657 Chr.17: 16348739-16348739 on Build GRCh38
rs897526404 Chr.17: 16348872-16348872 on Build GRCh38
rs565008939 Chr.17: 16348578-16348578 on Build GRCh38
rs967268457 Chr.17: 16348769-16348769 on Build GRCh38
rs758546868 Chr.17: 16348590-16348590 on Build GRCh38
rs1001625816 Chr.17: 16348560-16348560 on Build GRCh38
rs994422556 Chr.17: 16348876-16348876 on Build GRCh38
rs750609187 Chr.17: 16348685-16348685 on Build GRCh38
rs760933806 Chr.17: 16348930-16348930 on Build GRCh38
rs769659925 Chr.17: 16348642-16348642 on Build GRCh38
rs899856512 Chr.17: 16348938-16348938 on Build GRCh38
rs374278548 Chr.17: 16348719-16348719 on Build GRCh38
rs753983349 Chr.17: 16348949-16348949 on Build GRCh38
rs1035971125 Chr.17: 16348859-16348859 on Build GRCh38
rs758815803 Chr.17: 16348747-16348747 on Build GRCh38
rs529692068 Chr.17: 16348793-16348793 on Build GRCh38
rs776930185 Chr.17: 16348609-16348609 on Build GRCh38
rs569598454 Chr.17: 16348981-16348981 on Build GRCh38
rs140194188 Chr.17: 16348638-16348638 on Build GRCh38
rs888584101 Chr.17: 16348925-16348925 on Build GRCh38
rs773310097 Chr.17: 16348649-16348649 on Build GRCh38
rs200499958 Chr.17: 16348851-16348856 on Build GRCh38
rs779977221 Chr.17: 16348595-16348595 on Build GRCh38
rs1030161488 Chr.17: 16348947-16348947 on Build GRCh38
rs911657041 Chr.17: 16348839-16348839 on Build GRCh38
rs773719329 Chr.17: 16348672-16348672 on Build GRCh38
rs998242899 Chr.17: 16348561-16348562 on Build GRCh38
rs2286666 Chr.17: 16348545-16348545 on Build GRCh38
rs370851181 Chr.17: 16348637-16348637 on Build GRCh38
rs762197295 Chr.17: 16348726-16348726 on Build GRCh38
rs751994315 Chr.17: 16348697-16348698 on Build GRCh38
rs913129417 Chr.17: 16348713-16348713 on Build GRCh38
rs1004694691 Chr.17: 16348979-16348979 on Build GRCh38
rs747973484 Chr.17: 16348626-16348626 on Build GRCh38
rs376117303 Chr.17: 16349001-16349001 on Build GRCh38
rs373451091 Chr.17: 16348600-16348600 on Build GRCh38
rs931845959 Chr.17: 16348819-16348819 on Build GRCh38
rs567030421 Chr.17: 16348748-16348748 on Build GRCh38
rs1049161761 Chr.17: 16348919-16348919 on Build GRCh38
rs767161153 Chr.17: 16348702-16348702 on Build GRCh38
rs944353431 Chr.17: 16348841-16348841 on Build GRCh38
rs759299317 Chr.17: 16348683-16348683 on Build GRCh38
rs751591786 Chr.17: 16348749-16348749 on Build GRCh38
rs915604770 Chr.17: 16348792-16348792 on Build GRCh38
rs548702393 Chr.17: 16348632-16348632 on Build GRCh38
rs1050141182 Chr.17: 16348832-16348832 on Build GRCh38
rs370067633 Chr.17: 16348569-16348569 on Build GRCh38
rs893953428 Chr.17: 16348871-16348871 on Build GRCh38
rs992840490 Chr.17: 16348596-16348596 on Build GRCh38
rs1045522306 Chr.17: 16348830-16348830 on Build GRCh38
rs2286665 Chr.17: 16348795-16348795 on Build GRCh38
rs765555443 Chr.17: 16348728-16348728 on Build GRCh38
rs951532441 Chr.17: 16348958-16348958 on Build GRCh38
rs527525714 Chr.17: 16348601-16348601 on Build GRCh38
rs762765606 Chr.17: 16348655-16348655 on Build GRCh38
rs1034409584 Chr.17: 16348975-16348975 on Build GRCh38
rs765959647 Chr.17: 16348666-16348666 on Build GRCh38
rs545256555 Chr.17: 16348554-16348554 on Build GRCh38

View Full Product Details