Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16353273-16353761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPV
Entrez Gene ID
201161
Gene Name
centromere protein V
Gene Aliases
3110013H01Rik, CENP-V, PRR6, p30
UniGene
Hs.433422
Gene Chromosome Location
Chr.17: 16342534-16353498 on Build GRCh38
Transcripts
NM_181716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7220858 c.82G>C p.A28P Chr.17: 16353355-16353355 on Build GRCh38
1609879 c.79T>C p.S27P Chr.17: 16353358-16353358 on Build GRCh38
5446031 c.51_52insTCCGGGGCC p.S21_A22insGAS Chr.17: 16353385-16353386 on Build GRCh38
4129614 c.65C>G p.A22G Chr.17: 16353372-16353372 on Build GRCh38
5957947 c.40G>T p.G14W Chr.17: 16353397-16353397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006491298 Chr.17: 16353615-16353615 on Build GRCh38
rs1052919981 Chr.17: 16353735-16353735 on Build GRCh38
rs145798583 Chr.17: 16353591-16353591 on Build GRCh38
rs969615446 Chr.17: 16353644-16353644 on Build GRCh38
rs936617332 Chr.17: 16353535-16353535 on Build GRCh38
rs950698625 Chr.17: 16353327-16353327 on Build GRCh38
rs12951110 Chr.17: 16353357-16353357 on Build GRCh38
rs902016530 Chr.17: 16353433-16353433 on Build GRCh38
rs889465640 Chr.17: 16353555-16353555 on Build GRCh38
rs1019334620 Chr.17: 16353430-16353430 on Build GRCh38
rs917967255 Chr.17: 16353525-16353525 on Build GRCh38
rs772935654 Chr.17: 16353388-16353388 on Build GRCh38
rs768049448 Chr.17: 16353385-16353386 on Build GRCh38
rs939537245 Chr.17: 16353712-16353712 on Build GRCh38
rs1041194745 Chr.17: 16353585-16353585 on Build GRCh38
rs1016490335 Chr.17: 16353661-16353661 on Build GRCh38
rs747816930 Chr.17: 16353361-16353361 on Build GRCh38
rs1016290299 Chr.17: 16353457-16353457 on Build GRCh38
rs950979198 Chr.17: 16353451-16353451 on Build GRCh38
rs984829824 Chr.17: 16353338-16353338 on Build GRCh38
rs776046857 Chr.17: 16353358-16353359 on Build GRCh38
rs1053415209 Chr.17: 16353404-16353404 on Build GRCh38
rs573928905 Chr.17: 16353534-16353534 on Build GRCh38
rs903668338 Chr.17: 16353473-16353492 on Build GRCh38
rs999250068 Chr.17: 16353530-16353531 on Build GRCh38
rs942349540 Chr.17: 16353564-16353564 on Build GRCh38
rs1043529116 Chr.17: 16353350-16353367 on Build GRCh38
rs1053609323 Chr.17: 16353542-16353542 on Build GRCh38
rs540495470 Chr.17: 16353340-16353340 on Build GRCh38
rs985021514 Chr.17: 16353454-16353454 on Build GRCh38
rs747644694 Chr.17: 16353363-16353363 on Build GRCh38
rs754483275 Chr.17: 16353358-16353358 on Build GRCh38
rs959372180 Chr.17: 16353472-16353472 on Build GRCh38
rs977926559 Chr.17: 16353532-16353532 on Build GRCh38
rs909206158 Chr.17: 16353345-16353353 on Build GRCh38
rs1047954327 Chr.17: 16353598-16353598 on Build GRCh38
rs902476817 Chr.17: 16353377-16353377 on Build GRCh38
rs1026482631 Chr.17: 16353450-16353450 on Build GRCh38
rs889433286 Chr.17: 16353426-16353426 on Build GRCh38
rs578198570 Chr.17: 16353323-16353323 on Build GRCh38
rs539910742 Chr.17: 16353372-16353373 on Build GRCh38
rs780735851 Chr.17: 16353360-16353360 on Build GRCh38
rs928113552 Chr.17: 16353694-16353694 on Build GRCh38
rs560390150 Chr.17: 16353363-16353363 on Build GRCh38
rs900961098 Chr.17: 16353587-16353587 on Build GRCh38
rs990895753 Chr.17: 16353355-16353355 on Build GRCh38
rs538010702 Chr.17: 16353683-16353683 on Build GRCh38
rs976756584 Chr.17: 16353299-16353299 on Build GRCh38
rs886582069 Chr.17: 16353613-16353613 on Build GRCh38
rs936583008 Chr.17: 16353385-16353385 on Build GRCh38
rs769584320 Chr.17: 16353365-16353372 on Build GRCh38
rs1006300536 Chr.17: 16353506-16353506 on Build GRCh38
rs997655810 Chr.17: 16353438-16353438 on Build GRCh38
rs970864449 Chr.17: 16353723-16353723 on Build GRCh38
rs149829221 Chr.17: 16353372-16353372 on Build GRCh38
rs562885252 Chr.17: 16353611-16353611 on Build GRCh38
rs1012285808 Chr.17: 16353705-16353705 on Build GRCh38
rs188652843 Chr.17: 16353367-16353367 on Build GRCh38

View Full Product Details