Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27885203-27885708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
UniGene
Hs.10649
Gene Chromosome Location
Chr.1: 27872531-27886685 on Build GRCh38
Transcripts
NM_001039477.2 NM_001105556.2 NM_001286113.1 NM_001286115.1 NM_004848.3 XM_005246041.3 XM_006711050.1 XM_011542445.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7170725 c.748G>T p.D250Y Chr.1: 27885396-27885396 on Build GRCh38
907932 c.668C>T p.A223V Chr.1: 27885316-27885316 on Build GRCh38
426024 c.1871A>T p.D624V Chr.1: 27885446-27885446 on Build GRCh38
7170724 c.1821G>T p.K607N Chr.1: 27885396-27885396 on Build GRCh38
907931 c.1741C>T p.Q581* Chr.1: 27885316-27885316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751933529 Chr.1: 27885304-27885304 on Build GRCh38
rs367554537 Chr.1: 27885426-27885426 on Build GRCh38
rs981939649 Chr.1: 27885587-27885587 on Build GRCh38
rs767891402 Chr.1: 27885419-27885419 on Build GRCh38
rs907645790 Chr.1: 27885596-27885596 on Build GRCh38
rs890641882 Chr.1: 27885367-27885367 on Build GRCh38
rs755384916 Chr.1: 27885310-27885310 on Build GRCh38
rs544599415 Chr.1: 27885551-27885551 on Build GRCh38
rs563532099 Chr.1: 27885623-27885623 on Build GRCh38
rs747770523 Chr.1: 27885471-27885471 on Build GRCh38
rs1035453824 Chr.1: 27885526-27885526 on Build GRCh38
rs781326582 Chr.1: 27885321-27885321 on Build GRCh38
rs763418997 Chr.1: 27885303-27885303 on Build GRCh38
rs867568376 Chr.1: 27885496-27885496 on Build GRCh38
rs750893121 Chr.1: 27885302-27885302 on Build GRCh38
rs144780385 Chr.1: 27885468-27885468 on Build GRCh38
rs758209141 Chr.1: 27885657-27885657 on Build GRCh38
rs185853171 Chr.1: 27885266-27885266 on Build GRCh38
rs779786821 Chr.1: 27885463-27885463 on Build GRCh38
rs970999166 Chr.1: 27885515-27885515 on Build GRCh38
rs149827349 Chr.1: 27885348-27885348 on Build GRCh38
rs377661678 Chr.1: 27885325-27885325 on Build GRCh38
rs1012775365 Chr.1: 27885554-27885554 on Build GRCh38
rs551480656 Chr.1: 27885229-27885229 on Build GRCh38
rs771825849 Chr.1: 27885291-27885292 on Build GRCh38
rs769635654 Chr.1: 27885487-27885487 on Build GRCh38
rs776378400 Chr.1: 27885343-27885343 on Build GRCh38
rs916432349 Chr.1: 27885227-27885227 on Build GRCh38
rs772763797 Chr.1: 27885503-27885503 on Build GRCh38
rs1062813 Chr.1: 27885326-27885326 on Build GRCh38
rs745628507 Chr.1: 27885264-27885264 on Build GRCh38
rs768572278 Chr.1: 27885340-27885340 on Build GRCh38
rs1048078307 Chr.1: 27885421-27885421 on Build GRCh38
rs777829767 Chr.1: 27885433-27885433 on Build GRCh38
rs200906638 Chr.1: 27885409-27885409 on Build GRCh38
rs780025757 Chr.1: 27885365-27885367 on Build GRCh38
rs748304300 Chr.1: 27885588-27885588 on Build GRCh38
rs147739833 Chr.1: 27885427-27885427 on Build GRCh38
rs1050692096 Chr.1: 27885358-27885358 on Build GRCh38
rs777965598 Chr.1: 27885466-27885466 on Build GRCh38
rs960105713 Chr.1: 27885539-27885539 on Build GRCh38
rs1000153809 Chr.1: 27885422-27885422 on Build GRCh38
rs61744137 Chr.1: 27885324-27885324 on Build GRCh38
rs183024802 Chr.1: 27885265-27885265 on Build GRCh38
rs371915338 Chr.1: 27885386-27885386 on Build GRCh38
rs763894590 Chr.1: 27885246-27885246 on Build GRCh38
rs1023536758 Chr.1: 27885491-27885491 on Build GRCh38
rs373684077 Chr.1: 27885408-27885408 on Build GRCh38
rs778581261 Chr.1: 27885250-27885250 on Build GRCh38
rs757368674 Chr.1: 27885460-27885460 on Build GRCh38
rs1022861837 Chr.1: 27885631-27885631 on Build GRCh38
rs753926351 Chr.1: 27885449-27885449 on Build GRCh38
rs571140318 Chr.1: 27885230-27885230 on Build GRCh38
rs368016870 Chr.1: 27885329-27885329 on Build GRCh38
rs200183022 Chr.1: 27885467-27885467 on Build GRCh38
rs141747760 Chr.1: 27885382-27885382 on Build GRCh38
rs764867238 Chr.1: 27885291-27885291 on Build GRCh38
rs763510634 Chr.1: 27885387-27885387 on Build GRCh38
rs529216604 Chr.1: 27885681-27885681 on Build GRCh38
rs536413633 Chr.1: 27885328-27885328 on Build GRCh38

View Full Product Details