Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28691461-28691874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
UniGene
Hs.632373
Gene Chromosome Location
Chr.1: 28668728-28715603 on Build GRCh38
Transcripts
NM_001319674.1 NM_006582.3 NM_024482.2 XM_011540518.2 XM_011540519.2 XM_011540521.2 XM_011540522.2 XM_017000087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360819 c.315A>T p.G105G Chr.1: 28691688-28691688 on Build GRCh38
6671971 c.336+2T>C p.? Chr.1: 28691711-28691711 on Build GRCh38
5552567 c.238G>C p.D80H Chr.1: 28691611-28691611 on Build GRCh38
5779891 c.354G>A p.V118V Chr.1: 28691697-28691697 on Build GRCh38
5552566 c.268G>C p.D90H Chr.1: 28691611-28691611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912943491 Chr.1: 28691761-28691761 on Build GRCh38
rs937243526 Chr.1: 28691799-28691799 on Build GRCh38
rs762177732 Chr.1: 28691604-28691604 on Build GRCh38
rs1028191066 Chr.1: 28691555-28691558 on Build GRCh38
rs374526303 Chr.1: 28691804-28691805 on Build GRCh38
rs371609132 Chr.1: 28691672-28691672 on Build GRCh38
rs185884040 Chr.1: 28691725-28691725 on Build GRCh38
rs770199182 Chr.1: 28691606-28691606 on Build GRCh38
rs528350265 Chr.1: 28691798-28691799 on Build GRCh38
rs1004851926 Chr.1: 28691568-28691568 on Build GRCh38
rs201596426 Chr.1: 28691694-28691694 on Build GRCh38
rs751746178 Chr.1: 28691643-28691643 on Build GRCh38
rs771795991 Chr.1: 28691565-28691565 on Build GRCh38
rs746587161 Chr.1: 28691596-28691596 on Build GRCh38
rs925642014 Chr.1: 28691787-28691787 on Build GRCh38
rs745543228 Chr.1: 28691560-28691560 on Build GRCh38
rs746165929 Chr.1: 28691489-28691489 on Build GRCh38
rs967108550 Chr.1: 28691763-28691763 on Build GRCh38
rs762768161 Chr.1: 28691574-28691575 on Build GRCh38
rs71586854 Chr.1: 28691810-28691813 on Build GRCh38
rs200962078 Chr.1: 28691805-28691808 on Build GRCh38
rs778536677 Chr.1: 28691552-28691552 on Build GRCh38
rs577018963 Chr.1: 28691531-28691531 on Build GRCh38
rs779843026 Chr.1: 28691592-28691592 on Build GRCh38
rs750247034 Chr.1: 28691740-28691740 on Build GRCh38
rs543902062 Chr.1: 28691848-28691851 on Build GRCh38
rs963597578 Chr.1: 28691662-28691662 on Build GRCh38
rs773390447 Chr.1: 28691611-28691611 on Build GRCh38
rs532047660 Chr.1: 28691800-28691800 on Build GRCh38
rs1014076895 Chr.1: 28691503-28691503 on Build GRCh38
rs143019580 Chr.1: 28691783-28691783 on Build GRCh38
rs978369274 Chr.1: 28691767-28691767 on Build GRCh38
rs779894528 Chr.1: 28691760-28691760 on Build GRCh38
rs777245328 Chr.1: 28691603-28691603 on Build GRCh38
rs766289029 Chr.1: 28691752-28691752 on Build GRCh38
rs753693030 Chr.1: 28691541-28691541 on Build GRCh38
rs568295814 Chr.1: 28691808-28691809 on Build GRCh38
rs758265525 Chr.1: 28691759-28691759 on Build GRCh38
rs766652452 Chr.1: 28691614-28691614 on Build GRCh38
rs757094352 Chr.1: 28691542-28691542 on Build GRCh38
rs376853603 Chr.1: 28691695-28691695 on Build GRCh38
rs987623028 Chr.1: 28691734-28691734 on Build GRCh38
rs759658289 Chr.1: 28691679-28691679 on Build GRCh38
rs757080657 Chr.1: 28691697-28691697 on Build GRCh38
rs367810886 Chr.1: 28691602-28691602 on Build GRCh38
rs1046511871 Chr.1: 28691538-28691538 on Build GRCh38
rs757234187 Chr.1: 28691613-28691613 on Build GRCh38
rs1016283762 Chr.1: 28691652-28691652 on Build GRCh38
rs944712594 Chr.1: 28691828-28691828 on Build GRCh38

View Full Product Details