Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28710403-28710932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
UniGene
Hs.632373
Gene Chromosome Location
Chr.1: 28668728-28715603 on Build GRCh38
Transcripts
NM_001319674.1 NM_006582.3 NM_024482.2 XM_011540518.2 XM_011540519.2 XM_011540521.2 XM_011540522.2 XM_017000087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5040360 c.982G>A p.V328I Chr.1: 28710603-28710603 on Build GRCh38
6671974 c.980delA p.Y327fs*3 Chr.1: 28710631-28710631 on Build GRCh38
7171681 c.954G>A p.K318K Chr.1: 28710575-28710575 on Build GRCh38
5040361 c.952G>A p.V318I Chr.1: 28710603-28710603 on Build GRCh38
6487133 c.882C>G p.L294L Chr.1: 28710533-28710533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575055406 Chr.1: 28710820-28710820 on Build GRCh38
rs762225313 Chr.1: 28710676-28710676 on Build GRCh38
rs1046014199 Chr.1: 28710825-28710825 on Build GRCh38
rs369717346 Chr.1: 28710668-28710668 on Build GRCh38
rs757382803 Chr.1: 28710486-28710486 on Build GRCh38
rs760436748 Chr.1: 28710468-28710468 on Build GRCh38
rs768552083 Chr.1: 28710600-28710600 on Build GRCh38
rs369966949 Chr.1: 28710512-28710512 on Build GRCh38
rs79108316 Chr.1: 28710745-28710745 on Build GRCh38
rs980704998 Chr.1: 28710779-28710780 on Build GRCh38
rs776106665 Chr.1: 28710528-28710528 on Build GRCh38
rs770479035 Chr.1: 28710502-28710502 on Build GRCh38
rs764370987 Chr.1: 28710609-28710610 on Build GRCh38
rs865901012 Chr.1: 28710602-28710602 on Build GRCh38
rs750614860 Chr.1: 28710692-28710692 on Build GRCh38
rs771689691 Chr.1: 28710660-28710660 on Build GRCh38
rs949316014 Chr.1: 28710450-28710450 on Build GRCh38
rs926497771 Chr.1: 28710808-28710808 on Build GRCh38
rs776804192 Chr.1: 28710669-28710669 on Build GRCh38
rs116093303 Chr.1: 28710477-28710477 on Build GRCh38
rs200616220 Chr.1: 28710551-28710551 on Build GRCh38
rs745407705 Chr.1: 28710658-28710658 on Build GRCh38
rs768004583 Chr.1: 28710476-28710476 on Build GRCh38
rs11415888 Chr.1: 28710728-28710729 on Build GRCh38
rs1014318548 Chr.1: 28710714-28710714 on Build GRCh38
rs377551807 Chr.1: 28710898-28710898 on Build GRCh38
rs377667689 Chr.1: 28710661-28710661 on Build GRCh38
rs540193587 Chr.1: 28710831-28710831 on Build GRCh38
rs148296057 Chr.1: 28710596-28710596 on Build GRCh38
rs748004808 Chr.1: 28710498-28710498 on Build GRCh38
rs765373586 Chr.1: 28710581-28710581 on Build GRCh38
rs781094336 Chr.1: 28710497-28710497 on Build GRCh38
rs913583672 Chr.1: 28710833-28710834 on Build GRCh38
rs772769682 Chr.1: 28710521-28710521 on Build GRCh38
rs185336924 Chr.1: 28710783-28710783 on Build GRCh38
rs761172616 Chr.1: 28710539-28710539 on Build GRCh38
rs761051239 Chr.1: 28710666-28710666 on Build GRCh38
rs569104061 Chr.1: 28710645-28710645 on Build GRCh38
rs752580412 Chr.1: 28710613-28710613 on Build GRCh38
rs201698667 Chr.1: 28710636-28710636 on Build GRCh38
rs764318801 Chr.1: 28710544-28710544 on Build GRCh38
rs548492759 Chr.1: 28710480-28710480 on Build GRCh38
rs143549501 Chr.1: 28710817-28710817 on Build GRCh38
rs1043028937 Chr.1: 28710655-28710655 on Build GRCh38
rs761833123 Chr.1: 28710779-28710779 on Build GRCh38
rs3214953 Chr.1: 28710770-28710770 on Build GRCh38
rs145849888 Chr.1: 28710894-28710894 on Build GRCh38
rs201363668 Chr.1: 28710496-28710496 on Build GRCh38
rs397754554 Chr.1: 28710740-28710741 on Build GRCh38
rs1024788480 Chr.1: 28710724-28710724 on Build GRCh38
rs963270867 Chr.1: 28710909-28710909 on Build GRCh38
rs1054907972 Chr.1: 28710680-28710680 on Build GRCh38
rs372974630 Chr.1: 28710678-28710678 on Build GRCh38
rs945090110 Chr.1: 28710870-28710870 on Build GRCh38
rs761875078 Chr.1: 28710560-28710560 on Build GRCh38
rs562652729 Chr.1: 28710442-28710442 on Build GRCh38
rs199745759 Chr.1: 28710495-28710495 on Build GRCh38
rs1055273925 Chr.1: 28710445-28710445 on Build GRCh38
rs199899562 Chr.1: 28710741-28710741 on Build GRCh38
rs778701267 Chr.1: 28710649-28710649 on Build GRCh38
rs894840616 Chr.1: 28710457-28710457 on Build GRCh38
rs147476962 Chr.1: 28710631-28710631 on Build GRCh38
rs772940436 Chr.1: 28710500-28710500 on Build GRCh38
rs778822563 Chr.1: 28710488-28710488 on Build GRCh38
rs143592261 Chr.1: 28710632-28710632 on Build GRCh38
rs894876596 Chr.1: 28710700-28710700 on Build GRCh38
rs776581232 Chr.1: 28710763-28710763 on Build GRCh38

View Full Product Details