Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28996828-28997354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
UniGene
Hs.175437
Gene Chromosome Location
Chr.1: 28887091-29120046 on Build GRCh38
Transcripts
NM_001166005.1 NM_001166006.1 NM_001166007.1 NM_004437.3 NM_203342.2 NM_203343.2 XM_005245753.1 XM_005245757.1 XM_005245760.1 XM_005245761.1 XM_005245763.1 XM_005245764.1 XM_005245765.1 XM_005245768.1 XM_005245769.1 XM_005245770.1 XM_005245772.4 XM_005245773.4 XM_005245774.1 XM_006710434.1 XM_006710439.1 XM_011540956.1 XM_011540957.1 XM_011540958.1 XM_011540959.1 XM_011540960.1 XM_011540961.1 XM_011540962.1 XM_011540963.2 XM_011540964.1 XM_011540965.2 XM_017000581.1 XM_017000582.1 XM_017000583.1 XM_017000584.1 XM_017000585.1 XM_017000586.1 XM_017000587.1 XM_017000588.1 XM_017000589.1 XM_017000590.1 XM_017000591.1 XM_017000592.1 XM_017000593.1 XM_017000594.1 XM_017000595.1 XM_017000596.1 XM_017000597.1 XM_017000598.1 XM_017000599.1 XM_017000600.1 XM_017000601.1 XM_017000602.1 XM_017000603.1 XM_017000604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386075 c.142G>C p.D48H Chr.1: 28997302-28997302 on Build GRCh38
6336285 c.159+8G>T p.? Chr.1: 28997327-28997327 on Build GRCh38
3719606 c.159+1G>A p.? Chr.1: 28997320-28997320 on Build GRCh38
7309726 c.684A>T p.K228N Chr.1: 28997217-28997217 on Build GRCh38
1341621 c.738G>A p.L246L Chr.1: 28997271-28997271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932368342 Chr.1: 28997039-28997039 on Build GRCh38
rs920931279 Chr.1: 28997028-28997028 on Build GRCh38
rs747566443 Chr.1: 28997294-28997294 on Build GRCh38
rs566540981 Chr.1: 28997308-28997308 on Build GRCh38
rs767443466 Chr.1: 28997205-28997205 on Build GRCh38
rs779683074 Chr.1: 28997169-28997171 on Build GRCh38
rs929933368 Chr.1: 28997321-28997321 on Build GRCh38
rs367951854 Chr.1: 28996889-28996889 on Build GRCh38
rs1024117747 Chr.1: 28996962-28996962 on Build GRCh38
rs386629877 Chr.1: 28996904-28996906 on Build GRCh38
rs1050813968 Chr.1: 28997042-28997042 on Build GRCh38
rs780730982 Chr.1: 28997288-28997288 on Build GRCh38
rs914889954 Chr.1: 28996945-28996945 on Build GRCh38
rs751877537 Chr.1: 28997312-28997312 on Build GRCh38
rs373464494 Chr.1: 28997264-28997264 on Build GRCh38
rs781668873 Chr.1: 28997307-28997307 on Build GRCh38
rs982657318 Chr.1: 28996987-28996987 on Build GRCh38
rs548455058 Chr.1: 28997190-28997190 on Build GRCh38
rs368673412 Chr.1: 28996923-28996923 on Build GRCh38
rs777112119 Chr.1: 28997193-28997193 on Build GRCh38
rs941320405 Chr.1: 28997010-28997010 on Build GRCh38
rs930007450 Chr.1: 28997037-28997037 on Build GRCh38
rs769091318 Chr.1: 28997188-28997188 on Build GRCh38
rs771862783 Chr.1: 28997168-28997171 on Build GRCh38
rs898487062 Chr.1: 28997033-28997033 on Build GRCh38
rs749954369 Chr.1: 28997274-28997274 on Build GRCh38
rs531441869 Chr.1: 28997123-28997123 on Build GRCh38
rs990569895 Chr.1: 28996943-28996943 on Build GRCh38
rs369527719 Chr.1: 28997250-28997250 on Build GRCh38
rs973969764 Chr.1: 28997326-28997326 on Build GRCh38
rs760586185 Chr.1: 28997235-28997235 on Build GRCh38
rs1033980330 Chr.1: 28996900-28996917 on Build GRCh38
rs1034698412 Chr.1: 28996924-28996924 on Build GRCh38
rs1024379477 Chr.1: 28997246-28997246 on Build GRCh38
rs770064316 Chr.1: 28997311-28997311 on Build GRCh38
rs769853791 Chr.1: 28996860-28996860 on Build GRCh38
rs552908580 Chr.1: 28997296-28997296 on Build GRCh38
rs773706226 Chr.1: 28997318-28997318 on Build GRCh38
rs149693862 Chr.1: 28996993-28996993 on Build GRCh38
rs746871828 Chr.1: 28997182-28997186 on Build GRCh38
rs752588508 Chr.1: 28997219-28997219 on Build GRCh38
rs147787352 Chr.1: 28996994-28996994 on Build GRCh38
rs971657764 Chr.1: 28996982-28996982 on Build GRCh38
rs756652002 Chr.1: 28997229-28997229 on Build GRCh38
rs958741377 Chr.1: 28996942-28996942 on Build GRCh38
rs866959406 Chr.1: 28997016-28997016 on Build GRCh38
rs529713075 Chr.1: 28996906-28996906 on Build GRCh38
rs895005592 Chr.1: 28997131-28997131 on Build GRCh38
rs146976856 Chr.1: 28997322-28997322 on Build GRCh38
rs756996105 Chr.1: 28997253-28997253 on Build GRCh38
rs141351853 Chr.1: 28997201-28997201 on Build GRCh38
rs898535942 Chr.1: 28996856-28996856 on Build GRCh38
rs1011936653 Chr.1: 28997160-28997160 on Build GRCh38
rs763919964 Chr.1: 28997248-28997248 on Build GRCh38
rs762880453 Chr.1: 28997203-28997203 on Build GRCh38
rs924232608 Chr.1: 28996980-28996980 on Build GRCh38
rs773187250 Chr.1: 28996961-28996961 on Build GRCh38
rs978718370 Chr.1: 28996969-28996969 on Build GRCh38
rs552009502 Chr.1: 28997077-28997077 on Build GRCh38
rs1022011759 Chr.1: 28997166-28997166 on Build GRCh38
rs769903319 Chr.1: 28997200-28997200 on Build GRCh38
rs747411155 Chr.1: 28997187-28997187 on Build GRCh38
rs749414197 Chr.1: 28997315-28997315 on Build GRCh38
rs745518328 Chr.1: 28997167-28997171 on Build GRCh38
rs12354050 Chr.1: 28997083-28997083 on Build GRCh38
rs757942511 Chr.1: 28997284-28997284 on Build GRCh38
rs369745237 Chr.1: 28997056-28997056 on Build GRCh38
rs377669103 Chr.1: 28997196-28997196 on Build GRCh38

View Full Product Details