Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31652155-31652654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
UniGene
Hs.368921
Gene Chromosome Location
Chr.1: 31652247-31704167 on Build GRCh38
Transcripts
NM_001856.3 XM_005270481.1 XM_011540722.1 XM_011540723.1 XM_011540724.1 XM_011540726.2 XM_011540727.2 XM_011540728.2 XM_017000338.1 XM_017000339.1 XM_017000340.1 XM_017000341.1

SNP Detail

SNP ID Chromosome Location
rs1046788476 Chr.1: 31652276-31652276 on Build GRCh38
rs531601728 Chr.1: 31652301-31652301 on Build GRCh38
rs767213558 Chr.1: 31652342-31652342 on Build GRCh38
rs986922737 Chr.1: 31652431-31652431 on Build GRCh38
rs929039739 Chr.1: 31652408-31652408 on Build GRCh38
rs911207484 Chr.1: 31652438-31652438 on Build GRCh38
rs964320390 Chr.1: 31652360-31652360 on Build GRCh38
rs1029779262 Chr.1: 31652545-31652547 on Build GRCh38
rs942709668 Chr.1: 31652450-31652450 on Build GRCh38
rs906735236 Chr.1: 31652208-31652208 on Build GRCh38
rs548138581 Chr.1: 31652303-31652303 on Build GRCh38
rs760849938 Chr.1: 31652617-31652617 on Build GRCh38
rs938428642 Chr.1: 31652576-31652576 on Build GRCh38
rs79928805 Chr.1: 31652464-31652464 on Build GRCh38
rs141442362 Chr.1: 31652223-31652224 on Build GRCh38
rs181842391 Chr.1: 31652206-31652206 on Build GRCh38
rs368642761 Chr.1: 31652556-31652556 on Build GRCh38
rs950436227 Chr.1: 31652568-31652568 on Build GRCh38
rs973326368 Chr.1: 31652561-31652562 on Build GRCh38
rs920308980 Chr.1: 31652398-31652398 on Build GRCh38
rs977675287 Chr.1: 31652397-31652397 on Build GRCh38
rs377302775 Chr.1: 31652559-31652559 on Build GRCh38
rs895402625 Chr.1: 31652500-31652500 on Build GRCh38
rs895134243 Chr.1: 31652250-31652250 on Build GRCh38
rs748047647 Chr.1: 31652430-31652430 on Build GRCh38
rs966794043 Chr.1: 31652277-31652277 on Build GRCh38
rs2139539 Chr.1: 31652178-31652178 on Build GRCh38
rs867074314 Chr.1: 31652221-31652222 on Build GRCh38
rs1048367404 Chr.1: 31652194-31652194 on Build GRCh38
rs1010930543 Chr.1: 31652274-31652274 on Build GRCh38
rs898652548 Chr.1: 31652179-31652179 on Build GRCh38
rs1047681941 Chr.1: 31652529-31652529 on Build GRCh38
rs964605624 Chr.1: 31652560-31652560 on Build GRCh38
rs967207466 Chr.1: 31652384-31652384 on Build GRCh38
rs1043739369 Chr.1: 31652508-31652511 on Build GRCh38
rs906948151 Chr.1: 31652287-31652287 on Build GRCh38
rs902557429 Chr.1: 31652510-31652510 on Build GRCh38
rs186187190 Chr.1: 31652225-31652225 on Build GRCh38
rs1021989469 Chr.1: 31652370-31652370 on Build GRCh38
rs930118987 Chr.1: 31652507-31652507 on Build GRCh38
rs1017433733 Chr.1: 31652559-31652560 on Build GRCh38
rs1055761334 Chr.1: 31652476-31652476 on Build GRCh38
rs538892841 Chr.1: 31652323-31652323 on Build GRCh38
rs764202561 Chr.1: 31652618-31652618 on Build GRCh38
rs1031052493 Chr.1: 31652412-31652412 on Build GRCh38
rs1061392 Chr.1: 31652559-31652559 on Build GRCh38
rs928103042 Chr.1: 31652441-31652441 on Build GRCh38
rs987783384 Chr.1: 31652341-31652341 on Build GRCh38
rs907412517 Chr.1: 31652562-31652562 on Build GRCh38
rs776555093 Chr.1: 31652619-31652619 on Build GRCh38
rs369128729 Chr.1: 31652556-31652557 on Build GRCh38
rs553961024 Chr.1: 31652604-31652604 on Build GRCh38
rs974325108 Chr.1: 31652454-31652454 on Build GRCh38
rs759217107 Chr.1: 31652310-31652310 on Build GRCh38

View Full Product Details