Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28884138-28884645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT2
Entrez Gene ID
2319
Gene Name
flotillin 2
Gene Aliases
ECS-1, ECS1, ESA, ESA1, M17S1
UniGene
Hs.514038
Gene Chromosome Location
Chr.17: 28879339-28897728 on Build GRCh38
Transcripts
NM_004475.2 XM_005257950.3 XM_005257953.2 XM_017024394.1 XM_017024395.1 XM_017024396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685016 c.183G>A p.T61T Chr.17: 28884264-28884264 on Build GRCh38
7203705 c.177A>G p.V59V Chr.17: 28884270-28884270 on Build GRCh38
4065083 c.217G>A p.A73T Chr.17: 28884230-28884230 on Build GRCh38
1479378 c.183G>T p.T61T Chr.17: 28884264-28884264 on Build GRCh38
3819164 c.151A>G p.T51A Chr.17: 28884296-28884296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371665436 Chr.17: 28884261-28884261 on Build GRCh38
rs777995203 Chr.17: 28884345-28884345 on Build GRCh38
rs994451112 Chr.17: 28884376-28884376 on Build GRCh38
rs185758935 Chr.17: 28884336-28884336 on Build GRCh38
rs776917423 Chr.17: 28884194-28884194 on Build GRCh38
rs978138176 Chr.17: 28884327-28884327 on Build GRCh38
rs368311864 Chr.17: 28884176-28884176 on Build GRCh38
rs1018320729 Chr.17: 28884425-28884425 on Build GRCh38
rs748747160 Chr.17: 28884216-28884217 on Build GRCh38
rs753839776 Chr.17: 28884295-28884295 on Build GRCh38
rs375260312 Chr.17: 28884175-28884175 on Build GRCh38
rs368990152 Chr.17: 28884555-28884555 on Build GRCh38
rs373339466 Chr.17: 28884352-28884352 on Build GRCh38
rs945331834 Chr.17: 28884198-28884198 on Build GRCh38
rs552792143 Chr.17: 28884401-28884401 on Build GRCh38
rs528877539 Chr.17: 28884395-28884395 on Build GRCh38
rs763692285 Chr.17: 28884199-28884199 on Build GRCh38
rs1051293689 Chr.17: 28884182-28884182 on Build GRCh38
rs376560502 Chr.17: 28884354-28884354 on Build GRCh38
rs990925422 Chr.17: 28884610-28884610 on Build GRCh38
rs778774380 Chr.17: 28884303-28884303 on Build GRCh38
rs374933132 Chr.17: 28884202-28884202 on Build GRCh38
rs941590256 Chr.17: 28884400-28884400 on Build GRCh38
rs941143306 Chr.17: 28884580-28884580 on Build GRCh38
rs547509702 Chr.17: 28884480-28884480 on Build GRCh38
rs772212983 Chr.17: 28884266-28884266 on Build GRCh38
rs766280109 Chr.17: 28884294-28884294 on Build GRCh38
rs561751079 Chr.17: 28884366-28884366 on Build GRCh38
rs751056928 Chr.17: 28884200-28884200 on Build GRCh38
rs1020221503 Chr.17: 28884502-28884502 on Build GRCh38
rs572169033 Chr.17: 28884291-28884291 on Build GRCh38
rs1011461365 Chr.17: 28884495-28884495 on Build GRCh38
rs759866053 Chr.17: 28884279-28884279 on Build GRCh38
rs945274746 Chr.17: 28884474-28884474 on Build GRCh38
rs750006144 Chr.17: 28884285-28884285 on Build GRCh38
rs545992041 Chr.17: 28884311-28884311 on Build GRCh38
rs943183264 Chr.17: 28884168-28884168 on Build GRCh38
rs894893421 Chr.17: 28884520-28884520 on Build GRCh38
rs773832079 Chr.17: 28884281-28884281 on Build GRCh38
rs1050862076 Chr.17: 28884453-28884453 on Build GRCh38
rs999957478 Chr.17: 28884541-28884541 on Build GRCh38
rs373182015 Chr.17: 28884390-28884390 on Build GRCh38
rs770714663 Chr.17: 28884253-28884254 on Build GRCh38
rs1035106396 Chr.17: 28884601-28884601 on Build GRCh38
rs879126750 Chr.17: 28884172-28884172 on Build GRCh38
rs34236370 Chr.17: 28884511-28884512 on Build GRCh38
rs747505161 Chr.17: 28884273-28884273 on Build GRCh38
rs200366548 Chr.17: 28884230-28884230 on Build GRCh38
rs941013932 Chr.17: 28884162-28884162 on Build GRCh38
rs929676380 Chr.17: 28884422-28884422 on Build GRCh38
rs181122272 Chr.17: 28884193-28884193 on Build GRCh38
rs774090581 Chr.17: 28884261-28884261 on Build GRCh38
rs957724355 Chr.17: 28884426-28884426 on Build GRCh38
rs777022380 Chr.17: 28884278-28884278 on Build GRCh38
rs749079749 Chr.17: 28884238-28884238 on Build GRCh38
rs902501568 Chr.17: 28884538-28884538 on Build GRCh38
rs779898935 Chr.17: 28884237-28884237 on Build GRCh38
rs1039913554 Chr.17: 28884483-28884483 on Build GRCh38
rs374807903 Chr.17: 28884284-28884284 on Build GRCh38
rs761143929 Chr.17: 28884266-28884266 on Build GRCh38
rs1038031825 Chr.17: 28884164-28884164 on Build GRCh38
rs778955819 Chr.17: 28884258-28884258 on Build GRCh38
rs772839708 Chr.17: 28884193-28884194 on Build GRCh38
rs991421596 Chr.17: 28884370-28884370 on Build GRCh38
rs146169915 Chr.17: 28884355-28884355 on Build GRCh38
rs139785721 Chr.17: 28884602-28884602 on Build GRCh38
rs761456405 Chr.17: 28884283-28884283 on Build GRCh38
rs747338211 Chr.17: 28884417-28884417 on Build GRCh38
rs574722327 Chr.17: 28884163-28884163 on Build GRCh38
rs756053914 Chr.17: 28884231-28884231 on Build GRCh38
rs752000785 Chr.17: 28884479-28884479 on Build GRCh38
rs370996940 Chr.17: 28884173-28884173 on Build GRCh38
rs769600356 Chr.17: 28884194-28884194 on Build GRCh38
rs141542425 Chr.17: 28884349-28884349 on Build GRCh38
rs766551293 Chr.17: 28884201-28884201 on Build GRCh38
rs890892523 Chr.17: 28884473-28884473 on Build GRCh38
rs758751455 Chr.17: 28884339-28884339 on Build GRCh38
rs755208855 Chr.17: 28884242-28884242 on Build GRCh38
rs1054032175 Chr.17: 28884169-28884169 on Build GRCh38
rs918216174 Chr.17: 28884407-28884407 on Build GRCh38
rs989318803 Chr.17: 28884437-28884437 on Build GRCh38
rs897404515 Chr.17: 28884371-28884371 on Build GRCh38
rs778012620 Chr.17: 28884270-28884270 on Build GRCh38
rs372643690 Chr.17: 28884570-28884570 on Build GRCh38
rs770213730 Chr.17: 28884280-28884280 on Build GRCh38
rs922630601 Chr.17: 28884161-28884161 on Build GRCh38
rs892992673 Chr.17: 28884489-28884489 on Build GRCh38
rs767042568 Chr.17: 28884215-28884215 on Build GRCh38
rs190517951 Chr.17: 28884376-28884376 on Build GRCh38

View Full Product Details