Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28902661-28903190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS13
Entrez Gene ID
147015
Gene Name
dehydrogenase/reductase 13
Gene Aliases
SDR7C5
UniGene
Hs.631760
Gene Chromosome Location
Chr.17: 28897781-28903071 on Build GRCh38
Transcripts
NM_144683.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983340 c.49C>G p.L17V Chr.17: 28902896-28902896 on Build GRCh38
3771305 c.150G>C p.K50N Chr.17: 28902679-28902679 on Build GRCh38
1203518 c.7G>T p.A3S Chr.17: 28902938-28902938 on Build GRCh38
4993879 c.123C>A p.V41V Chr.17: 28902822-28902822 on Build GRCh38
5473864 c.1A>G p.M1V Chr.17: 28902944-28902944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943141781 Chr.17: 28902799-28902799 on Build GRCh38
rs368681405 Chr.17: 28902900-28902900 on Build GRCh38
rs752618993 Chr.17: 28902809-28902809 on Build GRCh38
rs750271207 Chr.17: 28902968-28902968 on Build GRCh38
rs780248630 Chr.17: 28902904-28902904 on Build GRCh38
rs546969625 Chr.17: 28903059-28903059 on Build GRCh38
rs748703296 Chr.17: 28902914-28902914 on Build GRCh38
rs757096571 Chr.17: 28902827-28902827 on Build GRCh38
rs760384662 Chr.17: 28902748-28902748 on Build GRCh38
rs577948182 Chr.17: 28902719-28902719 on Build GRCh38
rs1037563048 Chr.17: 28903057-28903057 on Build GRCh38
rs761868307 Chr.17: 28902963-28902963 on Build GRCh38
rs772490190 Chr.17: 28902864-28902864 on Build GRCh38
rs1037856551 Chr.17: 28902920-28902920 on Build GRCh38
rs761032358 Chr.17: 28903063-28903063 on Build GRCh38
rs764817263 Chr.17: 28902802-28902802 on Build GRCh38
rs767354285 Chr.17: 28902899-28902899 on Build GRCh38
rs866244018 Chr.17: 28902886-28902886 on Build GRCh38
rs954413967 Chr.17: 28902724-28902724 on Build GRCh38
rs1017745426 Chr.17: 28902853-28902853 on Build GRCh38
rs773539811 Chr.17: 28902879-28902880 on Build GRCh38
rs762407713 Chr.17: 28902943-28902943 on Build GRCh38
rs781230594 Chr.17: 28902844-28902844 on Build GRCh38
rs376838348 Chr.17: 28902815-28902815 on Build GRCh38
rs752247226 Chr.17: 28902880-28902880 on Build GRCh38
rs945309533 Chr.17: 28902818-28902818 on Build GRCh38
rs755141192 Chr.17: 28902907-28902907 on Build GRCh38
rs945409842 Chr.17: 28902696-28902696 on Build GRCh38
rs112491750 Chr.17: 28902713-28902713 on Build GRCh38
rs906642016 Chr.17: 28902722-28902722 on Build GRCh38
rs768966288 Chr.17: 28903089-28903089 on Build GRCh38
rs770346102 Chr.17: 28902793-28902808 on Build GRCh38
rs899104961 Chr.17: 28903061-28903061 on Build GRCh38
rs763595162 Chr.17: 28902947-28902947 on Build GRCh38
rs993439638 Chr.17: 28903071-28903071 on Build GRCh38
rs553376241 Chr.17: 28902700-28902700 on Build GRCh38
rs763967257 Chr.17: 28902882-28902882 on Build GRCh38
rs779497199 Chr.17: 28902909-28902909 on Build GRCh38
rs1008495705 Chr.17: 28903021-28903021 on Build GRCh38
rs879137526 Chr.17: 28903030-28903030 on Build GRCh38
rs1013036175 Chr.17: 28902919-28902919 on Build GRCh38
rs954961790 Chr.17: 28903019-28903019 on Build GRCh38
rs987235895 Chr.17: 28902819-28902819 on Build GRCh38
rs987105594 Chr.17: 28902757-28902757 on Build GRCh38
rs989673974 Chr.17: 28903073-28903073 on Build GRCh38
rs915210604 Chr.17: 28902690-28902690 on Build GRCh38
rs371122246 Chr.17: 28902778-28902778 on Build GRCh38
rs563774361 Chr.17: 28902781-28902781 on Build GRCh38
rs577270353 Chr.17: 28902854-28902854 on Build GRCh38
rs749804861 Chr.17: 28902856-28902856 on Build GRCh38
rs772905614 Chr.17: 28903124-28903124 on Build GRCh38
rs1055144571 Chr.17: 28902979-28902979 on Build GRCh38
rs977952134 Chr.17: 28903142-28903142 on Build GRCh38
rs939399171 Chr.17: 28902972-28902972 on Build GRCh38
rs964410890 Chr.17: 28903065-28903065 on Build GRCh38
rs898827580 Chr.17: 28902765-28902765 on Build GRCh38
rs1026948495 Chr.17: 28902977-28902977 on Build GRCh38
rs1017398246 Chr.17: 28903098-28903098 on Build GRCh38
rs775023116 Chr.17: 28902858-28902858 on Build GRCh38
rs993758919 Chr.17: 28902973-28902973 on Build GRCh38
rs762229292 Chr.17: 28902862-28902862 on Build GRCh38
rs762856868 Chr.17: 28902881-28902881 on Build GRCh38
rs756931614 Chr.17: 28902889-28902889 on Build GRCh38
rs749334823 Chr.17: 28902905-28902905 on Build GRCh38
rs758908418 Chr.17: 28902801-28902801 on Build GRCh38
rs894532634 Chr.17: 28902913-28902913 on Build GRCh38
rs915082159 Chr.17: 28902813-28902813 on Build GRCh38
rs915071925 Chr.17: 28903084-28903084 on Build GRCh38
rs978471594 Chr.17: 28902953-28902953 on Build GRCh38
rs766482491 Chr.17: 28902969-28902985 on Build GRCh38
rs772314440 Chr.17: 28902918-28902918 on Build GRCh38
rs894305670 Chr.17: 28903036-28903036 on Build GRCh38
rs773697372 Chr.17: 28902921-28902921 on Build GRCh38
rs964856902 Chr.17: 28903105-28903105 on Build GRCh38
rs374516726 Chr.17: 28902849-28902849 on Build GRCh38
rs769700518 Chr.17: 28902852-28902852 on Build GRCh38
rs899371306 Chr.17: 28902970-28902970 on Build GRCh38
rs775144015 Chr.17: 28902935-28902935 on Build GRCh38
rs751402262 Chr.17: 28902817-28902817 on Build GRCh38
rs756265900 Chr.17: 28902983-28902983 on Build GRCh38
rs764651976 Chr.17: 28902869-28902869 on Build GRCh38
rs767211784 Chr.17: 28902966-28902966 on Build GRCh38
rs776155895 Chr.17: 28903044-28903044 on Build GRCh38
rs758398199 Chr.17: 28902810-28902810 on Build GRCh38
rs766430746 Chr.17: 28902879-28902879 on Build GRCh38
rs201933739 Chr.17: 28902885-28902885 on Build GRCh38
rs528828150 Chr.17: 28902960-28902960 on Build GRCh38
rs759295428 Chr.17: 28902865-28902865 on Build GRCh38
rs1012753271 Chr.17: 28902752-28902752 on Build GRCh38
rs766670294 Chr.17: 28902985-28902985 on Build GRCh38
rs1056870286 Chr.17: 28902747-28902747 on Build GRCh38
rs879928661 Chr.17: 28903165-28903165 on Build GRCh38
rs1016167997 Chr.17: 28902755-28902755 on Build GRCh38
rs561447167 Chr.17: 28902924-28902924 on Build GRCh38
rs763049103 Chr.17: 28902904-28902921 on Build GRCh38
rs1025979145 Chr.17: 28902717-28902717 on Build GRCh38
rs945386289 Chr.17: 28902911-28902911 on Build GRCh38
rs571872239 Chr.17: 28903079-28903079 on Build GRCh38
rs769324384 Chr.17: 28902857-28902857 on Build GRCh38
rs993593482 Chr.17: 28902776-28902776 on Build GRCh38

View Full Product Details