Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29616190-29616692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO6
Entrez Gene ID
84940
Gene Name
coronin 6
Gene Aliases
-
UniGene
Hs.143046
Gene Chromosome Location
Chr.17: 29614756-29622912 on Build GRCh38
Transcripts
NM_032854.3 XM_005258047.3 XM_005258048.4 XM_005258056.4 XM_011525385.2 XM_011525386.2 XM_011525387.2 XM_011525388.2 XM_011525390.2 XM_011525391.2 XM_017025235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5529108 c.1021G>A p.E341K Chr.17: 29616320-29616320 on Build GRCh38
6249609 c.1015C>A p.L339I Chr.17: 29616326-29616326 on Build GRCh38
460386 c.1025G>C p.R342T Chr.17: 29616316-29616316 on Build GRCh38
6634257 c.1020C>T p.H340H Chr.17: 29616321-29616321 on Build GRCh38
4974068 c.1057C>T p.R353C Chr.17: 29616284-29616284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745872126 Chr.17: 29616380-29616381 on Build GRCh38
rs898920597 Chr.17: 29616568-29616568 on Build GRCh38
rs551582326 Chr.17: 29616369-29616369 on Build GRCh38
rs771888486 Chr.17: 29616213-29616213 on Build GRCh38
rs1031357840 Chr.17: 29616667-29616667 on Build GRCh38
rs1043387339 Chr.17: 29616646-29616646 on Build GRCh38
rs899701442 Chr.17: 29616640-29616640 on Build GRCh38
rs761673821 Chr.17: 29616212-29616212 on Build GRCh38
rs554061613 Chr.17: 29616595-29616595 on Build GRCh38
rs562301918 Chr.17: 29616670-29616670 on Build GRCh38
rs776247867 Chr.17: 29616671-29616671 on Build GRCh38
rs770617846 Chr.17: 29616317-29616317 on Build GRCh38
rs757527797 Chr.17: 29616271-29616271 on Build GRCh38
rs776924661 Chr.17: 29616319-29616319 on Build GRCh38
rs372891094 Chr.17: 29616385-29616385 on Build GRCh38
rs904173633 Chr.17: 29616651-29616651 on Build GRCh38
rs772993131 Chr.17: 29616311-29616311 on Build GRCh38
rs929080315 Chr.17: 29616587-29616587 on Build GRCh38
rs146808043 Chr.17: 29616409-29616409 on Build GRCh38
rs948385885 Chr.17: 29616509-29616509 on Build GRCh38
rs3029571 Chr.17: 29616391-29616394 on Build GRCh38
rs191308299 Chr.17: 29616286-29616286 on Build GRCh38
rs113833180 Chr.17: 29616511-29616511 on Build GRCh38
rs753259381 Chr.17: 29616241-29616241 on Build GRCh38
rs977610738 Chr.17: 29616413-29616413 on Build GRCh38
rs796412073 Chr.17: 29616302-29616302 on Build GRCh38
rs201994744 Chr.17: 29616266-29616266 on Build GRCh38
rs111572312 Chr.17: 29616338-29616338 on Build GRCh38
rs779117454 Chr.17: 29616295-29616297 on Build GRCh38
rs765349848 Chr.17: 29616240-29616240 on Build GRCh38
rs758712172 Chr.17: 29616388-29616390 on Build GRCh38
rs890316454 Chr.17: 29616596-29616596 on Build GRCh38
rs915550768 Chr.17: 29616496-29616496 on Build GRCh38
rs753561854 Chr.17: 29616375-29616375 on Build GRCh38
rs981466066 Chr.17: 29616407-29616407 on Build GRCh38
rs770854544 Chr.17: 29616304-29616304 on Build GRCh38
rs780377164 Chr.17: 29616653-29616654 on Build GRCh38
rs932583250 Chr.17: 29616645-29616645 on Build GRCh38
rs1017495483 Chr.17: 29616624-29616624 on Build GRCh38
rs370489778 Chr.17: 29616214-29616214 on Build GRCh38
rs1040906984 Chr.17: 29616632-29616632 on Build GRCh38
rs1031410044 Chr.17: 29616310-29616310 on Build GRCh38
rs878960480 Chr.17: 29616602-29616602 on Build GRCh38
rs572442949 Chr.17: 29616631-29616631 on Build GRCh38
rs760367073 Chr.17: 29616215-29616215 on Build GRCh38
rs778358180 Chr.17: 29616290-29616290 on Build GRCh38
rs547733920 Chr.17: 29616238-29616238 on Build GRCh38
rs1037413797 Chr.17: 29616558-29616558 on Build GRCh38
rs757691636 Chr.17: 29616288-29616289 on Build GRCh38
rs752002811 Chr.17: 29616357-29616357 on Build GRCh38
rs746890219 Chr.17: 29616313-29616313 on Build GRCh38
rs767712452 Chr.17: 29616373-29616373 on Build GRCh38
rs772048161 Chr.17: 29616305-29616305 on Build GRCh38
rs529028062 Chr.17: 29616600-29616600 on Build GRCh38
rs779817411 Chr.17: 29616277-29616277 on Build GRCh38
rs878887456 Chr.17: 29616441-29616441 on Build GRCh38
rs919936323 Chr.17: 29616416-29616416 on Build GRCh38
rs778302525 Chr.17: 29616263-29616263 on Build GRCh38
rs759718653 Chr.17: 29616239-29616239 on Build GRCh38
rs748107280 Chr.17: 29616292-29616292 on Build GRCh38
rs924814694 Chr.17: 29616418-29616418 on Build GRCh38
rs556343915 Chr.17: 29616426-29616426 on Build GRCh38
rs1025143536 Chr.17: 29616393-29616393 on Build GRCh38
rs748839216 Chr.17: 29616279-29616279 on Build GRCh38
rs368623066 Chr.17: 29616378-29616378 on Build GRCh38
rs535431920 Chr.17: 29616488-29616488 on Build GRCh38
rs201030691 Chr.17: 29616655-29616655 on Build GRCh38
rs746834698 Chr.17: 29616668-29616668 on Build GRCh38
rs763101202 Chr.17: 29616339-29616339 on Build GRCh38
rs1012657713 Chr.17: 29616230-29616230 on Build GRCh38
rs569747580 Chr.17: 29616396-29616396 on Build GRCh38
rs775659192 Chr.17: 29616323-29616323 on Build GRCh38
rs533247552 Chr.17: 29616353-29616353 on Build GRCh38
rs1028702440 Chr.17: 29616334-29616355 on Build GRCh38
rs754579051 Chr.17: 29616376-29616376 on Build GRCh38
rs765437209 Chr.17: 29616321-29616321 on Build GRCh38
rs908642192 Chr.17: 29616588-29616588 on Build GRCh38
rs759763909 Chr.17: 29616320-29616320 on Build GRCh38
rs943812200 Chr.17: 29616615-29616615 on Build GRCh38
rs374753616 Chr.17: 29616284-29616284 on Build GRCh38
rs758932234 Chr.17: 29616242-29616242 on Build GRCh38
rs568118705 Chr.17: 29616448-29616448 on Build GRCh38

View Full Product Details