Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30834363-30834864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD5
Entrez Gene ID
79915
Gene Name
ATPase family, AAA domain containing 5
Gene Aliases
C17orf41, ELG1, FRAG1
UniGene
Hs.528902
Gene Chromosome Location
Chr.17: 30830932-30896424 on Build GRCh38
Transcripts
NM_024857.4 XM_011525269.2 XM_011525270.2 XM_011525273.2 XM_017025111.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6571448 c.427G>C p.E143Q Chr.17: 30834508-30834508 on Build GRCh38
5981184 c.777T>A p.N259K Chr.17: 30834858-30834858 on Build GRCh38
4663946 c.479G>A p.R160H Chr.17: 30834560-30834560 on Build GRCh38
1749942 c.698G>T p.G233V Chr.17: 30834779-30834779 on Build GRCh38
6859445 c.352A>G p.N118D Chr.17: 30834433-30834433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760661424 Chr.17: 30834677-30834677 on Build GRCh38
rs367699401 Chr.17: 30834724-30834724 on Build GRCh38
rs373741401 Chr.17: 30834494-30834494 on Build GRCh38
rs774332249 Chr.17: 30834765-30834765 on Build GRCh38
rs372570681 Chr.17: 30834720-30834720 on Build GRCh38
rs756535617 Chr.17: 30834441-30834441 on Build GRCh38
rs138260353 Chr.17: 30834781-30834781 on Build GRCh38
rs769598020 Chr.17: 30834496-30834496 on Build GRCh38
rs745340855 Chr.17: 30834620-30834620 on Build GRCh38
rs781362252 Chr.17: 30834430-30834430 on Build GRCh38
rs1042030653 Chr.17: 30834694-30834694 on Build GRCh38
rs780484748 Chr.17: 30834449-30834449 on Build GRCh38
rs768662290 Chr.17: 30834761-30834761 on Build GRCh38
rs762145513 Chr.17: 30834663-30834663 on Build GRCh38
rs1047706402 Chr.17: 30834606-30834606 on Build GRCh38
rs755590738 Chr.17: 30834743-30834743 on Build GRCh38
rs564125064 Chr.17: 30834559-30834559 on Build GRCh38
rs1018482018 Chr.17: 30834652-30834652 on Build GRCh38
rs548306992 Chr.17: 30834816-30834816 on Build GRCh38
rs766428559 Chr.17: 30834680-30834680 on Build GRCh38
rs769837150 Chr.17: 30834623-30834623 on Build GRCh38
rs763286624 Chr.17: 30834418-30834418 on Build GRCh38
rs914996435 Chr.17: 30834629-30834629 on Build GRCh38
rs759319232 Chr.17: 30834671-30834671 on Build GRCh38
rs202218286 Chr.17: 30834498-30834498 on Build GRCh38
rs773461644 Chr.17: 30834672-30834672 on Build GRCh38
rs767734234 Chr.17: 30834541-30834541 on Build GRCh38
rs149194938 Chr.17: 30834480-30834480 on Build GRCh38
rs762057400 Chr.17: 30834518-30834518 on Build GRCh38
rs750477216 Chr.17: 30834542-30834542 on Build GRCh38
rs150061485 Chr.17: 30834420-30834420 on Build GRCh38
rs746696655 Chr.17: 30834568-30834568 on Build GRCh38
rs374681958 Chr.17: 30834786-30834786 on Build GRCh38
rs749190277 Chr.17: 30834632-30834632 on Build GRCh38
rs375111527 Chr.17: 30834560-30834560 on Build GRCh38
rs774726632 Chr.17: 30834515-30834515 on Build GRCh38
rs755474549 Chr.17: 30834701-30834701 on Build GRCh38
rs780816463 Chr.17: 30834732-30834732 on Build GRCh38
rs780730225 Chr.17: 30834588-30834588 on Build GRCh38
rs577386724 Chr.17: 30834530-30834530 on Build GRCh38
rs760087393 Chr.17: 30834819-30834819 on Build GRCh38
rs899243141 Chr.17: 30834835-30834835 on Build GRCh38
rs181925781 Chr.17: 30834573-30834573 on Build GRCh38
rs954559133 Chr.17: 30834398-30834398 on Build GRCh38
rs779548775 Chr.17: 30834747-30834747 on Build GRCh38
rs775379351 Chr.17: 30834441-30834443 on Build GRCh38
rs764432631 Chr.17: 30834507-30834507 on Build GRCh38
rs147909444 Chr.17: 30834499-30834499 on Build GRCh38
rs777394586 Chr.17: 30834471-30834471 on Build GRCh38
rs17826219 Chr.17: 30834827-30834827 on Build GRCh38
rs201064814 Chr.17: 30834812-30834812 on Build GRCh38
rs150438654 Chr.17: 30834729-30834729 on Build GRCh38
rs560055952 Chr.17: 30834458-30834458 on Build GRCh38
rs145376542 Chr.17: 30834478-30834478 on Build GRCh38
rs756690738 Chr.17: 30834551-30834551 on Build GRCh38
rs751755706 Chr.17: 30834426-30834426 on Build GRCh38
rs1013100163 Chr.17: 30834818-30834818 on Build GRCh38
rs752385383 Chr.17: 30834413-30834413 on Build GRCh38
rs923747996 Chr.17: 30834817-30834817 on Build GRCh38
rs768574370 Chr.17: 30834639-30834639 on Build GRCh38
rs76545689 Chr.17: 30834764-30834764 on Build GRCh38
rs774244442 Chr.17: 30834662-30834662 on Build GRCh38
rs147045641 Chr.17: 30834800-30834800 on Build GRCh38
rs761143646 Chr.17: 30834834-30834834 on Build GRCh38
rs757397900 Chr.17: 30834429-30834429 on Build GRCh38
rs894616667 Chr.17: 30834807-30834807 on Build GRCh38
rs775276466 Chr.17: 30834624-30834624 on Build GRCh38
rs745441598 Chr.17: 30834738-30834738 on Build GRCh38
rs11080134 Chr.17: 30834485-30834485 on Build GRCh38
rs373317723 Chr.17: 30834459-30834459 on Build GRCh38
rs754290871 Chr.17: 30834557-30834557 on Build GRCh38
rs367906557 Chr.17: 30834495-30834495 on Build GRCh38
rs749211440 Chr.17: 30834756-30834756 on Build GRCh38
rs878985290 Chr.17: 30834813-30834813 on Build GRCh38
rs1016372285 Chr.17: 30834452-30834452 on Build GRCh38
rs766795863 Chr.17: 30834556-30834556 on Build GRCh38
rs747917492 Chr.17: 30834780-30834780 on Build GRCh38

View Full Product Details