Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30948826-30949371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAP2
Entrez Gene ID
55803
Gene Name
ArfGAP with dual PH domains 2
Gene Aliases
CENTA2, HSA272195, cent-b
UniGene
Hs.514063
Gene Chromosome Location
Chr.17: 30921736-30959193 on Build GRCh38
Transcripts
NM_018404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5793642 c.726A>T p.E242D Chr.17: 30949355-30949355 on Build GRCh38
1679692 c.685C>T p.R229C Chr.17: 30949314-30949314 on Build GRCh38
229302 c.714G>A p.M238I Chr.17: 30949343-30949343 on Build GRCh38
6198812 c.695G>A p.R232H Chr.17: 30949324-30949324 on Build GRCh38
5526335 c.684C>T p.L228L Chr.17: 30949313-30949313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535391181 Chr.17: 30949283-30949283 on Build GRCh38
rs950138204 Chr.17: 30949101-30949101 on Build GRCh38
rs1004728202 Chr.17: 30949183-30949184 on Build GRCh38
rs974644975 Chr.17: 30949216-30949216 on Build GRCh38
rs886461536 Chr.17: 30949173-30949173 on Build GRCh38
rs2232273 Chr.17: 30949150-30949150 on Build GRCh38
rs977249492 Chr.17: 30949015-30949015 on Build GRCh38
rs368594067 Chr.17: 30948902-30948902 on Build GRCh38
rs569393396 Chr.17: 30948988-30948988 on Build GRCh38
rs111431912 Chr.17: 30948901-30948901 on Build GRCh38
rs763876179 Chr.17: 30949139-30949139 on Build GRCh38
rs750080743 Chr.17: 30949289-30949289 on Build GRCh38
rs1017913407 Chr.17: 30948967-30948967 on Build GRCh38
rs991911809 Chr.17: 30949091-30949091 on Build GRCh38
rs765737517 Chr.17: 30949268-30949268 on Build GRCh38
rs755710322 Chr.17: 30949306-30949306 on Build GRCh38
rs897957722 Chr.17: 30948879-30948879 on Build GRCh38
rs1029153629 Chr.17: 30949225-30949225 on Build GRCh38
rs537666328 Chr.17: 30949241-30949241 on Build GRCh38
rs774117783 Chr.17: 30948877-30948877 on Build GRCh38
rs1007787013 Chr.17: 30949223-30949223 on Build GRCh38
rs1028363464 Chr.17: 30948897-30948897 on Build GRCh38
rs779427042 Chr.17: 30949314-30949314 on Build GRCh38
rs946374729 Chr.17: 30948983-30948983 on Build GRCh38
rs778773284 Chr.17: 30949332-30949332 on Build GRCh38
rs1005660770 Chr.17: 30948919-30948919 on Build GRCh38
rs552471362 Chr.17: 30949194-30949194 on Build GRCh38
rs185220687 Chr.17: 30948882-30948882 on Build GRCh38
rs113055344 Chr.17: 30949282-30949282 on Build GRCh38
rs950252218 Chr.17: 30949112-30949112 on Build GRCh38
rs368026623 Chr.17: 30949269-30949269 on Build GRCh38
rs993717724 Chr.17: 30948888-30948888 on Build GRCh38
rs746897235 Chr.17: 30949255-30949256 on Build GRCh38
rs966232994 Chr.17: 30949215-30949215 on Build GRCh38
rs570298715 Chr.17: 30949202-30949202 on Build GRCh38
rs1027640094 Chr.17: 30949161-30949161 on Build GRCh38
rs373389287 Chr.17: 30949278-30949278 on Build GRCh38
rs967238966 Chr.17: 30949012-30949012 on Build GRCh38
rs533840681 Chr.17: 30949151-30949151 on Build GRCh38
rs375059370 Chr.17: 30949100-30949100 on Build GRCh38
rs747968126 Chr.17: 30949345-30949345 on Build GRCh38
rs770891630 Chr.17: 30949264-30949264 on Build GRCh38
rs7213182 Chr.17: 30949029-30949029 on Build GRCh38
rs1029510788 Chr.17: 30948993-30948993 on Build GRCh38
rs935920385 Chr.17: 30949061-30949061 on Build GRCh38
rs754885895 Chr.17: 30949326-30949326 on Build GRCh38
rs370875162 Chr.17: 30949025-30949025 on Build GRCh38
rs370140572 Chr.17: 30949324-30949324 on Build GRCh38
rs555813486 Chr.17: 30949254-30949254 on Build GRCh38
rs202013881 Chr.17: 30949277-30949277 on Build GRCh38
rs1053381622 Chr.17: 30949049-30949049 on Build GRCh38
rs923576050 Chr.17: 30949273-30949273 on Build GRCh38
rs548301121 Chr.17: 30949133-30949133 on Build GRCh38
rs144736072 Chr.17: 30949323-30949323 on Build GRCh38
rs561106544 Chr.17: 30948989-30948989 on Build GRCh38

View Full Product Details