Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30953048-30953448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAP2
Entrez Gene ID
55803
Gene Name
ArfGAP with dual PH domains 2
Gene Aliases
CENTA2, HSA272195, cent-b
UniGene
Hs.514063
Gene Chromosome Location
Chr.17: 30921736-30959193 on Build GRCh38
Transcripts
NM_018404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3515805 c.796G>A p.G266R Chr.17: 30953342-30953342 on Build GRCh38
5504282 c.765C>T p.N255N Chr.17: 30953311-30953311 on Build GRCh38
4065302 c.742C>T p.L248F Chr.17: 30953288-30953288 on Build GRCh38
5762692 c.745G>A p.V249M Chr.17: 30953291-30953291 on Build GRCh38
3515804 c.762G>A p.R254R Chr.17: 30953308-30953308 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs560918709 Chr.17: 30953164-30953164 on Build GRCh38
rs745979784 Chr.17: 30953338-30953338 on Build GRCh38
rs974289121 Chr.17: 30953112-30953112 on Build GRCh38
rs2232275 Chr.17: 30953403-30953403 on Build GRCh38
rs769522049 Chr.17: 30953180-30953180 on Build GRCh38
rs758246178 Chr.17: 30953291-30953291 on Build GRCh38
rs768057029 Chr.17: 30953109-30953109 on Build GRCh38
rs1039584325 Chr.17: 30953110-30953110 on Build GRCh38
rs746749267 Chr.17: 30953325-30953325 on Build GRCh38
rs776316242 Chr.17: 30953396-30953396 on Build GRCh38
rs752607136 Chr.17: 30953290-30953290 on Build GRCh38
rs892942822 Chr.17: 30953418-30953418 on Build GRCh38
rs765979239 Chr.17: 30953392-30953392 on Build GRCh38
rs762190583 Chr.17: 30953384-30953384 on Build GRCh38
rs868036809 Chr.17: 30953270-30953270 on Build GRCh38
rs754492353 Chr.17: 30953259-30953259 on Build GRCh38
rs920053382 Chr.17: 30953169-30953169 on Build GRCh38
rs777537718 Chr.17: 30953308-30953308 on Build GRCh38
rs184174155 Chr.17: 30953192-30953192 on Build GRCh38
rs2232274 Chr.17: 30953262-30953262 on Build GRCh38
rs28449298 Chr.17: 30953184-30953184 on Build GRCh38
rs1023729164 Chr.17: 30953284-30953284 on Build GRCh38
rs749810909 Chr.17: 30953364-30953364 on Build GRCh38
rs754445355 Chr.17: 30953269-30953269 on Build GRCh38
rs892227877 Chr.17: 30953250-30953250 on Build GRCh38
rs780679367 Chr.17: 30953267-30953268 on Build GRCh38
rs985742018 Chr.17: 30953214-30953214 on Build GRCh38
rs2232276 Chr.17: 30953423-30953423 on Build GRCh38
rs769221913 Chr.17: 30953381-30953381 on Build GRCh38
rs112480699 Chr.17: 30953357-30953357 on Build GRCh38
rs764107717 Chr.17: 30953216-30953216 on Build GRCh38
rs528111044 Chr.17: 30953176-30953176 on Build GRCh38
rs1015708004 Chr.17: 30953090-30953090 on Build GRCh38
rs769975921 Chr.17: 30953354-30953354 on Build GRCh38
rs776201148 Chr.17: 30953360-30953360 on Build GRCh38
rs991050613 Chr.17: 30953275-30953275 on Build GRCh38
rs760755537 Chr.17: 30953209-30953209 on Build GRCh38
rs954230325 Chr.17: 30953421-30953421 on Build GRCh38
rs138525120 Chr.17: 30953179-30953179 on Build GRCh38
rs573944016 Chr.17: 30953382-30953382 on Build GRCh38
rs112682010 Chr.17: 30953228-30953228 on Build GRCh38
rs778861207 Chr.17: 30953277-30953277 on Build GRCh38
rs371471867 Chr.17: 30953230-30953230 on Build GRCh38
rs1001991044 Chr.17: 30953329-30953329 on Build GRCh38
rs961731701 Chr.17: 30953097-30953097 on Build GRCh38
rs974531501 Chr.17: 30953398-30953398 on Build GRCh38
rs766000180 Chr.17: 30953254-30953254 on Build GRCh38
rs367955205 Chr.17: 30953408-30953408 on Build GRCh38
rs548853111 Chr.17: 30953074-30953074 on Build GRCh38
rs912813798 Chr.17: 30953215-30953215 on Build GRCh38
rs771069186 Chr.17: 30953330-30953330 on Build GRCh38
rs902810409 Chr.17: 30953227-30953227 on Build GRCh38
rs139594986 Chr.17: 30953331-30953331 on Build GRCh38
rs1039658244 Chr.17: 30953410-30953410 on Build GRCh38
rs372525207 Chr.17: 30953294-30953294 on Build GRCh38
rs141540697 Chr.17: 30953246-30953246 on Build GRCh38

View Full Product Details