Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30954390-30954857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAP2
Entrez Gene ID
55803
Gene Name
ArfGAP with dual PH domains 2
Gene Aliases
CENTA2, HSA272195, cent-b
UniGene
Hs.514063
Gene Chromosome Location
Chr.17: 30921736-30959193 on Build GRCh38
Transcripts
NM_018404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1381999 c.811G>T p.E271* Chr.17: 30954484-30954484 on Build GRCh38
5775753 c.812A>T p.E271V Chr.17: 30954485-30954485 on Build GRCh38
3958165 c.837C>T p.A279A Chr.17: 30954510-30954510 on Build GRCh38
4065303 c.805-1G>A p.? Chr.17: 30954477-30954477 on Build GRCh38
4510723 c.846C>T p.C282C Chr.17: 30954519-30954519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776203025 Chr.17: 30954527-30954527 on Build GRCh38
rs751329619 Chr.17: 30954470-30954470 on Build GRCh38
rs915642474 Chr.17: 30954567-30954567 on Build GRCh38
rs746077502 Chr.17: 30954487-30954487 on Build GRCh38
rs972994479 Chr.17: 30954739-30954739 on Build GRCh38
rs762397571 Chr.17: 30954558-30954558 on Build GRCh38
rs774669803 Chr.17: 30954607-30954607 on Build GRCh38
rs759182146 Chr.17: 30954534-30954534 on Build GRCh38
rs531086712 Chr.17: 30954677-30954677 on Build GRCh38
rs775931806 Chr.17: 30954801-30954801 on Build GRCh38
rs770558642 Chr.17: 30954445-30954445 on Build GRCh38
rs928992877 Chr.17: 30954772-30954772 on Build GRCh38
rs751496637 Chr.17: 30954563-30954563 on Build GRCh38
rs973137561 Chr.17: 30954731-30954731 on Build GRCh38
rs757049256 Chr.17: 30954474-30954474 on Build GRCh38
rs199813260 Chr.17: 30954431-30954431 on Build GRCh38
rs748501963 Chr.17: 30954524-30954524 on Build GRCh38
rs757062249 Chr.17: 30954578-30954578 on Build GRCh38
rs764031082 Chr.17: 30954469-30954469 on Build GRCh38
rs556426891 Chr.17: 30954493-30954493 on Build GRCh38
rs764174762 Chr.17: 30954830-30954830 on Build GRCh38
rs889756492 Chr.17: 30954426-30954426 on Build GRCh38
rs758334259 Chr.17: 30954455-30954455 on Build GRCh38
rs774881193 Chr.17: 30954515-30954515 on Build GRCh38
rs756331978 Chr.17: 30954501-30954501 on Build GRCh38
rs1047761801 Chr.17: 30954774-30954774 on Build GRCh38
rs567889601 Chr.17: 30954745-30954745 on Build GRCh38
rs772459818 Chr.17: 30954526-30954526 on Build GRCh38
rs371476409 Chr.17: 30954438-30954438 on Build GRCh38
rs144506344 Chr.17: 30954678-30954678 on Build GRCh38
rs201617793 Chr.17: 30954560-30954560 on Build GRCh38
rs2232279 Chr.17: 30954507-30954507 on Build GRCh38
rs769309988 Chr.17: 30954509-30954509 on Build GRCh38
rs920230319 Chr.17: 30954766-30954766 on Build GRCh38
rs201799866 Chr.17: 30954508-30954508 on Build GRCh38
rs1025171994 Chr.17: 30954640-30954640 on Build GRCh38
rs984071586 Chr.17: 30954777-30954777 on Build GRCh38
rs774990978 Chr.17: 30954554-30954554 on Build GRCh38
rs766577829 Chr.17: 30954539-30954539 on Build GRCh38
rs764712927 Chr.17: 30954436-30954436 on Build GRCh38
rs199913340 Chr.17: 30954429-30954429 on Build GRCh38
rs772452066 Chr.17: 30954682-30954682 on Build GRCh38
rs746284010 Chr.17: 30954652-30954652 on Build GRCh38
rs781321318 Chr.17: 30954478-30954478 on Build GRCh38
rs760813115 Chr.17: 30954764-30954764 on Build GRCh38
rs960136802 Chr.17: 30954538-30954538 on Build GRCh38
rs944191682 Chr.17: 30954788-30954788 on Build GRCh38
rs764801341 Chr.17: 30954542-30954542 on Build GRCh38
rs2232280 Chr.17: 30954608-30954608 on Build GRCh38
rs767343992 Chr.17: 30954592-30954592 on Build GRCh38
rs931225277 Chr.17: 30954747-30954747 on Build GRCh38
rs761086214 Chr.17: 30954804-30954804 on Build GRCh38

View Full Product Details