Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 31305465-31305972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI2B
Entrez Gene ID
2124
Gene Name
ecotropic viral integration site 2B
Gene Aliases
CD361, D17S376, EVDB
UniGene
Hs.5509
Gene Chromosome Location
Chr.17: 31303766-31314149 on Build GRCh38
Transcripts
NM_006495.3 XM_005257946.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Gene Symbol
NF1
Entrez Gene ID
4763
Gene Name
neurofibromin 1
Gene Aliases
NFNS, VRNF, WSS
UniGene
Hs.113577
Gene Chromosome Location
Chr.17: 31094927-31377677 on Build GRCh38
Transcripts
NM_006495.3 XM_005257946.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
977461 c.32T>G p.F11C Chr.17: 31305578-31305578 on Build GRCh38
6556278 c.188C>A p.S63Y Chr.17: 31305467-31305467 on Build GRCh38
6556277 c.143C>A p.S48Y Chr.17: 31305467-31305467 on Build GRCh38
4172206 c.53C>A p.T18K Chr.17: 31305557-31305557 on Build GRCh38
4612364 c.106delT p.S36fs*92 Chr.17: 31305549-31305549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026472883 Chr.17: 31305885-31305885 on Build GRCh38
rs777799482 Chr.17: 31305638-31305638 on Build GRCh38
rs941071081 Chr.17: 31305558-31305558 on Build GRCh38
rs907472476 Chr.17: 31305806-31305806 on Build GRCh38
rs754101275 Chr.17: 31305625-31305625 on Build GRCh38
rs768495636 Chr.17: 31305668-31305668 on Build GRCh38
rs921186585 Chr.17: 31305622-31305622 on Build GRCh38
rs750146352 Chr.17: 31305590-31305591 on Build GRCh38
rs748117738 Chr.17: 31305536-31305536 on Build GRCh38
rs780540629 Chr.17: 31305491-31305491 on Build GRCh38
rs753236143 Chr.17: 31305637-31305637 on Build GRCh38
rs748094180 Chr.17: 31305670-31305670 on Build GRCh38
rs950975367 Chr.17: 31305892-31305892 on Build GRCh38
rs1003013442 Chr.17: 31305822-31305822 on Build GRCh38
rs564349361 Chr.17: 31305554-31305554 on Build GRCh38
rs755399921 Chr.17: 31305506-31305506 on Build GRCh38
rs576969125 Chr.17: 31305557-31305557 on Build GRCh38
rs374306730 Chr.17: 31305533-31305533 on Build GRCh38
rs150267160 Chr.17: 31305776-31305776 on Build GRCh38
rs903105392 Chr.17: 31305581-31305581 on Build GRCh38
rs368793224 Chr.17: 31305833-31305833 on Build GRCh38
rs1039835047 Chr.17: 31305879-31305879 on Build GRCh38
rs772172313 Chr.17: 31305680-31305680 on Build GRCh38
rs766161104 Chr.17: 31305623-31305623 on Build GRCh38
rs374849042 Chr.17: 31305633-31305633 on Build GRCh38
rs919474444 Chr.17: 31305713-31305713 on Build GRCh38
rs531300031 Chr.17: 31305636-31305636 on Build GRCh38
rs747123373 Chr.17: 31305641-31305641 on Build GRCh38
rs762833164 Chr.17: 31305565-31305565 on Build GRCh38
rs774353882 Chr.17: 31305567-31305567 on Build GRCh38
rs554250562 Chr.17: 31305685-31305685 on Build GRCh38
rs982551083 Chr.17: 31305910-31305910 on Build GRCh38
rs16972176 Chr.17: 31305646-31305646 on Build GRCh38
rs77954806 Chr.17: 31305647-31305647 on Build GRCh38
rs777889615 Chr.17: 31305556-31305556 on Build GRCh38
rs998770690 Chr.17: 31305936-31305940 on Build GRCh38
rs754289943 Chr.17: 31305492-31305492 on Build GRCh38
rs375405082 Chr.17: 31305489-31305489 on Build GRCh38
rs771183458 Chr.17: 31305559-31305559 on Build GRCh38
rs903053344 Chr.17: 31305934-31305934 on Build GRCh38
rs760368575 Chr.17: 31305620-31305620 on Build GRCh38
rs565290323 Chr.17: 31305850-31305850 on Build GRCh38
rs772378094 Chr.17: 31305570-31305570 on Build GRCh38
rs1050924360 Chr.17: 31305933-31305933 on Build GRCh38
rs774069085 Chr.17: 31305669-31305669 on Build GRCh38
rs929550361 Chr.17: 31305723-31305723 on Build GRCh38
rs1012930421 Chr.17: 31305711-31305711 on Build GRCh38
rs749998773 Chr.17: 31305618-31305618 on Build GRCh38
rs768424280 Chr.17: 31305566-31305566 on Build GRCh38
rs772195960 Chr.17: 31305540-31305540 on Build GRCh38
rs1050900648 Chr.17: 31305628-31305628 on Build GRCh38
rs541453289 Chr.17: 31305681-31305681 on Build GRCh38
rs540818953 Chr.17: 31305809-31305809 on Build GRCh38
rs141059622 Chr.17: 31305626-31305626 on Build GRCh38
rs370014149 Chr.17: 31305597-31305597 on Build GRCh38
rs200921973 Chr.17: 31305562-31305562 on Build GRCh38
rs561229698 Chr.17: 31305709-31305709 on Build GRCh38
rs766932041 Chr.17: 31305662-31305665 on Build GRCh38
rs190016033 Chr.17: 31305658-31305658 on Build GRCh38
rs941066938 Chr.17: 31305797-31305797 on Build GRCh38
rs145887114 Chr.17: 31305915-31305915 on Build GRCh38
rs1023184373 Chr.17: 31305742-31305742 on Build GRCh38

View Full Product Details