Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 31313842-31314365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI2B
Entrez Gene ID
2124
Gene Name
ecotropic viral integration site 2B
Gene Aliases
CD361, D17S376, EVDB
UniGene
Hs.5509
Gene Chromosome Location
Chr.17: 31303766-31314149 on Build GRCh38
Transcripts
NM_006495.3 XM_005257946.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Gene Symbol
NF1
Entrez Gene ID
4763
Gene Name
neurofibromin 1
Gene Aliases
NFNS, VRNF, WSS
UniGene
Hs.113577
Gene Chromosome Location
Chr.17: 31094927-31377677 on Build GRCh38
Transcripts
NM_006495.3 XM_005257946.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
27386 c.1_*del p.0 Chr.17: 31095311-31374156 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572820304 Chr.17: 31314143-31314143 on Build GRCh38
rs1001391733 Chr.17: 31314136-31314136 on Build GRCh38
rs983709385 Chr.17: 31314175-31314175 on Build GRCh38
rs970438683 Chr.17: 31314112-31314112 on Build GRCh38
rs1021713299 Chr.17: 31314096-31314096 on Build GRCh38
rs912070818 Chr.17: 31314316-31314316 on Build GRCh38
rs368521049 Chr.17: 31314227-31314227 on Build GRCh38
rs988105708 Chr.17: 31313997-31313997 on Build GRCh38
rs538574131 Chr.17: 31314182-31314182 on Build GRCh38
rs905664341 Chr.17: 31314108-31314108 on Build GRCh38
rs754266992 Chr.17: 31314083-31314083 on Build GRCh38
rs565544412 Chr.17: 31313890-31313890 on Build GRCh38
rs912939045 Chr.17: 31314078-31314078 on Build GRCh38
rs987643903 Chr.17: 31314284-31314284 on Build GRCh38
rs1038489625 Chr.17: 31314184-31314184 on Build GRCh38
rs1050085511 Chr.17: 31313870-31313870 on Build GRCh38
rs912161316 Chr.17: 31314024-31314024 on Build GRCh38
rs935611178 Chr.17: 31314050-31314050 on Build GRCh38
rs189487695 Chr.17: 31314147-31314147 on Build GRCh38
rs919706419 Chr.17: 31314198-31314198 on Build GRCh38
rs866914049 Chr.17: 31313883-31313883 on Build GRCh38
rs922062977 Chr.17: 31313886-31313886 on Build GRCh38
rs994885568 Chr.17: 31314258-31314258 on Build GRCh38
rs781488322 Chr.17: 31313950-31313950 on Build GRCh38
rs1031505805 Chr.17: 31313928-31313928 on Build GRCh38
rs895717160 Chr.17: 31314169-31314169 on Build GRCh38
rs1029101713 Chr.17: 31314272-31314272 on Build GRCh38
rs575030370 Chr.17: 31314289-31314289 on Build GRCh38
rs890819616 Chr.17: 31314275-31314275 on Build GRCh38
rs534465728 Chr.17: 31313926-31313926 on Build GRCh38
rs980138241 Chr.17: 31314119-31314119 on Build GRCh38
rs748366805 Chr.17: 31314339-31314339 on Build GRCh38
rs9898921 Chr.17: 31313876-31313876 on Build GRCh38
rs751017909 Chr.17: 31314214-31314214 on Build GRCh38
rs908045398 Chr.17: 31314177-31314177 on Build GRCh38
rs559310309 Chr.17: 31313985-31313985 on Build GRCh38
rs1008177631 Chr.17: 31313891-31313891 on Build GRCh38
rs932282232 Chr.17: 31314213-31314213 on Build GRCh38
rs956974465 Chr.17: 31314331-31314331 on Build GRCh38
rs577487206 Chr.17: 31314070-31314070 on Build GRCh38
rs947129921 Chr.17: 31314084-31314084 on Build GRCh38

View Full Product Details