Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 31316894-31317440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI2A
Entrez Gene ID
2123
Gene Name
ecotropic viral integration site 2A
Gene Aliases
EVDA, EVI-2A, EVI2
UniGene
Hs.591198
Gene Chromosome Location
Chr.17: 31316410-31321749 on Build GRCh38
Transcripts
NM_001003927.2 NM_014210.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Gene Symbol
NF1
Entrez Gene ID
4763
Gene Name
neurofibromin 1
Gene Aliases
NFNS, VRNF, WSS
UniGene
Hs.113577
Gene Chromosome Location
Chr.17: 31094927-31377677 on Build GRCh38
Transcripts
NM_001003927.2 NM_014210.3 NM_000267.3 NM_001042492.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
27386 c.1_*del p.0 Chr.17: 31095311-31374156 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs536028438 Chr.17: 31317008-31317008 on Build GRCh38
rs911466925 Chr.17: 31317085-31317085 on Build GRCh38
rs867757222 Chr.17: 31317194-31317194 on Build GRCh38
rs546367698 Chr.17: 31317166-31317166 on Build GRCh38
rs879074792 Chr.17: 31317385-31317385 on Build GRCh38
rs77652736 Chr.17: 31317198-31317198 on Build GRCh38
rs141481799 Chr.17: 31317236-31317236 on Build GRCh38
rs866279339 Chr.17: 31317376-31317376 on Build GRCh38
rs766765911 Chr.17: 31316952-31316952 on Build GRCh38
rs200395855 Chr.17: 31317405-31317405 on Build GRCh38
rs895043832 Chr.17: 31317269-31317270 on Build GRCh38
rs529606388 Chr.17: 31317325-31317325 on Build GRCh38
rs34766876 Chr.17: 31317368-31317369 on Build GRCh38
rs3138611 Chr.17: 31317370-31317371 on Build GRCh38
rs571315603 Chr.17: 31317403-31317404 on Build GRCh38
rs952803827 Chr.17: 31317390-31317391 on Build GRCh38
rs1000666845 Chr.17: 31317234-31317234 on Build GRCh38
rs56932012 Chr.17: 31317404-31317405 on Build GRCh38
rs993785828 Chr.17: 31316926-31316926 on Build GRCh38
rs35744604 Chr.17: 31317313-31317314 on Build GRCh38
rs938291389 Chr.17: 31317146-31317146 on Build GRCh38
rs878999114 Chr.17: 31317387-31317387 on Build GRCh38
rs984346922 Chr.17: 31317052-31317052 on Build GRCh38
rs1007915350 Chr.17: 31317386-31317386 on Build GRCh38
rs980054393 Chr.17: 31317093-31317093 on Build GRCh38
rs970723230 Chr.17: 31317329-31317329 on Build GRCh38
rs112194571 Chr.17: 31317403-31317403 on Build GRCh38
rs567367085 Chr.17: 31316980-31316980 on Build GRCh38
rs770454886 Chr.17: 31317259-31317259 on Build GRCh38
rs147552041 Chr.17: 31317394-31317395 on Build GRCh38
rs970833649 Chr.17: 31316927-31316927 on Build GRCh38
rs796212150 Chr.17: 31317369-31317370 on Build GRCh38
rs1027573612 Chr.17: 31316934-31316934 on Build GRCh38
rs183131835 Chr.17: 31317220-31317220 on Build GRCh38
rs796212451 Chr.17: 31317103-31317103 on Build GRCh38
rs566363619 Chr.17: 31317371-31317371 on Build GRCh38
rs1017479913 Chr.17: 31316948-31316948 on Build GRCh38
rs542219039 Chr.17: 31317238-31317238 on Build GRCh38
rs763163263 Chr.17: 31316946-31316946 on Build GRCh38
rs66803216 Chr.17: 31317369-31317370 on Build GRCh38
rs994212133 Chr.17: 31317361-31317361 on Build GRCh38
rs897443055 Chr.17: 31317254-31317254 on Build GRCh38
rs867097777 Chr.17: 31317078-31317078 on Build GRCh38
rs1012617282 Chr.17: 31317271-31317271 on Build GRCh38
rs976206892 Chr.17: 31316957-31316957 on Build GRCh38
rs148316543 Chr.17: 31316935-31316935 on Build GRCh38
rs147491890 Chr.17: 31317313-31317313 on Build GRCh38
rs774781504 Chr.17: 31316977-31316977 on Build GRCh38
rs564807370 Chr.17: 31317282-31317286 on Build GRCh38
rs993194886 Chr.17: 31317360-31317360 on Build GRCh38

View Full Product Details