Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17212793-17213297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLCN
Entrez Gene ID
201163
Gene Name
folliculin
Gene Aliases
BHD, FLCL
UniGene
Hs.31652
Gene Chromosome Location
Chr.17: 17206945-17237188 on Build GRCh38
Transcripts
NM_144997.5 XM_011523714.2 XM_011523716.1 XM_011523718.2 XM_011523719.2 XM_011523721.2 XM_017024304.1 XM_017024305.1 XM_017024306.1 XM_017024307.1 XM_017024309.1

SNP Detail

SNP ID Chromosome Location
rs574547835 Chr.17: 17213129-17213129 on Build GRCh38
rs977629513 Chr.17: 17213193-17213193 on Build GRCh38
rs188840802 Chr.17: 17213252-17213252 on Build GRCh38
rs988015663 Chr.17: 17213159-17213159 on Build GRCh38
rs566100851 Chr.17: 17213075-17213075 on Build GRCh38
rs991236394 Chr.17: 17212955-17212955 on Build GRCh38
rs935920439 Chr.17: 17213006-17213006 on Build GRCh38
rs947184141 Chr.17: 17213015-17213015 on Build GRCh38
rs550746288 Chr.17: 17212938-17212938 on Build GRCh38
rs775700421 Chr.17: 17212960-17212960 on Build GRCh38
rs1019110164 Chr.17: 17212885-17212885 on Build GRCh38
rs963700738 Chr.17: 17213134-17213134 on Build GRCh38
rs574471240 Chr.17: 17213173-17213173 on Build GRCh38
rs750900491 Chr.17: 17212827-17212827 on Build GRCh38
rs7224335 Chr.17: 17213230-17213230 on Build GRCh38
rs3803761 Chr.17: 17213098-17213098 on Build GRCh38
rs558952732 Chr.17: 17212916-17212916 on Build GRCh38
rs780240981 Chr.17: 17213017-17213017 on Build GRCh38
rs924500249 Chr.17: 17212985-17212985 on Build GRCh38
rs1033116770 Chr.17: 17212837-17212837 on Build GRCh38
rs770643935 Chr.17: 17212834-17212834 on Build GRCh38
rs537259057 Chr.17: 17213145-17213145 on Build GRCh38
rs886052658 Chr.17: 17212959-17212960 on Build GRCh38
rs974975943 Chr.17: 17213256-17213256 on Build GRCh38
rs912113115 Chr.17: 17212919-17212919 on Build GRCh38
rs1026941756 Chr.17: 17212894-17212894 on Build GRCh38
rs1056992450 Chr.17: 17213113-17213113 on Build GRCh38
rs184006653 Chr.17: 17213127-17213127 on Build GRCh38
rs7224213 Chr.17: 17213213-17213213 on Build GRCh38
rs963455195 Chr.17: 17212853-17212853 on Build GRCh38
rs189888135 Chr.17: 17212941-17212941 on Build GRCh38
rs994970871 Chr.17: 17213140-17213140 on Build GRCh38
rs917123087 Chr.17: 17212959-17212960 on Build GRCh38
rs889112411 Chr.17: 17213030-17213030 on Build GRCh38
rs1026777733 Chr.17: 17213141-17213141 on Build GRCh38
rs919468352 Chr.17: 17212862-17212862 on Build GRCh38
rs139529824 Chr.17: 17212861-17212861 on Build GRCh38
rs935955441 Chr.17: 17213225-17213225 on Build GRCh38
rs539709518 Chr.17: 17212975-17212975 on Build GRCh38
rs1041638151 Chr.17: 17212984-17212984 on Build GRCh38
rs12602675 Chr.17: 17213262-17213262 on Build GRCh38
rs181935335 Chr.17: 17213115-17213115 on Build GRCh38
rs972349338 Chr.17: 17212887-17212887 on Build GRCh38
rs1012768985 Chr.17: 17213128-17213128 on Build GRCh38
rs937098560 Chr.17: 17212933-17212933 on Build GRCh38
rs966172286 Chr.17: 17212886-17212886 on Build GRCh38
rs1040401893 Chr.17: 17213056-17213056 on Build GRCh38
rs909539407 Chr.17: 17212922-17212922 on Build GRCh38
rs920912515 Chr.17: 17212967-17212967 on Build GRCh38
rs952429267 Chr.17: 17213110-17213110 on Build GRCh38
rs903021698 Chr.17: 17213093-17213093 on Build GRCh38
rs778371655 Chr.17: 17213156-17213159 on Build GRCh38
rs530771625 Chr.17: 17212914-17212914 on Build GRCh38
rs199572622 Chr.17: 17213021-17213021 on Build GRCh38
rs146851795 Chr.17: 17213175-17213175 on Build GRCh38
rs951094974 Chr.17: 17212898-17212898 on Build GRCh38
rs143768653 Chr.17: 17212869-17212869 on Build GRCh38
rs943697642 Chr.17: 17212959-17212959 on Build GRCh38

View Full Product Details