Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18039789-18040292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GID4
Entrez Gene ID
79018
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C17orf39, VID24
UniGene
Hs.187422
Gene Chromosome Location
Chr.17: 18039297-18068405 on Build GRCh38
Transcripts
NM_024052.4 XM_005256799.4 XM_005256800.1 XM_011524008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425208 c.352G>A p.A118T Chr.17: 18039816-18039816 on Build GRCh38
1381268 c.353C>T p.A118V Chr.17: 18039817-18039817 on Build GRCh38
4745716 c.384C>T p.F128F Chr.17: 18039848-18039848 on Build GRCh38
6364245 c.411C>A p.N137K Chr.17: 18039875-18039875 on Build GRCh38
6403039 c.346G>A p.G116S Chr.17: 18039810-18039810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771301487 Chr.17: 18039908-18039908 on Build GRCh38
rs536242201 Chr.17: 18040248-18040248 on Build GRCh38
rs372694605 Chr.17: 18040208-18040208 on Build GRCh38
rs948695030 Chr.17: 18040255-18040255 on Build GRCh38
rs955487495 Chr.17: 18039921-18039921 on Build GRCh38
rs1005606069 Chr.17: 18040242-18040242 on Build GRCh38
rs750720978 Chr.17: 18039908-18039909 on Build GRCh38
rs753642479 Chr.17: 18039920-18039920 on Build GRCh38
rs922953799 Chr.17: 18040055-18040055 on Build GRCh38
rs752632729 Chr.17: 18039914-18039914 on Build GRCh38
rs970748378 Chr.17: 18039927-18039927 on Build GRCh38
rs772264496 Chr.17: 18040268-18040268 on Build GRCh38
rs778674493 Chr.17: 18039973-18039973 on Build GRCh38
rs948847581 Chr.17: 18039965-18039965 on Build GRCh38
rs892363735 Chr.17: 18040091-18040091 on Build GRCh38
rs915914517 Chr.17: 18040251-18040251 on Build GRCh38
rs961902465 Chr.17: 18040259-18040259 on Build GRCh38
rs760457321 Chr.17: 18039915-18039915 on Build GRCh38
rs1046628119 Chr.17: 18040143-18040143 on Build GRCh38
rs975539508 Chr.17: 18039895-18039895 on Build GRCh38
rs778457004 Chr.17: 18039929-18039929 on Build GRCh38
rs781586887 Chr.17: 18039850-18039850 on Build GRCh38
rs748538986 Chr.17: 18039942-18039942 on Build GRCh38
rs910319292 Chr.17: 18040102-18040102 on Build GRCh38
rs1050020829 Chr.17: 18040097-18040097 on Build GRCh38
rs529229797 Chr.17: 18039951-18039951 on Build GRCh38
rs982947644 Chr.17: 18040169-18040169 on Build GRCh38
rs766966067 Chr.17: 18039940-18039941 on Build GRCh38
rs571269621 Chr.17: 18040128-18040128 on Build GRCh38
rs955560785 Chr.17: 18040149-18040149 on Build GRCh38
rs936383609 Chr.17: 18040110-18040110 on Build GRCh38
rs551910203 Chr.17: 18040117-18040117 on Build GRCh38
rs138787084 Chr.17: 18039893-18039893 on Build GRCh38
rs1038358825 Chr.17: 18039943-18039943 on Build GRCh38
rs763213923 Chr.17: 18039905-18039905 on Build GRCh38
rs964026872 Chr.17: 18040112-18040112 on Build GRCh38
rs192426094 Chr.17: 18040024-18040024 on Build GRCh38
rs915867626 Chr.17: 18039953-18039953 on Build GRCh38
rs1032447528 Chr.17: 18039841-18039841 on Build GRCh38
rs932974180 Chr.17: 18040095-18040095 on Build GRCh38
rs553878929 Chr.17: 18040180-18040180 on Build GRCh38
rs966063010 Chr.17: 18040035-18040035 on Build GRCh38
rs778333717 Chr.17: 18039815-18039815 on Build GRCh38
rs909007948 Chr.17: 18039978-18039978 on Build GRCh38
rs1002220807 Chr.17: 18040188-18040188 on Build GRCh38
rs1025428106 Chr.17: 18039917-18039917 on Build GRCh38
rs908820160 Chr.17: 18040215-18040215 on Build GRCh38
rs1003323270 Chr.17: 18040008-18040008 on Build GRCh38
rs748535840 Chr.17: 18039857-18039857 on Build GRCh38
rs76769873 Chr.17: 18040241-18040241 on Build GRCh38
rs996613500 Chr.17: 18040116-18040116 on Build GRCh38
rs1033658634 Chr.17: 18040198-18040198 on Build GRCh38
rs377621102 Chr.17: 18039837-18039837 on Build GRCh38
rs759576226 Chr.17: 18039911-18039911 on Build GRCh38
rs1045751943 Chr.17: 18039986-18039986 on Build GRCh38
rs1022488172 Chr.17: 18039836-18039836 on Build GRCh38
rs971999037 Chr.17: 18040037-18040037 on Build GRCh38
rs906383026 Chr.17: 18039992-18039992 on Build GRCh38
rs749915459 Chr.17: 18039932-18039932 on Build GRCh38
rs370576830 Chr.17: 18039944-18039944 on Build GRCh38
rs906558681 Chr.17: 18040182-18040182 on Build GRCh38
rs773720979 Chr.17: 18039896-18039896 on Build GRCh38
rs769163999 Chr.17: 18039839-18039839 on Build GRCh38
rs763810094 Chr.17: 18039919-18039919 on Build GRCh38
rs569198470 Chr.17: 18039988-18039988 on Build GRCh38
rs1016546483 Chr.17: 18040256-18040256 on Build GRCh38
rs756850367 Chr.17: 18039925-18039925 on Build GRCh38
rs1021976343 Chr.17: 18040107-18040107 on Build GRCh38
rs767468920 Chr.17: 18039912-18039912 on Build GRCh38
rs533855570 Chr.17: 18040141-18040141 on Build GRCh38
rs774449450 Chr.17: 18040157-18040157 on Build GRCh38
rs774450115 Chr.17: 18039909-18039909 on Build GRCh38
rs763382338 Chr.17: 18039922-18039922 on Build GRCh38
rs553431769 Chr.17: 18039963-18039963 on Build GRCh38
rs757891822 Chr.17: 18039937-18039937 on Build GRCh38
rs374149732 Chr.17: 18039830-18039830 on Build GRCh38

View Full Product Details