Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18066640-18067133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GID4
Entrez Gene ID
79018
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C17orf39, VID24
UniGene
Hs.187422
Gene Chromosome Location
Chr.17: 18039297-18068405 on Build GRCh38
Transcripts
NM_024052.4 XM_005256800.1

SNP Detail

SNP ID Chromosome Location
rs937644456 Chr.17: 18067031-18067031 on Build GRCh38
rs955237481 Chr.17: 18066934-18066934 on Build GRCh38
rs1006263362 Chr.17: 18066869-18066869 on Build GRCh38
rs962033154 Chr.17: 18066873-18066873 on Build GRCh38
rs942394011 Chr.17: 18066698-18066698 on Build GRCh38
rs8073401 Chr.17: 18067057-18067057 on Build GRCh38
rs994008413 Chr.17: 18066891-18066891 on Build GRCh38
rs968677620 Chr.17: 18066721-18066721 on Build GRCh38
rs918420258 Chr.17: 18066665-18066665 on Build GRCh38
rs76619863 Chr.17: 18067091-18067091 on Build GRCh38
rs150150097 Chr.17: 18066961-18066961 on Build GRCh38
rs919208664 Chr.17: 18066867-18066867 on Build GRCh38
rs566265544 Chr.17: 18066696-18066696 on Build GRCh38
rs1000902940 Chr.17: 18066725-18066725 on Build GRCh38
rs911171876 Chr.17: 18067018-18067018 on Build GRCh38
rs758117520 Chr.17: 18066896-18066897 on Build GRCh38
rs1056591861 Chr.17: 18066685-18066685 on Build GRCh38
rs1003027870 Chr.17: 18066812-18066812 on Build GRCh38
rs896216932 Chr.17: 18067062-18067062 on Build GRCh38
rs1016688535 Chr.17: 18066870-18066870 on Build GRCh38
rs1054041768 Chr.17: 18066715-18066715 on Build GRCh38
rs926282810 Chr.17: 18066951-18066951 on Build GRCh38
rs546083763 Chr.17: 18067008-18067008 on Build GRCh38
rs552553846 Chr.17: 18066693-18066693 on Build GRCh38
rs140839416 Chr.17: 18066858-18066858 on Build GRCh38
rs188988227 Chr.17: 18066733-18066733 on Build GRCh38
rs933803828 Chr.17: 18066680-18066680 on Build GRCh38
rs1042938609 Chr.17: 18067076-18067076 on Build GRCh38
rs986693404 Chr.17: 18066690-18066690 on Build GRCh38
rs1009751407 Chr.17: 18067066-18067066 on Build GRCh38
rs772653659 Chr.17: 18066916-18066916 on Build GRCh38
rs74633559 Chr.17: 18066728-18066728 on Build GRCh38
rs2955372 Chr.17: 18066915-18066915 on Build GRCh38
rs963937600 Chr.17: 18067035-18067035 on Build GRCh38
rs907376608 Chr.17: 18066789-18066789 on Build GRCh38
rs574177034 Chr.17: 18066801-18066801 on Build GRCh38
rs912618979 Chr.17: 18066780-18066780 on Build GRCh38
rs370898767 Chr.17: 18066785-18066785 on Build GRCh38
rs1056079470 Chr.17: 18066813-18066813 on Build GRCh38
rs541380701 Chr.17: 18066962-18066962 on Build GRCh38
rs892767764 Chr.17: 18066727-18066727 on Build GRCh38
rs573036200 Chr.17: 18067085-18067085 on Build GRCh38

View Full Product Details