Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18389025-18389519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVPLL
Entrez Gene ID
645027
Gene Name
envoplakin like
Gene Aliases
-
UniGene
Hs.462400
Gene Chromosome Location
Chr.17: 18377673-18389652 on Build GRCh38
Transcripts
NM_001145127.1 XM_011523982.2 XM_011523985.2 XM_011523986.2

SNP Detail

SNP ID Chromosome Location
rs1055780075 Chr.17: 18389133-18389133 on Build GRCh38
rs1032534041 Chr.17: 18389209-18389209 on Build GRCh38
rs957790709 Chr.17: 18389225-18389226 on Build GRCh38
rs1056249024 Chr.17: 18389357-18389357 on Build GRCh38
rs909981011 Chr.17: 18389325-18389325 on Build GRCh38
rs1001970310 Chr.17: 18389112-18389112 on Build GRCh38
rs536637026 Chr.17: 18389406-18389406 on Build GRCh38
rs950028173 Chr.17: 18389383-18389383 on Build GRCh38
rs199697056 Chr.17: 18389140-18389140 on Build GRCh38
rs902416101 Chr.17: 18389430-18389430 on Build GRCh38
rs1013516319 Chr.17: 18389145-18389145 on Build GRCh38
rs930108021 Chr.17: 18389047-18389047 on Build GRCh38
rs985587559 Chr.17: 18389227-18389227 on Build GRCh38
rs984293970 Chr.17: 18389318-18389318 on Build GRCh38
rs600657 Chr.17: 18389044-18389044 on Build GRCh38
rs662914 Chr.17: 18389498-18389498 on Build GRCh38
rs1048997769 Chr.17: 18389060-18389060 on Build GRCh38
rs1017911450 Chr.17: 18389241-18389241 on Build GRCh38
rs937460413 Chr.17: 18389327-18389327 on Build GRCh38
rs964983810 Chr.17: 18389254-18389254 on Build GRCh38
rs565563998 Chr.17: 18389093-18389093 on Build GRCh38
rs528338526 Chr.17: 18389094-18389094 on Build GRCh38
rs532648577 Chr.17: 18389045-18389045 on Build GRCh38
rs895821847 Chr.17: 18389374-18389374 on Build GRCh38
rs930172206 Chr.17: 18389309-18389309 on Build GRCh38
rs976977406 Chr.17: 18389284-18389284 on Build GRCh38
rs568042985 Chr.17: 18389207-18389207 on Build GRCh38
rs373278706 Chr.17: 18389117-18389117 on Build GRCh38
rs575354117 Chr.17: 18389162-18389162 on Build GRCh38
rs556954863 Chr.17: 18389465-18389465 on Build GRCh38
rs902386428 Chr.17: 18389190-18389190 on Build GRCh38
rs551906591 Chr.17: 18389053-18389053 on Build GRCh38

View Full Product Details