Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18797383-18797885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TVP23B
Entrez Gene ID
51030
Gene Name
trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
Gene Aliases
CGI-148, FAM18B, FAM18B1, NPD008, YDR084C
UniGene
Hs.87295
Gene Chromosome Location
Chr.17: 18780995-18806713 on Build GRCh38
Transcripts
NM_001316919.1 NM_001316920.1 NM_001316921.1 NM_001316922.1 NM_001316923.1 NM_001316924.1 NM_016078.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260787 c.294A>G p.E98E Chr.17: 18797632-18797632 on Build GRCh38
298088 c.272G>A p.R91H Chr.17: 18797610-18797610 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112599260 Chr.17: 18797741-18797741 on Build GRCh38
rs779077584 Chr.17: 18797453-18797453 on Build GRCh38
rs765156300 Chr.17: 18797611-18797611 on Build GRCh38
rs577643984 Chr.17: 18797498-18797498 on Build GRCh38
rs9908423 Chr.17: 18797530-18797530 on Build GRCh38
rs907052861 Chr.17: 18797722-18797722 on Build GRCh38
rs752880071 Chr.17: 18797678-18797679 on Build GRCh38
rs1022258745 Chr.17: 18797815-18797815 on Build GRCh38
rs762752544 Chr.17: 18797641-18797641 on Build GRCh38
rs998768752 Chr.17: 18797735-18797735 on Build GRCh38
rs772508212 Chr.17: 18797706-18797706 on Build GRCh38
rs528437306 Chr.17: 18797739-18797739 on Build GRCh38
rs779564909 Chr.17: 18797696-18797696 on Build GRCh38
rs767567168 Chr.17: 18797676-18797676 on Build GRCh38
rs771538573 Chr.17: 18797554-18797554 on Build GRCh38
rs761798180 Chr.17: 18797610-18797610 on Build GRCh38
rs191789765 Chr.17: 18797495-18797495 on Build GRCh38
rs1012684057 Chr.17: 18797684-18797684 on Build GRCh38
rs750296616 Chr.17: 18797631-18797631 on Build GRCh38
rs542314521 Chr.17: 18797753-18797753 on Build GRCh38
rs750694069 Chr.17: 18797843-18797843 on Build GRCh38
rs760758636 Chr.17: 18797600-18797600 on Build GRCh38
rs776551141 Chr.17: 18797606-18797606 on Build GRCh38
rs780799440 Chr.17: 18797677-18797677 on Build GRCh38
rs779474959 Chr.17: 18797562-18797562 on Build GRCh38
rs964075945 Chr.17: 18797854-18797854 on Build GRCh38
rs766043089 Chr.17: 18797643-18797643 on Build GRCh38
rs754560826 Chr.17: 18797659-18797659 on Build GRCh38
rs751259631 Chr.17: 18797651-18797651 on Build GRCh38
rs746227826 Chr.17: 18797565-18797565 on Build GRCh38
rs558658841 Chr.17: 18797521-18797521 on Build GRCh38
rs559803655 Chr.17: 18797725-18797725 on Build GRCh38
rs768812296 Chr.17: 18797601-18797601 on Build GRCh38
rs975525363 Chr.17: 18797460-18797460 on Build GRCh38
rs746317594 Chr.17: 18797705-18797705 on Build GRCh38
rs375069393 Chr.17: 18797593-18797593 on Build GRCh38
rs538655359 Chr.17: 18797456-18797456 on Build GRCh38
rs1010821262 Chr.17: 18797778-18797778 on Build GRCh38
rs780240860 Chr.17: 18797716-18797716 on Build GRCh38
rs956727991 Chr.17: 18797767-18797767 on Build GRCh38
rs752237750 Chr.17: 18797680-18797680 on Build GRCh38
rs934243314 Chr.17: 18797525-18797525 on Build GRCh38
rs757903447 Chr.17: 18797690-18797690 on Build GRCh38
rs775793170 Chr.17: 18797597-18797597 on Build GRCh38

View Full Product Details