Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18976666-18977165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83G
Entrez Gene ID
644815
Gene Name
family with sequence similarity 83 member G
Gene Aliases
PAWS1
UniGene
Hs.710727
Gene Chromosome Location
Chr.17: 18968789-19006144 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Gene Symbol
SLC5A10
Entrez Gene ID
125206
Gene Name
solute carrier family 5 member 10
Gene Aliases
SGLT-5, SGLT5
UniGene
Hs.462418
Gene Chromosome Location
Chr.17: 18950676-19020691 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7390711 c.704C>T p.P235L Chr.17: 18976960-18976960 on Build GRCh38
6408505 c.943A>G p.I315V Chr.17: 18976950-18976950 on Build GRCh38
4064579 c.970G>A p.A324T Chr.17: 18976977-18976977 on Build GRCh38
4064580 c.721G>A p.A241T Chr.17: 18976977-18976977 on Build GRCh38
5092859 c.896C>T p.A299V Chr.17: 18976903-18976903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547247312 Chr.17: 18976996-18976996 on Build GRCh38
rs770542664 Chr.17: 18977073-18977073 on Build GRCh38
rs968083627 Chr.17: 18977032-18977032 on Build GRCh38
rs771681407 Chr.17: 18976980-18976980 on Build GRCh38
rs761410107 Chr.17: 18976866-18976866 on Build GRCh38
rs187379476 Chr.17: 18976758-18976758 on Build GRCh38
rs978219876 Chr.17: 18976717-18976717 on Build GRCh38
rs746157510 Chr.17: 18977041-18977041 on Build GRCh38
rs749776194 Chr.17: 18977055-18977055 on Build GRCh38
rs531697809 Chr.17: 18976888-18976888 on Build GRCh38
rs758459672 Chr.17: 18976935-18976935 on Build GRCh38
rs748152199 Chr.17: 18976914-18976914 on Build GRCh38
rs574050907 Chr.17: 18976777-18976777 on Build GRCh38
rs778918413 Chr.17: 18976808-18976808 on Build GRCh38
rs560493309 Chr.17: 18976726-18976726 on Build GRCh38
rs376643051 Chr.17: 18976806-18976806 on Build GRCh38
rs746598709 Chr.17: 18976997-18976997 on Build GRCh38
rs1046841367 Chr.17: 18976975-18976975 on Build GRCh38
rs778828313 Chr.17: 18976903-18976903 on Build GRCh38
rs772217660 Chr.17: 18976830-18976830 on Build GRCh38
rs146394624 Chr.17: 18976839-18976839 on Build GRCh38
rs781569572 Chr.17: 18976960-18976960 on Build GRCh38
rs1031124909 Chr.17: 18977072-18977072 on Build GRCh38
rs747223859 Chr.17: 18976954-18976954 on Build GRCh38
rs765865991 Chr.17: 18976860-18976860 on Build GRCh38
rs751611936 Chr.17: 18977018-18977018 on Build GRCh38
rs17842753 Chr.17: 18976693-18976693 on Build GRCh38
rs947695051 Chr.17: 18976895-18976895 on Build GRCh38
rs759236995 Chr.17: 18977083-18977083 on Build GRCh38
rs767198712 Chr.17: 18976880-18976880 on Build GRCh38
rs920358449 Chr.17: 18976786-18976786 on Build GRCh38
rs750073061 Chr.17: 18976881-18976881 on Build GRCh38
rs777763111 Chr.17: 18976952-18976952 on Build GRCh38
rs978166167 Chr.17: 18977064-18977064 on Build GRCh38
rs763451815 Chr.17: 18976872-18976873 on Build GRCh38
rs781271717 Chr.17: 18977026-18977026 on Build GRCh38
rs549340867 Chr.17: 18977046-18977046 on Build GRCh38
rs747253351 Chr.17: 18976833-18976833 on Build GRCh38
rs973455050 Chr.17: 18976780-18976780 on Build GRCh38
rs752518946 Chr.17: 18977094-18977094 on Build GRCh38
rs760047028 Chr.17: 18976853-18976853 on Build GRCh38
rs1027724922 Chr.17: 18977010-18977010 on Build GRCh38
rs757441937 Chr.17: 18977023-18977023 on Build GRCh38
rs752609210 Chr.17: 18977006-18977006 on Build GRCh38
rs201925576 Chr.17: 18976796-18976796 on Build GRCh38
rs771303153 Chr.17: 18976834-18976834 on Build GRCh38
rs118127777 Chr.17: 18976703-18976703 on Build GRCh38
rs538343611 Chr.17: 18977109-18977109 on Build GRCh38
rs140249808 Chr.17: 18977066-18977068 on Build GRCh38
rs946089718 Chr.17: 18976904-18976904 on Build GRCh38
rs753603133 Chr.17: 18977003-18977003 on Build GRCh38
rs551594244 Chr.17: 18976953-18976953 on Build GRCh38
rs961748949 Chr.17: 18976709-18976709 on Build GRCh38
rs764948983 Chr.17: 18976959-18976971 on Build GRCh38
rs766162266 Chr.17: 18976884-18976884 on Build GRCh38
rs773937697 Chr.17: 18976863-18976863 on Build GRCh38
rs1008050842 Chr.17: 18976998-18976998 on Build GRCh38
rs770355239 Chr.17: 18976970-18976970 on Build GRCh38
rs2472715 Chr.17: 18976955-18976955 on Build GRCh38
rs770265271 Chr.17: 18977043-18977043 on Build GRCh38
rs61738464 Chr.17: 18976976-18976976 on Build GRCh38
rs202168324 Chr.17: 18976974-18976974 on Build GRCh38
rs753372898 Chr.17: 18976879-18976879 on Build GRCh38
rs776136950 Chr.17: 18977077-18977077 on Build GRCh38
rs772723125 Chr.17: 18977060-18977060 on Build GRCh38
rs772632240 Chr.17: 18976990-18976990 on Build GRCh38
rs769070332 Chr.17: 18977059-18977059 on Build GRCh38
rs374026119 Chr.17: 18977004-18977004 on Build GRCh38
rs748241996 Chr.17: 18976828-18976828 on Build GRCh38
rs754854911 Chr.17: 18976897-18976897 on Build GRCh38
rs199950392 Chr.17: 18976940-18976940 on Build GRCh38
rs761435407 Chr.17: 18976957-18976957 on Build GRCh38
rs569763556 Chr.17: 18977070-18977070 on Build GRCh38
rs183581661 Chr.17: 18977120-18977120 on Build GRCh38
rs113728377 Chr.17: 18976781-18976781 on Build GRCh38
rs773418136 Chr.17: 18976831-18976831 on Build GRCh38
rs992363887 Chr.17: 18977092-18977092 on Build GRCh38
rs373153498 Chr.17: 18977009-18977009 on Build GRCh38
rs755766055 Chr.17: 18976882-18976882 on Build GRCh38
rs764984495 Chr.17: 18977084-18977084 on Build GRCh38
rs939537784 Chr.17: 18976767-18976767 on Build GRCh38
rs143883763 Chr.17: 18976722-18976723 on Build GRCh38
rs927813598 Chr.17: 18977135-18977135 on Build GRCh38
rs987664329 Chr.17: 18976697-18976697 on Build GRCh38
rs112313380 Chr.17: 18977007-18977007 on Build GRCh38
rs746229122 Chr.17: 18976961-18976961 on Build GRCh38

View Full Product Details