Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19343034-19343526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B9D1
Entrez Gene ID
27077
Gene Name
B9 domain containing 1
Gene Aliases
B9, EPPB9, MKS9, MKSR1
UniGene
Hs.309467
Gene Chromosome Location
Chr.17: 19335372-19378193 on Build GRCh38
Transcripts
NM_001243473.2 NM_001243475.2 NM_001321214.1 NM_001321215.1 NM_001321217.1 NM_001321218.1 NM_001321219.1 NM_015681.4 XM_005256607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735183 c.515A>G p.N172S Chr.17: 19343419-19343419 on Build GRCh38
3958090 c.588C>A p.P196P Chr.17: 19343346-19343346 on Build GRCh38
5484078 c.590G>C p.S197T Chr.17: 19343344-19343344 on Build GRCh38
976381 c.544G>A p.G182S Chr.17: 19343390-19343390 on Build GRCh38
297267 c.558G>A p.G186G Chr.17: 19343376-19343376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030339874 Chr.17: 19343082-19343082 on Build GRCh38
rs769710052 Chr.17: 19343300-19343301 on Build GRCh38
rs375701897 Chr.17: 19343252-19343252 on Build GRCh38
rs777925046 Chr.17: 19343281-19343281 on Build GRCh38
rs1036392632 Chr.17: 19343111-19343111 on Build GRCh38
rs926375160 Chr.17: 19343309-19343309 on Build GRCh38
rs549042881 Chr.17: 19343129-19343129 on Build GRCh38
rs773826781 Chr.17: 19343306-19343306 on Build GRCh38
rs771494755 Chr.17: 19343392-19343392 on Build GRCh38
rs997083657 Chr.17: 19343072-19343072 on Build GRCh38
rs200124070 Chr.17: 19343272-19343272 on Build GRCh38
rs752818836 Chr.17: 19343347-19343347 on Build GRCh38
rs915603351 Chr.17: 19343181-19343181 on Build GRCh38
rs886038206 Chr.17: 19343412-19343414 on Build GRCh38
rs764112875 Chr.17: 19343429-19343429 on Build GRCh38
rs776537748 Chr.17: 19343417-19343417 on Build GRCh38
rs1048267763 Chr.17: 19343080-19343080 on Build GRCh38
rs182767811 Chr.17: 19343456-19343456 on Build GRCh38
rs776039881 Chr.17: 19343367-19343367 on Build GRCh38
rs777059613 Chr.17: 19343302-19343302 on Build GRCh38
rs142346405 Chr.17: 19343449-19343449 on Build GRCh38
rs538949071 Chr.17: 19343455-19343455 on Build GRCh38
rs372240519 Chr.17: 19343262-19343262 on Build GRCh38
rs747138868 Chr.17: 19343454-19343454 on Build GRCh38
rs1036359666 Chr.17: 19343406-19343406 on Build GRCh38
rs140933644 Chr.17: 19343131-19343131 on Build GRCh38
rs969720058 Chr.17: 19343375-19343375 on Build GRCh38
rs781361422 Chr.17: 19343146-19343146 on Build GRCh38
rs754944872 Chr.17: 19343338-19343338 on Build GRCh38
rs887041798 Chr.17: 19343098-19343098 on Build GRCh38
rs547869203 Chr.17: 19343331-19343331 on Build GRCh38
rs934506646 Chr.17: 19343248-19343248 on Build GRCh38
rs752593719 Chr.17: 19343426-19343426 on Build GRCh38
rs1001445188 Chr.17: 19343246-19343246 on Build GRCh38
rs779534156 Chr.17: 19343168-19343168 on Build GRCh38
rs199683823 Chr.17: 19343467-19343467 on Build GRCh38
rs748154295 Chr.17: 19343270-19343271 on Build GRCh38
rs981355151 Chr.17: 19343205-19343205 on Build GRCh38
rs766184795 Chr.17: 19343407-19343407 on Build GRCh38
rs939940954 Chr.17: 19343400-19343400 on Build GRCh38
rs371387811 Chr.17: 19343496-19343496 on Build GRCh38
rs1010902726 Chr.17: 19343058-19343058 on Build GRCh38
rs151295524 Chr.17: 19343328-19343328 on Build GRCh38
rs973698037 Chr.17: 19343161-19343161 on Build GRCh38
rs530321513 Chr.17: 19343357-19343357 on Build GRCh38
rs758466581 Chr.17: 19343350-19343350 on Build GRCh38
rs757444799 Chr.17: 19343452-19343452 on Build GRCh38
rs765307040 Chr.17: 19343346-19343346 on Build GRCh38
rs7212549 Chr.17: 19343354-19343354 on Build GRCh38
rs771218470 Chr.17: 19343294-19343294 on Build GRCh38
rs758440762 Chr.17: 19343427-19343427 on Build GRCh38
rs757604145 Chr.17: 19343359-19343359 on Build GRCh38
rs747677339 Chr.17: 19343384-19343384 on Build GRCh38
rs201986754 Chr.17: 19343317-19343317 on Build GRCh38
rs201805359 Chr.17: 19343480-19343480 on Build GRCh38
rs753785719 Chr.17: 19343337-19343337 on Build GRCh38
rs760275062 Chr.17: 19343404-19343404 on Build GRCh38
rs534428777 Chr.17: 19343261-19343261 on Build GRCh38
rs772619551 Chr.17: 19343495-19343495 on Build GRCh38
rs760070960 Chr.17: 19343304-19343304 on Build GRCh38
rs780551918 Chr.17: 19343476-19343476 on Build GRCh38
rs772797594 Chr.17: 19343322-19343322 on Build GRCh38
rs201498591 Chr.17: 19343321-19343321 on Build GRCh38
rs982915821 Chr.17: 19343100-19343100 on Build GRCh38
rs772851974 Chr.17: 19343398-19343398 on Build GRCh38
rs572607311 Chr.17: 19343062-19343062 on Build GRCh38
rs940643115 Chr.17: 19343101-19343101 on Build GRCh38
rs868683895 Chr.17: 19343457-19343457 on Build GRCh38
rs1022331962 Chr.17: 19343059-19343059 on Build GRCh38
rs955532616 Chr.17: 19343097-19343097 on Build GRCh38
rs771568799 Chr.17: 19343492-19343492 on Build GRCh38
rs948410736 Chr.17: 19343193-19343193 on Build GRCh38
rs574693620 Chr.17: 19343211-19343211 on Build GRCh38
rs928552442 Chr.17: 19343229-19343229 on Build GRCh38
rs1015745292 Chr.17: 19343339-19343339 on Build GRCh38
rs147684440 Chr.17: 19343366-19343366 on Build GRCh38
rs781561961 Chr.17: 19343362-19343362 on Build GRCh38
rs149758860 Chr.17: 19343418-19343418 on Build GRCh38
rs150434901 Chr.17: 19343365-19343365 on Build GRCh38
rs765253677 Chr.17: 19343419-19343419 on Build GRCh38
rs770704711 Chr.17: 19343504-19343504 on Build GRCh38
rs779157234 Chr.17: 19343271-19343271 on Build GRCh38
rs962470942 Chr.17: 19343137-19343137 on Build GRCh38
rs997262238 Chr.17: 19343323-19343323 on Build GRCh38
rs980524660 Chr.17: 19343275-19343275 on Build GRCh38
rs552182629 Chr.17: 19343293-19343293 on Build GRCh38
rs957193278 Chr.17: 19343254-19343254 on Build GRCh38

View Full Product Details