Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19347692-19348189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B9D1
Entrez Gene ID
27077
Gene Name
B9 domain containing 1
Gene Aliases
B9, EPPB9, MKS9, MKSR1
UniGene
Hs.309467
Gene Chromosome Location
Chr.17: 19335372-19378193 on Build GRCh38
Transcripts
NM_001243473.2 NM_001243475.2 NM_001321214.1 NM_001321215.1 NM_001321216.1 NM_001321217.1 NM_001321218.1 NM_001321219.1 NM_015681.4 XM_005256607.2 XM_005256610.1 XM_017024452.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
417153 c.302G>A p.R101Q Chr.17: 19347823-19347823 on Build GRCh38
6609490 c.341G>A p.R114Q Chr.17: 19347784-19347784 on Build GRCh38
5952042 c.321C>T p.H107H Chr.17: 19347804-19347804 on Build GRCh38
6609485 c.315C>T p.A105A Chr.17: 19347810-19347810 on Build GRCh38
6609486 c.315C>T p.A105A Chr.17: 19347810-19347810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376515485 Chr.17: 19347911-19347911 on Build GRCh38
rs996417216 Chr.17: 19347849-19347849 on Build GRCh38
rs571518695 Chr.17: 19347728-19347728 on Build GRCh38
rs11869088 Chr.17: 19348063-19348063 on Build GRCh38
rs759444627 Chr.17: 19347870-19347870 on Build GRCh38
rs143149764 Chr.17: 19347782-19347782 on Build GRCh38
rs777500443 Chr.17: 19347860-19347860 on Build GRCh38
rs536966537 Chr.17: 19347923-19347923 on Build GRCh38
rs780676505 Chr.17: 19347846-19347846 on Build GRCh38
rs776662206 Chr.17: 19347810-19347810 on Build GRCh38
rs751788862 Chr.17: 19347839-19347839 on Build GRCh38
rs141244130 Chr.17: 19347858-19347858 on Build GRCh38
rs944381101 Chr.17: 19348154-19348154 on Build GRCh38
rs779979020 Chr.17: 19347897-19347898 on Build GRCh38
rs970227141 Chr.17: 19347975-19347975 on Build GRCh38
rs770853834 Chr.17: 19347809-19347809 on Build GRCh38
rs770757538 Chr.17: 19347867-19347867 on Build GRCh38
rs980279491 Chr.17: 19348004-19348004 on Build GRCh38
rs766736724 Chr.17: 19347916-19347916 on Build GRCh38
rs1027895963 Chr.17: 19348031-19348031 on Build GRCh38
rs376529489 Chr.17: 19347980-19347980 on Build GRCh38
rs1040604454 Chr.17: 19347836-19347836 on Build GRCh38
rs991049495 Chr.17: 19347950-19347950 on Build GRCh38
rs771617247 Chr.17: 19347792-19347792 on Build GRCh38
rs1014306050 Chr.17: 19347934-19347934 on Build GRCh38
rs942362205 Chr.17: 19348097-19348097 on Build GRCh38
rs1053478935 Chr.17: 19347753-19347753 on Build GRCh38
rs746997996 Chr.17: 19348124-19348124 on Build GRCh38
rs752872131 Chr.17: 19347735-19347735 on Build GRCh38
rs774504181 Chr.17: 19347899-19347899 on Build GRCh38
rs146218883 Chr.17: 19347861-19347861 on Build GRCh38
rs766582587 Chr.17: 19348094-19348094 on Build GRCh38
rs773056053 Chr.17: 19347803-19347803 on Build GRCh38
rs575785540 Chr.17: 19348001-19348001 on Build GRCh38
rs907568766 Chr.17: 19348066-19348066 on Build GRCh38
rs752718131 Chr.17: 19347824-19347824 on Build GRCh38
rs144339542 Chr.17: 19347995-19347995 on Build GRCh38
rs763153642 Chr.17: 19347829-19347829 on Build GRCh38
rs778039369 Chr.17: 19347769-19347769 on Build GRCh38
rs997980384 Chr.17: 19347834-19347834 on Build GRCh38
rs746402205 Chr.17: 19347780-19347780 on Build GRCh38
rs764245755 Chr.17: 19347833-19347833 on Build GRCh38
rs997957766 Chr.17: 19348165-19348165 on Build GRCh38
rs34895642 Chr.17: 19348056-19348057 on Build GRCh38
rs764194377 Chr.17: 19347897-19347897 on Build GRCh38
rs778260923 Chr.17: 19347784-19347784 on Build GRCh38
rs755154925 Chr.17: 19347928-19347928 on Build GRCh38
rs753632387 Chr.17: 19348099-19348099 on Build GRCh38
rs775514861 Chr.17: 19347889-19347889 on Build GRCh38
rs759369800 Chr.17: 19347812-19347812 on Build GRCh38
rs747731978 Chr.17: 19347785-19347785 on Build GRCh38
rs751911513 Chr.17: 19347773-19347773 on Build GRCh38
rs377672945 Chr.17: 19347804-19347804 on Build GRCh38
rs758461043 Chr.17: 19347835-19347835 on Build GRCh38
rs370603654 Chr.17: 19347922-19347922 on Build GRCh38
rs952298543 Chr.17: 19348045-19348045 on Build GRCh38
rs923667517 Chr.17: 19348014-19348014 on Build GRCh38
rs762878521 Chr.17: 19347892-19347892 on Build GRCh38
rs930266587 Chr.17: 19347739-19347739 on Build GRCh38
rs758519832 Chr.17: 19347736-19347736 on Build GRCh38
rs1024813357 Chr.17: 19347939-19347939 on Build GRCh38
rs148240978 Chr.17: 19347823-19347823 on Build GRCh38
rs544439306 Chr.17: 19348044-19348044 on Build GRCh38
rs769608300 Chr.17: 19347872-19347872 on Build GRCh38
rs901130039 Chr.17: 19348159-19348161 on Build GRCh38
rs754741861 Chr.17: 19348101-19348101 on Build GRCh38
rs935123325 Chr.17: 19348029-19348029 on Build GRCh38
rs149626690 Chr.17: 19347723-19347723 on Build GRCh38
rs779846554 Chr.17: 19348096-19348097 on Build GRCh38
rs757635419 Chr.17: 19347778-19347778 on Build GRCh38
rs750480029 Chr.17: 19347951-19347951 on Build GRCh38
rs756557597 Chr.17: 19347915-19347915 on Build GRCh38
rs373478202 Chr.17: 19347840-19347840 on Build GRCh38
rs35236507 Chr.17: 19347811-19347812 on Build GRCh38
rs747596672 Chr.17: 19347856-19347856 on Build GRCh38
rs750471365 Chr.17: 19347790-19347792 on Build GRCh38
rs776537604 Chr.17: 19347869-19347869 on Build GRCh38
rs781617046 Chr.17: 19347779-19347779 on Build GRCh38
rs778716707 Chr.17: 19348152-19348152 on Build GRCh38
rs750746499 Chr.17: 19347903-19347903 on Build GRCh38
rs767764163 Chr.17: 19347900-19347900 on Build GRCh38

View Full Product Details