Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 20580098-20580595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT15L2
Entrez Gene ID
256223
Gene Name
CMT1A duplicated region transcript 15-like 2
Gene Aliases
-
UniGene
Hs.131916
Gene Chromosome Location
Chr.17: 20579724-20580911 on Build GRCh38
Transcripts
NM_001190790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5812361 c.325G>A p.A109T Chr.17: 20580208-20580208 on Build GRCh38
5812820 c.326C>A p.A109E Chr.17: 20580209-20580209 on Build GRCh38
1381498 c.647G>A p.G216D Chr.17: 20580530-20580530 on Build GRCh38
6016541 c.574C>T p.Q192* Chr.17: 20580457-20580457 on Build GRCh38
5658250 c.312C>T p.V104V Chr.17: 20580195-20580195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993196308 Chr.17: 20580189-20580189 on Build GRCh38
rs199616807 Chr.17: 20580196-20580196 on Build GRCh38
rs577650693 Chr.17: 20580458-20580458 on Build GRCh38
rs1030037562 Chr.17: 20580410-20580410 on Build GRCh38
rs372505001 Chr.17: 20580233-20580233 on Build GRCh38
rs116532204 Chr.17: 20580505-20580505 on Build GRCh38
rs1053769110 Chr.17: 20580126-20580126 on Build GRCh38
rs546590517 Chr.17: 20580157-20580157 on Build GRCh38
rs888642261 Chr.17: 20580307-20580307 on Build GRCh38
rs760859987 Chr.17: 20580228-20580229 on Build GRCh38
rs761533381 Chr.17: 20580502-20580502 on Build GRCh38
rs993562656 Chr.17: 20580398-20580398 on Build GRCh38
rs1037220972 Chr.17: 20580370-20580370 on Build GRCh38
rs755162537 Chr.17: 20580240-20580240 on Build GRCh38
rs1005780498 Chr.17: 20580164-20580164 on Build GRCh38
rs954520069 Chr.17: 20580195-20580195 on Build GRCh38
rs567175573 Chr.17: 20580241-20580241 on Build GRCh38
rs1007339568 Chr.17: 20580434-20580434 on Build GRCh38
rs941492374 Chr.17: 20580523-20580523 on Build GRCh38
rs560861774 Chr.17: 20580129-20580129 on Build GRCh38
rs201905293 Chr.17: 20580132-20580132 on Build GRCh38
rs765068476 Chr.17: 20580556-20580556 on Build GRCh38
rs778212142 Chr.17: 20580384-20580384 on Build GRCh38
rs966684557 Chr.17: 20580470-20580470 on Build GRCh38
rs556804326 Chr.17: 20580504-20580504 on Build GRCh38
rs550877583 Chr.17: 20580182-20580182 on Build GRCh38
rs763875823 Chr.17: 20580155-20580155 on Build GRCh38
rs538756855 Chr.17: 20580468-20580468 on Build GRCh38
rs11868057 Chr.17: 20580569-20580569 on Build GRCh38
rs768413380 Chr.17: 20580491-20580491 on Build GRCh38
rs1020851455 Chr.17: 20580469-20580469 on Build GRCh38
rs1055329969 Chr.17: 20580162-20580162 on Build GRCh38
rs747422850 Chr.17: 20580442-20580442 on Build GRCh38
rs183902062 Chr.17: 20580134-20580134 on Build GRCh38
rs766343415 Chr.17: 20580530-20580530 on Build GRCh38
rs114609020 Chr.17: 20580163-20580163 on Build GRCh38
rs542420281 Chr.17: 20580522-20580522 on Build GRCh38
rs569461537 Chr.17: 20580188-20580188 on Build GRCh38
rs753892895 Chr.17: 20580558-20580558 on Build GRCh38
rs990834821 Chr.17: 20580515-20580515 on Build GRCh38
rs1005687292 Chr.17: 20580369-20580369 on Build GRCh38
rs938147691 Chr.17: 20580253-20580253 on Build GRCh38
rs897397230 Chr.17: 20580567-20580567 on Build GRCh38
rs536466901 Chr.17: 20580214-20580214 on Build GRCh38
rs771593919 Chr.17: 20580447-20580447 on Build GRCh38
rs1017380315 Chr.17: 20580450-20580450 on Build GRCh38
rs565032829 Chr.17: 20580158-20580158 on Build GRCh38
rs1030460965 Chr.17: 20580192-20580192 on Build GRCh38
rs1055681300 Chr.17: 20580259-20580259 on Build GRCh38
rs553046415 Chr.17: 20580414-20580414 on Build GRCh38
rs1046897230 Chr.17: 20580144-20580144 on Build GRCh38
rs548860276 Chr.17: 20580236-20580236 on Build GRCh38
rs534598169 Chr.17: 20580343-20580343 on Build GRCh38
rs982515807 Chr.17: 20580501-20580501 on Build GRCh38
rs748541911 Chr.17: 20580340-20580340 on Build GRCh38

View Full Product Details