Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 20866306-20866813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC144NL
Entrez Gene ID
339184
Gene Name
coiled-coil domain containing 144 family, N-terminal like
Gene Aliases
-
UniGene
Hs.674830
Gene Chromosome Location
Chr.17: 20847348-20896587 on Build GRCh38
Transcripts
NM_001004306.2 XM_011523818.2 XM_011523819.2 XM_011523821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4552078 c.548G>A p.G183E Chr.17: 20866571-20866571 on Build GRCh38
3932515 c.566C>T p.P189L Chr.17: 20866553-20866553 on Build GRCh38
7359795 c.449G>T p.G150V Chr.17: 20866670-20866670 on Build GRCh38
7325765 c.571+7G>A p.? Chr.17: 20866541-20866541 on Build GRCh38
4129701 c.429G>C p.Q143H Chr.17: 20866690-20866690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142103578 Chr.17: 20866452-20866452 on Build GRCh38
rs369010517 Chr.17: 20866713-20866713 on Build GRCh38
rs759231648 Chr.17: 20866666-20866666 on Build GRCh38
rs377405045 Chr.17: 20866665-20866665 on Build GRCh38
rs918860745 Chr.17: 20866677-20866677 on Build GRCh38
rs779448500 Chr.17: 20866722-20866722 on Build GRCh38
rs1017453280 Chr.17: 20866387-20866387 on Build GRCh38
rs112187499 Chr.17: 20866409-20866409 on Build GRCh38
rs749649006 Chr.17: 20866707-20866707 on Build GRCh38
rs1034844121 Chr.17: 20866771-20866771 on Build GRCh38
rs550849477 Chr.17: 20866541-20866541 on Build GRCh38
rs913715434 Chr.17: 20866468-20866468 on Build GRCh38
rs376930164 Chr.17: 20866604-20866604 on Build GRCh38
rs909883383 Chr.17: 20866658-20866658 on Build GRCh38
rs376698213 Chr.17: 20866585-20866585 on Build GRCh38
rs1021262267 Chr.17: 20866745-20866745 on Build GRCh38
rs755765309 Chr.17: 20866536-20866536 on Build GRCh38
rs577903345 Chr.17: 20866382-20866382 on Build GRCh38
rs113718085 Chr.17: 20866334-20866334 on Build GRCh38
rs763949344 Chr.17: 20866676-20866676 on Build GRCh38
rs1055372472 Chr.17: 20866340-20866340 on Build GRCh38
rs748557108 Chr.17: 20866717-20866717 on Build GRCh38
rs562798700 Chr.17: 20866772-20866772 on Build GRCh38
rs569328064 Chr.17: 20866550-20866550 on Build GRCh38
rs555013237 Chr.17: 20866333-20866333 on Build GRCh38
rs79189629 Chr.17: 20866775-20866775 on Build GRCh38
rs746038158 Chr.17: 20866640-20866640 on Build GRCh38
rs749935115 Chr.17: 20866535-20866535 on Build GRCh38
rs779863997 Chr.17: 20866500-20866500 on Build GRCh38
rs746104584 Chr.17: 20866576-20866576 on Build GRCh38
rs938017642 Chr.17: 20866335-20866335 on Build GRCh38
rs536624039 Chr.17: 20866561-20866561 on Build GRCh38
rs9900791 Chr.17: 20866455-20866455 on Build GRCh38
rs777878006 Chr.17: 20866554-20866554 on Build GRCh38
rs1003747642 Chr.17: 20866767-20866767 on Build GRCh38
rs780315624 Chr.17: 20866704-20866704 on Build GRCh38
rs79930314 Chr.17: 20866586-20866586 on Build GRCh38
rs79843086 Chr.17: 20866583-20866583 on Build GRCh38
rs373533608 Chr.17: 20866718-20866718 on Build GRCh38
rs748236768 Chr.17: 20866507-20866507 on Build GRCh38
rs777628286 Chr.17: 20866628-20866628 on Build GRCh38
rs372322537 Chr.17: 20866563-20866563 on Build GRCh38
rs369730543 Chr.17: 20866601-20866601 on Build GRCh38
rs781342781 Chr.17: 20866639-20866639 on Build GRCh38
rs532709070 Chr.17: 20866763-20866763 on Build GRCh38
rs575673736 Chr.17: 20866460-20866460 on Build GRCh38
rs1017230199 Chr.17: 20866598-20866598 on Build GRCh38
rs982584484 Chr.17: 20866492-20866492 on Build GRCh38
rs752525867 Chr.17: 20866669-20866669 on Build GRCh38
rs74863563 Chr.17: 20866591-20866591 on Build GRCh38
rs756311124 Chr.17: 20866693-20866693 on Build GRCh38
rs565375881 Chr.17: 20866736-20866736 on Build GRCh38
rs545266475 Chr.17: 20866397-20866397 on Build GRCh38
rs76332742 Chr.17: 20866519-20866519 on Build GRCh38
rs77065992 Chr.17: 20866641-20866641 on Build GRCh38
rs765030905 Chr.17: 20866667-20866667 on Build GRCh38
rs1005699164 Chr.17: 20866348-20866348 on Build GRCh38
rs373597919 Chr.17: 20866653-20866653 on Build GRCh38
rs534710244 Chr.17: 20866584-20866584 on Build GRCh38
rs199843054 Chr.17: 20866514-20866514 on Build GRCh38
rs747767771 Chr.17: 20866539-20866539 on Build GRCh38
rs774787674 Chr.17: 20866654-20866654 on Build GRCh38
rs565472810 Chr.17: 20866522-20866522 on Build GRCh38
rs772213164 Chr.17: 20866508-20866508 on Build GRCh38
rs79395366 Chr.17: 20866651-20866651 on Build GRCh38
rs1003267372 Chr.17: 20866753-20866753 on Build GRCh38
rs754692381 Chr.17: 20866594-20866594 on Build GRCh38
rs139970268 Chr.17: 20866647-20866647 on Build GRCh38
rs9906916 Chr.17: 20866592-20866592 on Build GRCh38
rs367548776 Chr.17: 20866520-20866520 on Build GRCh38
rs534476003 Chr.17: 20866367-20866367 on Build GRCh38
rs768837121 Chr.17: 20866709-20866709 on Build GRCh38
rs80298820 Chr.17: 20866689-20866689 on Build GRCh38
rs768617921 Chr.17: 20866502-20866502 on Build GRCh38
rs377494043 Chr.17: 20866728-20866728 on Build GRCh38
rs530317827 Chr.17: 20866774-20866774 on Build GRCh38
rs779052633 Chr.17: 20866506-20866506 on Build GRCh38
rs377422349 Chr.17: 20866607-20866607 on Build GRCh38
rs76947654 Chr.17: 20866690-20866690 on Build GRCh38
rs775906865 Chr.17: 20866525-20866525 on Build GRCh38
rs544528466 Chr.17: 20866770-20866770 on Build GRCh38
rs548612127 Chr.17: 20866782-20866782 on Build GRCh38
rs78361528 Chr.17: 20866729-20866729 on Build GRCh38
rs998395652 Chr.17: 20866557-20866557 on Build GRCh38
rs757213624 Chr.17: 20866635-20866635 on Build GRCh38
rs954366892 Chr.17: 20866379-20866379 on Build GRCh38
rs79433424 Chr.17: 20866549-20866549 on Build GRCh38
rs561258808 Chr.17: 20866719-20866719 on Build GRCh38
rs77472832 Chr.17: 20866377-20866377 on Build GRCh38
rs773446965 Chr.17: 20866512-20866512 on Build GRCh38
rs770083619 Chr.17: 20866577-20866577 on Build GRCh38
rs78792130 Chr.17: 20866655-20866655 on Build GRCh38
rs567288956 Chr.17: 20866564-20866564 on Build GRCh38
rs76435951 Chr.17: 20866532-20866532 on Build GRCh38
rs79678106 Chr.17: 20866553-20866553 on Build GRCh38
rs765999869 Chr.17: 20866537-20866537 on Build GRCh38
rs543165132 Chr.17: 20866501-20866501 on Build GRCh38
rs765082270 Chr.17: 20866597-20866597 on Build GRCh38
rs888712241 Chr.17: 20866341-20866341 on Build GRCh38
rs762903822 Chr.17: 20866670-20866670 on Build GRCh38
rs796156687 Chr.17: 20866373-20866373 on Build GRCh38
rs966670045 Chr.17: 20866758-20866758 on Build GRCh38
rs553120641 Chr.17: 20866374-20866374 on Build GRCh38
rs775857996 Chr.17: 20866582-20866582 on Build GRCh38
rs766968913 Chr.17: 20866534-20866534 on Build GRCh38
rs1055146978 Chr.17: 20866524-20866524 on Build GRCh38
rs1020892974 Chr.17: 20866616-20866616 on Build GRCh38
rs538572804 Chr.17: 20866643-20866643 on Build GRCh38
rs757237708 Chr.17: 20866683-20866683 on Build GRCh38
rs752578734 Chr.17: 20866602-20866602 on Build GRCh38
rs934132588 Chr.17: 20866688-20866688 on Build GRCh38
rs72838133 Chr.17: 20866346-20866346 on Build GRCh38
rs575712239 Chr.17: 20866708-20866708 on Build GRCh38
rs747106199 Chr.17: 20866560-20866560 on Build GRCh38
rs957953345 Chr.17: 20866396-20866396 on Build GRCh38
rs1016185240 Chr.17: 20866365-20866367 on Build GRCh38
rs769502233 Chr.17: 20866668-20866670 on Build GRCh38
rs78132860 Chr.17: 20866682-20866682 on Build GRCh38
rs75406219 Chr.17: 20866489-20866489 on Build GRCh38
rs747103708 Chr.17: 20866633-20866633 on Build GRCh38
rs993650891 Chr.17: 20866368-20866368 on Build GRCh38

View Full Product Details