Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21126836-21127332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7B
Entrez Gene ID
25979
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
CGI-93, SDR32C1
UniGene
Hs.386989
Gene Chromosome Location
Chr.17: 21123369-21191972 on Build GRCh38
Transcripts
NM_015510.4 XM_005256595.4 XM_011523786.2 XM_017024426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
84382 c.15T>A p.A5A Chr.17: 21126986-21126986 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755890301 Chr.17: 21127013-21127013 on Build GRCh38
rs751196487 Chr.17: 21126983-21126983 on Build GRCh38
rs537092573 Chr.17: 21126926-21126926 on Build GRCh38
rs762416571 Chr.17: 21126956-21126956 on Build GRCh38
rs767739473 Chr.17: 21127034-21127034 on Build GRCh38
rs1037198122 Chr.17: 21127209-21127209 on Build GRCh38
rs571131112 Chr.17: 21127040-21127040 on Build GRCh38
rs542860611 Chr.17: 21127186-21127186 on Build GRCh38
rs900628953 Chr.17: 21127226-21127226 on Build GRCh38
rs767055632 Chr.17: 21126946-21126946 on Build GRCh38
rs898122850 Chr.17: 21126867-21126867 on Build GRCh38
rs756278085 Chr.17: 21127062-21127062 on Build GRCh38
rs555428276 Chr.17: 21127125-21127125 on Build GRCh38
rs375926496 Chr.17: 21126931-21126931 on Build GRCh38
rs141302193 Chr.17: 21126904-21126904 on Build GRCh38
rs1022803600 Chr.17: 21127003-21127003 on Build GRCh38
rs939279164 Chr.17: 21127038-21127038 on Build GRCh38
rs940854668 Chr.17: 21127107-21127107 on Build GRCh38
rs996755894 Chr.17: 21127229-21127229 on Build GRCh38
rs370778794 Chr.17: 21127126-21127126 on Build GRCh38
rs1040938591 Chr.17: 21127225-21127225 on Build GRCh38
rs1005260451 Chr.17: 21126868-21126868 on Build GRCh38
rs34961086 Chr.17: 21127260-21127261 on Build GRCh38
rs866914287 Chr.17: 21127192-21127192 on Build GRCh38
rs767872901 Chr.17: 21126976-21126977 on Build GRCh38
rs937627057 Chr.17: 21127075-21127075 on Build GRCh38
rs199536359 Chr.17: 21127000-21127000 on Build GRCh38
rs145048621 Chr.17: 21126923-21126923 on Build GRCh38
rs78159258 Chr.17: 21126986-21126986 on Build GRCh38
rs960611459 Chr.17: 21127286-21127286 on Build GRCh38
rs1009135739 Chr.17: 21127216-21127216 on Build GRCh38
rs999514450 Chr.17: 21127244-21127244 on Build GRCh38
rs1046737307 Chr.17: 21127201-21127201 on Build GRCh38
rs762500541 Chr.17: 21126932-21126932 on Build GRCh38
rs559412412 Chr.17: 21127276-21127276 on Build GRCh38
rs916494229 Chr.17: 21127308-21127308 on Build GRCh38
rs887955305 Chr.17: 21127085-21127085 on Build GRCh38
rs965147589 Chr.17: 21126976-21126976 on Build GRCh38
rs1038091442 Chr.17: 21127112-21127112 on Build GRCh38
rs371187379 Chr.17: 21126969-21126969 on Build GRCh38
rs376837328 Chr.17: 21126963-21126963 on Build GRCh38
rs138073746 Chr.17: 21127137-21127137 on Build GRCh38
rs896932109 Chr.17: 21127212-21127212 on Build GRCh38
rs754379765 Chr.17: 21126955-21126955 on Build GRCh38
rs1019098779 Chr.17: 21126972-21126972 on Build GRCh38
rs886713506 Chr.17: 21126942-21126942 on Build GRCh38
rs992038878 Chr.17: 21127295-21127295 on Build GRCh38
rs780321689 Chr.17: 21127097-21127097 on Build GRCh38
rs1006812946 Chr.17: 21127274-21127274 on Build GRCh38
rs916457861 Chr.17: 21127090-21127090 on Build GRCh38
rs1038442600 Chr.17: 21126860-21126860 on Build GRCh38
rs367960380 Chr.17: 21127064-21127064 on Build GRCh38
rs1049240257 Chr.17: 21126864-21126864 on Build GRCh38
rs1045718225 Chr.17: 21127060-21127060 on Build GRCh38
rs768651501 Chr.17: 21127030-21127030 on Build GRCh38
rs534420841 Chr.17: 21126997-21126997 on Build GRCh38
rs575446014 Chr.17: 21127181-21127181 on Build GRCh38
rs988926049 Chr.17: 21127023-21127023 on Build GRCh38
rs978100573 Chr.17: 21127021-21127021 on Build GRCh38
rs968143397 Chr.17: 21127241-21127241 on Build GRCh38
rs985754110 Chr.17: 21126966-21126966 on Build GRCh38
rs749220845 Chr.17: 21127026-21127026 on Build GRCh38
rs886677653 Chr.17: 21127207-21127207 on Build GRCh38
rs573236914 Chr.17: 21127277-21127277 on Build GRCh38
rs761268515 Chr.17: 21126930-21126930 on Build GRCh38
rs1049738481 Chr.17: 21127076-21127076 on Build GRCh38
rs532722899 Chr.17: 21126891-21126891 on Build GRCh38
rs757812575 Chr.17: 21127017-21127017 on Build GRCh38
rs374076480 Chr.17: 21127014-21127014 on Build GRCh38
rs764853081 Chr.17: 21126933-21126933 on Build GRCh38
rs898254778 Chr.17: 21127169-21127169 on Build GRCh38
rs1030017604 Chr.17: 21126960-21126960 on Build GRCh38
rs554675639 Chr.17: 21127242-21127242 on Build GRCh38
rs750850282 Chr.17: 21127058-21127058 on Build GRCh38
rs1022349267 Chr.17: 21127239-21127239 on Build GRCh38

View Full Product Details