Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21184252-21184787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7B
Entrez Gene ID
25979
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
CGI-93, SDR32C1
UniGene
Hs.386989
Gene Chromosome Location
Chr.17: 21123369-21191972 on Build GRCh38
Transcripts
NM_015510.4 XM_005256595.4 XM_005256596.3 XM_011523786.2 XM_017024425.1 XM_017024426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3819039 c.556C>A p.Q186K Chr.17: 21184400-21184400 on Build GRCh38
7298537 c.584T>A p.I195N Chr.17: 21184428-21184428 on Build GRCh38
7364290 c.619+1G>A p.? Chr.17: 21184464-21184464 on Build GRCh38
4677323 c.610C>T p.R204* Chr.17: 21184454-21184454 on Build GRCh38
2798788 c.571G>A p.A191T Chr.17: 21184415-21184415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910991047 Chr.17: 21184671-21184671 on Build GRCh38
rs1050863995 Chr.17: 21184301-21184301 on Build GRCh38
rs771175775 Chr.17: 21184408-21184408 on Build GRCh38
rs755113065 Chr.17: 21184359-21184359 on Build GRCh38
rs778136873 Chr.17: 21184371-21184371 on Build GRCh38
rs753833880 Chr.17: 21184480-21184480 on Build GRCh38
rs755443912 Chr.17: 21184593-21184593 on Build GRCh38
rs1010201055 Chr.17: 21184387-21184387 on Build GRCh38
rs376243333 Chr.17: 21184746-21184746 on Build GRCh38
rs771774054 Chr.17: 21184446-21184446 on Build GRCh38
rs3744227 Chr.17: 21184302-21184302 on Build GRCh38
rs923596346 Chr.17: 21184715-21184715 on Build GRCh38
rs1026656815 Chr.17: 21184551-21184551 on Build GRCh38
rs541562372 Chr.17: 21184316-21184316 on Build GRCh38
rs529380535 Chr.17: 21184579-21184579 on Build GRCh38
rs1018800899 Chr.17: 21184655-21184655 on Build GRCh38
rs1020703046 Chr.17: 21184478-21184478 on Build GRCh38
rs867518186 Chr.17: 21184622-21184622 on Build GRCh38
rs977753067 Chr.17: 21184710-21184710 on Build GRCh38
rs3744224 Chr.17: 21184675-21184675 on Build GRCh38
rs964525191 Chr.17: 21184662-21184662 on Build GRCh38
rs1057500241 Chr.17: 21184736-21184736 on Build GRCh38
rs555480336 Chr.17: 21184707-21184707 on Build GRCh38
rs899190152 Chr.17: 21184523-21184523 on Build GRCh38
rs139308154 Chr.17: 21184367-21184367 on Build GRCh38
rs201154365 Chr.17: 21184360-21184360 on Build GRCh38
rs781742062 Chr.17: 21184414-21184414 on Build GRCh38
rs946369511 Chr.17: 21184693-21184693 on Build GRCh38
rs776672008 Chr.17: 21184379-21184380 on Build GRCh38
rs766045185 Chr.17: 21184463-21184463 on Build GRCh38
rs896861879 Chr.17: 21184282-21184282 on Build GRCh38
rs555939096 Chr.17: 21184290-21184290 on Build GRCh38
rs182284105 Chr.17: 21184669-21184669 on Build GRCh38
rs779085266 Chr.17: 21184361-21184361 on Build GRCh38
rs189960572 Chr.17: 21184340-21184340 on Build GRCh38
rs766967306 Chr.17: 21184455-21184455 on Build GRCh38
rs115794457 Chr.17: 21184415-21184415 on Build GRCh38
rs561781197 Chr.17: 21184545-21184545 on Build GRCh38
rs767306153 Chr.17: 21184326-21184326 on Build GRCh38
rs370153755 Chr.17: 21184729-21184729 on Build GRCh38
rs1034420712 Chr.17: 21184528-21184528 on Build GRCh38
rs962948954 Chr.17: 21184635-21184635 on Build GRCh38
rs879220173 Chr.17: 21184552-21184552 on Build GRCh38
rs564422597 Chr.17: 21184366-21184366 on Build GRCh38
rs3744225 Chr.17: 21184650-21184650 on Build GRCh38
rs59601337 Chr.17: 21184591-21184591 on Build GRCh38
rs754359691 Chr.17: 21184548-21184548 on Build GRCh38
rs981480145 Chr.17: 21184503-21184503 on Build GRCh38
rs369770695 Chr.17: 21184335-21184335 on Build GRCh38
rs117895288 Chr.17: 21184745-21184745 on Build GRCh38
rs150007102 Chr.17: 21184501-21184501 on Build GRCh38
rs991073329 Chr.17: 21184569-21184569 on Build GRCh38
rs768169268 Chr.17: 21184420-21184420 on Build GRCh38
rs946338525 Chr.17: 21184308-21184308 on Build GRCh38
rs747464710 Chr.17: 21184396-21184396 on Build GRCh38
rs772719945 Chr.17: 21184454-21184454 on Build GRCh38
rs372749594 Chr.17: 21184341-21184341 on Build GRCh38
rs895986083 Chr.17: 21184393-21184393 on Build GRCh38
rs3744226 Chr.17: 21184358-21184358 on Build GRCh38
rs1038781730 Chr.17: 21184300-21184300 on Build GRCh38
rs950695622 Chr.17: 21184603-21184603 on Build GRCh38
rs761311874 Chr.17: 21184444-21184444 on Build GRCh38
rs773784084 Chr.17: 21184426-21184426 on Build GRCh38
rs750580422 Chr.17: 21184315-21184315 on Build GRCh38

View Full Product Details