Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21188625-21189133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7B
Entrez Gene ID
25979
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
CGI-93, SDR32C1
UniGene
Hs.386989
Gene Chromosome Location
Chr.17: 21123369-21191972 on Build GRCh38
Transcripts
NM_015510.4 XM_005256595.4 XM_005256596.3 XM_011523786.2 XM_017024425.1 XM_017024426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5646685 c.691G>C p.E231Q Chr.17: 21188782-21188782 on Build GRCh38
5758000 c.620A>T p.Y207F Chr.17: 21188711-21188711 on Build GRCh38
248768 c.665G>A p.R222H Chr.17: 21188756-21188756 on Build GRCh38
6643355 c.700G>A p.V234I Chr.17: 21188791-21188791 on Build GRCh38
6643354 c.712G>A p.G238S Chr.17: 21188803-21188803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751820251 Chr.17: 21188774-21188774 on Build GRCh38
rs200890935 Chr.17: 21188852-21188852 on Build GRCh38
rs941465537 Chr.17: 21189068-21189068 on Build GRCh38
rs370896452 Chr.17: 21188733-21188733 on Build GRCh38
rs982345425 Chr.17: 21188893-21188893 on Build GRCh38
rs762927919 Chr.17: 21188864-21188864 on Build GRCh38
rs1028650836 Chr.17: 21188912-21188912 on Build GRCh38
rs113030470 Chr.17: 21188669-21188669 on Build GRCh38
rs765409007 Chr.17: 21188662-21188662 on Build GRCh38
rs1048538917 Chr.17: 21188798-21188799 on Build GRCh38
rs746675850 Chr.17: 21188826-21188826 on Build GRCh38
rs991429907 Chr.17: 21188974-21188974 on Build GRCh38
rs763164881 Chr.17: 21188764-21188764 on Build GRCh38
rs752891452 Chr.17: 21188667-21188667 on Build GRCh38
rs918127836 Chr.17: 21189064-21189064 on Build GRCh38
rs538971297 Chr.17: 21188687-21188687 on Build GRCh38
rs748677748 Chr.17: 21188975-21188975 on Build GRCh38
rs764039935 Chr.17: 21188766-21188766 on Build GRCh38
rs780583208 Chr.17: 21188809-21188809 on Build GRCh38
rs557704274 Chr.17: 21188720-21188720 on Build GRCh38
rs756505562 Chr.17: 21188880-21188880 on Build GRCh38
rs756767266 Chr.17: 21188913-21188913 on Build GRCh38
rs775599763 Chr.17: 21188856-21188856 on Build GRCh38
rs754351755 Chr.17: 21188891-21188891 on Build GRCh38
rs779467418 Chr.17: 21188902-21188902 on Build GRCh38
rs376241779 Chr.17: 21188727-21188727 on Build GRCh38
rs373037825 Chr.17: 21188670-21188670 on Build GRCh38
rs750847780 Chr.17: 21188791-21188791 on Build GRCh38
rs776332619 Chr.17: 21188842-21188842 on Build GRCh38
rs1035193769 Chr.17: 21188938-21188938 on Build GRCh38
rs760193362 Chr.17: 21188728-21188728 on Build GRCh38
rs756450457 Chr.17: 21188803-21188803 on Build GRCh38
rs757923245 Chr.17: 21188821-21188821 on Build GRCh38
rs748031839 Chr.17: 21188676-21188679 on Build GRCh38
rs1007099443 Chr.17: 21188820-21188820 on Build GRCh38
rs755369449 Chr.17: 21188897-21188897 on Build GRCh38
rs565974903 Chr.17: 21188654-21188654 on Build GRCh38
rs918605180 Chr.17: 21189102-21189102 on Build GRCh38
rs746556079 Chr.17: 21188903-21188903 on Build GRCh38
rs145432891 Chr.17: 21188843-21188843 on Build GRCh38
rs200124826 Chr.17: 21188674-21188674 on Build GRCh38
rs766684838 Chr.17: 21188890-21188890 on Build GRCh38
rs572869432 Chr.17: 21188950-21188950 on Build GRCh38
rs767684176 Chr.17: 21188873-21188873 on Build GRCh38
rs115830740 Chr.17: 21188844-21188844 on Build GRCh38
rs776219075 Chr.17: 21188748-21188748 on Build GRCh38
rs571452442 Chr.17: 21188678-21188678 on Build GRCh38
rs201946422 Chr.17: 21188725-21188725 on Build GRCh38
rs774363342 Chr.17: 21188872-21188872 on Build GRCh38
rs746391576 Chr.17: 21188685-21188685 on Build GRCh38
rs187236331 Chr.17: 21188871-21188871 on Build GRCh38
rs892910928 Chr.17: 21188845-21188845 on Build GRCh38
rs1050975713 Chr.17: 21188760-21188760 on Build GRCh38
rs780529889 Chr.17: 21188694-21188694 on Build GRCh38
rs781355838 Chr.17: 21188683-21188683 on Build GRCh38
rs1032071863 Chr.17: 21188951-21188951 on Build GRCh38
rs375889358 Chr.17: 21188942-21188942 on Build GRCh38
rs750571861 Chr.17: 21188811-21188811 on Build GRCh38
rs926653400 Chr.17: 21188696-21188696 on Build GRCh38
rs770647008 Chr.17: 21189096-21189096 on Build GRCh38
rs777085078 Chr.17: 21188823-21188823 on Build GRCh38
rs749759123 Chr.17: 21188814-21188814 on Build GRCh38
rs577733383 Chr.17: 21188790-21188790 on Build GRCh38
rs752656764 Chr.17: 21188761-21188761 on Build GRCh38
rs769405006 Chr.17: 21188765-21188766 on Build GRCh38
rs550494048 Chr.17: 21188841-21188841 on Build GRCh38
rs543473052 Chr.17: 21188971-21188971 on Build GRCh38
rs750615122 Chr.17: 21188874-21188874 on Build GRCh38
rs988274522 Chr.17: 21189016-21189016 on Build GRCh38
rs757357586 Chr.17: 21188784-21188784 on Build GRCh38
rs756244631 Chr.17: 21188999-21188999 on Build GRCh38
rs949378115 Chr.17: 21188675-21188675 on Build GRCh38
rs747710075 Chr.17: 21188711-21188711 on Build GRCh38
rs1009986970 Chr.17: 21188865-21188865 on Build GRCh38
rs972827909 Chr.17: 21189080-21189080 on Build GRCh38
rs765323120 Chr.17: 21188756-21188756 on Build GRCh38
rs759397209 Chr.17: 21188755-21188755 on Build GRCh38
rs933925814 Chr.17: 21188707-21188707 on Build GRCh38

View Full Product Details