Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21242521-21243010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NATD1
Entrez Gene ID
256302
Gene Name
N-acetyltransferase domain containing 1
Gene Aliases
C17orf103, Gtlf3b
UniGene
Hs.654713
Gene Chromosome Location
Chr.17: 21238870-21253266 on Build GRCh38
Transcripts
NM_152914.2

SNP Detail

SNP ID Chromosome Location
rs774568693 Chr.17: 21242680-21242680 on Build GRCh38
rs895929256 Chr.17: 21242901-21242901 on Build GRCh38
rs1030818638 Chr.17: 21242699-21242699 on Build GRCh38
rs1018059593 Chr.17: 21242650-21242650 on Build GRCh38
rs992035280 Chr.17: 21242708-21242708 on Build GRCh38
rs1035021957 Chr.17: 21242881-21242881 on Build GRCh38
rs894685563 Chr.17: 21242892-21242892 on Build GRCh38
rs1021659881 Chr.17: 21242629-21242629 on Build GRCh38
rs998945645 Chr.17: 21242662-21242662 on Build GRCh38
rs1000677523 Chr.17: 21242549-21242549 on Build GRCh38
rs566867132 Chr.17: 21242541-21242541 on Build GRCh38
rs959140462 Chr.17: 21242679-21242679 on Build GRCh38
rs1053625401 Chr.17: 21242835-21242835 on Build GRCh38
rs986055440 Chr.17: 21242990-21242990 on Build GRCh38
rs77030980 Chr.17: 21242716-21242716 on Build GRCh38
rs888966921 Chr.17: 21242548-21242548 on Build GRCh38
rs900103981 Chr.17: 21242608-21242608 on Build GRCh38
rs1040346730 Chr.17: 21242608-21242609 on Build GRCh38
rs9906484 Chr.17: 21242717-21242717 on Build GRCh38
rs944264648 Chr.17: 21242600-21242600 on Build GRCh38
rs947668902 Chr.17: 21242837-21242837 on Build GRCh38
rs944190942 Chr.17: 21242781-21242781 on Build GRCh38
rs1046120144 Chr.17: 21242849-21242849 on Build GRCh38
rs564517628 Chr.17: 21242841-21242841 on Build GRCh38
rs1016762268 Chr.17: 21242687-21242687 on Build GRCh38
rs1009276892 Chr.17: 21242571-21242571 on Build GRCh38
rs954719980 Chr.17: 21242974-21242974 on Build GRCh38
rs923258999 Chr.17: 21242721-21242721 on Build GRCh38
rs143699778 Chr.17: 21242956-21242956 on Build GRCh38
rs891312722 Chr.17: 21242574-21242574 on Build GRCh38
rs1023955470 Chr.17: 21242714-21242714 on Build GRCh38
rs986025777 Chr.17: 21242759-21242759 on Build GRCh38
rs201183982 Chr.17: 21242615-21242616 on Build GRCh38
rs61502494 Chr.17: 21242667-21242669 on Build GRCh38
rs747988609 Chr.17: 21242965-21242965 on Build GRCh38
rs903205075 Chr.17: 21242879-21242879 on Build GRCh38
rs372441130 Chr.17: 21242872-21242872 on Build GRCh38
rs189202234 Chr.17: 21242924-21242924 on Build GRCh38
rs906216569 Chr.17: 21242865-21242865 on Build GRCh38
rs1024639490 Chr.17: 21242610-21242610 on Build GRCh38
rs750801575 Chr.17: 21242693-21242693 on Build GRCh38
rs903231328 Chr.17: 21242643-21242643 on Build GRCh38
rs542307844 Chr.17: 21242869-21242869 on Build GRCh38
rs369355573 Chr.17: 21242702-21242703 on Build GRCh38
rs936504962 Chr.17: 21242799-21242799 on Build GRCh38
rs574956602 Chr.17: 21242836-21242836 on Build GRCh38
rs933302751 Chr.17: 21242754-21242754 on Build GRCh38
rs558971703 Chr.17: 21242675-21242675 on Build GRCh38
rs534227917 Chr.17: 21242555-21242555 on Build GRCh38
rs963776496 Chr.17: 21242659-21242659 on Build GRCh38
rs865791060 Chr.17: 21242656-21242656 on Build GRCh38
rs971667900 Chr.17: 21242631-21242631 on Build GRCh38
rs865945882 Chr.17: 21242942-21242942 on Build GRCh38
rs1011986862 Chr.17: 21242626-21242626 on Build GRCh38
rs540236370 Chr.17: 21242945-21242945 on Build GRCh38
rs192063433 Chr.17: 21242941-21242941 on Build GRCh38
rs951111361 Chr.17: 21242771-21242771 on Build GRCh38
rs892177053 Chr.17: 21242560-21242560 on Build GRCh38
rs114030798 Chr.17: 21242776-21242776 on Build GRCh38
rs778577964 Chr.17: 21242673-21242673 on Build GRCh38
rs1053953979 Chr.17: 21242557-21242557 on Build GRCh38
rs979146443 Chr.17: 21242663-21242663 on Build GRCh38

View Full Product Details