Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28534980-28535466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN1
Entrez Gene ID
8456
Gene Name
forkhead box N1
Gene Aliases
FKHL20, RONU, WHN
UniGene
Hs.58611
Gene Chromosome Location
Chr.17: 28506211-28538900 on Build GRCh38
Transcripts
NM_003593.2 XM_005258046.3 XM_011525358.1 XM_011525359.1 XM_011525362.1 XM_011525367.1 XM_011525368.2 XM_011525369.2 XM_011525370.2 XM_017025228.1 XM_017025229.1 XM_017025230.1 XM_017025231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833254 c.1460delC p.Q489fs*61 Chr.17: 28535031-28535031 on Build GRCh38
4971016 c.1424C>T p.P475L Chr.17: 28534995-28534995 on Build GRCh38
2800273 c.1429G>A p.G477R Chr.17: 28535000-28535000 on Build GRCh38
3795371 c.1604C>T p.S535L Chr.17: 28535175-28535175 on Build GRCh38
4399098 c.1500C>T p.S500S Chr.17: 28535071-28535071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780302437 Chr.17: 28535060-28535061 on Build GRCh38
rs911400955 Chr.17: 28535044-28535044 on Build GRCh38
rs559274393 Chr.17: 28535401-28535401 on Build GRCh38
rs1022017967 Chr.17: 28535374-28535374 on Build GRCh38
rs34814444 Chr.17: 28535127-28535127 on Build GRCh38
rs558081551 Chr.17: 28535224-28535224 on Build GRCh38
rs778561319 Chr.17: 28535201-28535201 on Build GRCh38
rs759765753 Chr.17: 28535054-28535054 on Build GRCh38
rs777800751 Chr.17: 28535220-28535220 on Build GRCh38
rs753403074 Chr.17: 28535192-28535192 on Build GRCh38
rs761377489 Chr.17: 28535060-28535060 on Build GRCh38
rs760056095 Chr.17: 28535165-28535165 on Build GRCh38
rs914906056 Chr.17: 28535071-28535071 on Build GRCh38
rs541055674 Chr.17: 28535390-28535390 on Build GRCh38
rs145486134 Chr.17: 28535254-28535254 on Build GRCh38
rs939283306 Chr.17: 28535148-28535148 on Build GRCh38
rs750677729 Chr.17: 28535035-28535065 on Build GRCh38
rs774957376 Chr.17: 28535253-28535253 on Build GRCh38
rs200807422 Chr.17: 28535063-28535063 on Build GRCh38
rs897156140 Chr.17: 28535249-28535249 on Build GRCh38
rs770048065 Chr.17: 28535132-28535132 on Build GRCh38
rs556139208 Chr.17: 28535346-28535346 on Build GRCh38
rs905085296 Chr.17: 28535442-28535442 on Build GRCh38
rs146694795 Chr.17: 28535125-28535125 on Build GRCh38
rs200053524 Chr.17: 28535120-28535120 on Build GRCh38
rs978916727 Chr.17: 28535316-28535316 on Build GRCh38
rs146492596 Chr.17: 28535092-28535092 on Build GRCh38
rs921491072 Chr.17: 28535275-28535275 on Build GRCh38
rs773523562 Chr.17: 28535124-28535124 on Build GRCh38
rs140965751 Chr.17: 28535109-28535109 on Build GRCh38
rs142654668 Chr.17: 28535047-28535047 on Build GRCh38
rs746063530 Chr.17: 28535235-28535235 on Build GRCh38
rs954194706 Chr.17: 28535312-28535312 on Build GRCh38
rs1007569073 Chr.17: 28535270-28535271 on Build GRCh38
rs760121466 Chr.17: 28535258-28535258 on Build GRCh38
rs530723331 Chr.17: 28535006-28535006 on Build GRCh38
rs189886792 Chr.17: 28535118-28535118 on Build GRCh38
rs759981952 Chr.17: 28535130-28535130 on Build GRCh38
rs965653001 Chr.17: 28535034-28535034 on Build GRCh38
rs753110723 Chr.17: 28535058-28535058 on Build GRCh38
rs934663731 Chr.17: 28535102-28535102 on Build GRCh38
rs548817079 Chr.17: 28535014-28535014 on Build GRCh38
rs966451854 Chr.17: 28535381-28535381 on Build GRCh38
rs754872536 Chr.17: 28535196-28535196 on Build GRCh38
rs748095576 Chr.17: 28535114-28535114 on Build GRCh38
rs201850763 Chr.17: 28535043-28535043 on Build GRCh38
rs752486885 Chr.17: 28535204-28535204 on Build GRCh38
rs777081770 Chr.17: 28535053-28535053 on Build GRCh38
rs781540857 Chr.17: 28535228-28535228 on Build GRCh38
rs779911152 Chr.17: 28535005-28535005 on Build GRCh38
rs772251699 Chr.17: 28535042-28535042 on Build GRCh38
rs181483148 Chr.17: 28535189-28535189 on Build GRCh38
rs926104471 Chr.17: 28535344-28535344 on Build GRCh38
rs771004105 Chr.17: 28535051-28535051 on Build GRCh38
rs754708376 Chr.17: 28535106-28535106 on Build GRCh38
rs749877199 Chr.17: 28535065-28535065 on Build GRCh38
rs1046301958 Chr.17: 28535433-28535433 on Build GRCh38
rs143313551 Chr.17: 28535119-28535119 on Build GRCh38
rs755959486 Chr.17: 28535000-28535000 on Build GRCh38
rs778918211 Chr.17: 28535030-28535030 on Build GRCh38
rs917263947 Chr.17: 28535391-28535391 on Build GRCh38
rs749689030 Chr.17: 28535194-28535194 on Build GRCh38
rs748288117 Chr.17: 28535035-28535035 on Build GRCh38
rs927633114 Chr.17: 28535410-28535410 on Build GRCh38
rs977761196 Chr.17: 28535404-28535404 on Build GRCh38
rs554701510 Chr.17: 28535423-28535423 on Build GRCh38
rs1013487643 Chr.17: 28535143-28535143 on Build GRCh38
rs776033591 Chr.17: 28535138-28535138 on Build GRCh38
rs989251101 Chr.17: 28535067-28535068 on Build GRCh38
rs932776196 Chr.17: 28535064-28535064 on Build GRCh38
rs905054182 Chr.17: 28535205-28535205 on Build GRCh38
rs574374662 Chr.17: 28535358-28535358 on Build GRCh38
rs749967360 Chr.17: 28535161-28535161 on Build GRCh38
rs758418847 Chr.17: 28535214-28535214 on Build GRCh38
rs746838285 Chr.17: 28535065-28535065 on Build GRCh38
rs769998823 Chr.17: 28535248-28535248 on Build GRCh38
rs950033664 Chr.17: 28535396-28535396 on Build GRCh38
rs537641290 Chr.17: 28535345-28535345 on Build GRCh38
rs758660910 Chr.17: 28535058-28535060 on Build GRCh38
rs755766998 Chr.17: 28535084-28535084 on Build GRCh38
rs977014020 Chr.17: 28535269-28535269 on Build GRCh38
rs368261937 Chr.17: 28535093-28535093 on Build GRCh38
rs866327967 Chr.17: 28535251-28535251 on Build GRCh38
rs768671496 Chr.17: 28535016-28535016 on Build GRCh38
rs764517468 Chr.17: 28535154-28535154 on Build GRCh38
rs763464553 Chr.17: 28535149-28535149 on Build GRCh38
rs374049334 Chr.17: 28535025-28535025 on Build GRCh38
rs965028696 Chr.17: 28535263-28535263 on Build GRCh38

View Full Product Details