Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28573432-28573942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOC
Entrez Gene ID
230
Gene Name
aldolase, fructose-bisphosphate C
Gene Aliases
ALDC
UniGene
Hs.155247
Gene Chromosome Location
Chr.17: 28573115-28576963 on Build GRCh38
Transcripts
NM_005165.2 XM_005257949.2 XM_011524556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064937 c.801A>G p.G267G Chr.17: 28573933-28573933 on Build GRCh38
6202820 c.1066T>G p.S356A Chr.17: 28573555-28573555 on Build GRCh38
4064935 c.919C>A p.Q307K Chr.17: 28573815-28573815 on Build GRCh38
4064936 c.914C>T p.A305V Chr.17: 28573820-28573820 on Build GRCh38
1182510 c.890C>T p.A297V Chr.17: 28573844-28573844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372509862 Chr.17: 28573917-28573917 on Build GRCh38
rs545952798 Chr.17: 28573634-28573634 on Build GRCh38
rs376690171 Chr.17: 28573824-28573824 on Build GRCh38
rs780510976 Chr.17: 28573640-28573640 on Build GRCh38
rs758865156 Chr.17: 28573803-28573803 on Build GRCh38
rs550214011 Chr.17: 28573781-28573781 on Build GRCh38
rs776348298 Chr.17: 28573552-28573552 on Build GRCh38
rs368456725 Chr.17: 28573650-28573650 on Build GRCh38
rs768592729 Chr.17: 28573851-28573851 on Build GRCh38
rs185997454 Chr.17: 28573658-28573658 on Build GRCh38
rs572172987 Chr.17: 28573546-28573546 on Build GRCh38
rs886811462 Chr.17: 28573607-28573607 on Build GRCh38
rs776095497 Chr.17: 28573885-28573885 on Build GRCh38
rs775106212 Chr.17: 28573579-28573579 on Build GRCh38
rs1016433018 Chr.17: 28573557-28573557 on Build GRCh38
rs867498954 Chr.17: 28573854-28573854 on Build GRCh38
rs374220270 Chr.17: 28573639-28573639 on Build GRCh38
rs145671545 Chr.17: 28573799-28573799 on Build GRCh38
rs774246308 Chr.17: 28573852-28573852 on Build GRCh38
rs757418370 Chr.17: 28573621-28573621 on Build GRCh38
rs530097176 Chr.17: 28573790-28573790 on Build GRCh38
rs767734347 Chr.17: 28573767-28573767 on Build GRCh38
rs150324059 Chr.17: 28573844-28573844 on Build GRCh38
rs765100040 Chr.17: 28573574-28573574 on Build GRCh38
rs781560703 Chr.17: 28573482-28573482 on Build GRCh38
rs745563858 Chr.17: 28573700-28573700 on Build GRCh38
rs1031748302 Chr.17: 28573696-28573696 on Build GRCh38
rs773722392 Chr.17: 28573900-28573900 on Build GRCh38
rs1046810536 Chr.17: 28573606-28573606 on Build GRCh38
rs544206574 Chr.17: 28573701-28573701 on Build GRCh38
rs760663322 Chr.17: 28573880-28573880 on Build GRCh38
rs371687048 Chr.17: 28573782-28573782 on Build GRCh38
rs755344517 Chr.17: 28573791-28573791 on Build GRCh38
rs758954980 Chr.17: 28573916-28573916 on Build GRCh38
rs771878945 Chr.17: 28573864-28573864 on Build GRCh38
rs746010687 Chr.17: 28573488-28573488 on Build GRCh38
rs551895563 Chr.17: 28573485-28573485 on Build GRCh38
rs762986467 Chr.17: 28573580-28573580 on Build GRCh38
rs764100776 Chr.17: 28573613-28573613 on Build GRCh38
rs771206004 Chr.17: 28573500-28573500 on Build GRCh38
rs751747428 Chr.17: 28573616-28573616 on Build GRCh38
rs1041281888 Chr.17: 28573638-28573638 on Build GRCh38
rs761879736 Chr.17: 28573859-28573859 on Build GRCh38
rs756516507 Chr.17: 28573514-28573514 on Build GRCh38
rs759050092 Chr.17: 28573569-28573569 on Build GRCh38
rs1057129469 Chr.17: 28573915-28573915 on Build GRCh38
rs749834508 Chr.17: 28573538-28573538 on Build GRCh38
rs763914905 Chr.17: 28573743-28573743 on Build GRCh38
rs375869123 Chr.17: 28573896-28573896 on Build GRCh38
rs140292923 Chr.17: 28573755-28573755 on Build GRCh38
rs775952908 Chr.17: 28573666-28573666 on Build GRCh38
rs375775620 Chr.17: 28573473-28573473 on Build GRCh38
rs750437044 Chr.17: 28573769-28573769 on Build GRCh38
rs775402885 Chr.17: 28573735-28573735 on Build GRCh38
rs780234569 Chr.17: 28573526-28573526 on Build GRCh38
rs199985843 Chr.17: 28573843-28573843 on Build GRCh38
rs919757372 Chr.17: 28573486-28573486 on Build GRCh38
rs201358818 Chr.17: 28573750-28573750 on Build GRCh38
rs868173932 Chr.17: 28573779-28573779 on Build GRCh38
rs369962449 Chr.17: 28573906-28573906 on Build GRCh38
rs962481616 Chr.17: 28573578-28573578 on Build GRCh38
rs773395204 Chr.17: 28573869-28573869 on Build GRCh38
rs999383072 Chr.17: 28573683-28573683 on Build GRCh38
rs931154185 Chr.17: 28573868-28573868 on Build GRCh38
rs757601204 Chr.17: 28573476-28573476 on Build GRCh38
rs767648346 Chr.17: 28573625-28573625 on Build GRCh38
rs145405642 Chr.17: 28573819-28573819 on Build GRCh38
rs765624567 Chr.17: 28573913-28573913 on Build GRCh38
rs748583504 Chr.17: 28573659-28573659 on Build GRCh38
rs748579404 Chr.17: 28573603-28573603 on Build GRCh38
rs769299089 Chr.17: 28573702-28573702 on Build GRCh38
rs769124297 Chr.17: 28573544-28573544 on Build GRCh38
rs912511231 Chr.17: 28573776-28573776 on Build GRCh38
rs748891048 Chr.17: 28573850-28573850 on Build GRCh38
rs770437455 Chr.17: 28573664-28573664 on Build GRCh38
rs41280130 Chr.17: 28573635-28573635 on Build GRCh38
rs754077392 Chr.17: 28573902-28573902 on Build GRCh38
rs762828594 Chr.17: 28573742-28573742 on Build GRCh38
rs151291013 Chr.17: 28573890-28573890 on Build GRCh38
rs770077311 Chr.17: 28573711-28573713 on Build GRCh38

View Full Product Details