Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43493367-43493866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX8
Entrez Gene ID
1659
Gene Name
DEAH-box helicase 8
Gene Aliases
DDX8, HRH1, PRP22, PRPF22
UniGene
Hs.463105
Gene Chromosome Location
Chr.17: 43483865-43544787 on Build GRCh38
Transcripts
NM_001302623.1 NM_001322216.1 NM_001322217.1 NM_001322218.1 NM_001322219.1 NM_001322220.1 NM_001322221.1 NM_004941.2 NR_136225.1 NR_136226.1 NR_136227.1 NR_136228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3819709 c.871T>G p.W291G Chr.17: 43493452-43493452 on Build GRCh38
1563819 c.1051C>T p.R351W Chr.17: 43493725-43493725 on Build GRCh38
5342938 c.1149C>T p.D383D Chr.17: 43493823-43493823 on Build GRCh38
3787344 c.873G>A p.W291* Chr.17: 43493454-43493454 on Build GRCh38
3772930 c.1008+9G>A p.? Chr.17: 43493598-43493598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771713143 Chr.17: 43493394-43493394 on Build GRCh38
rs768286256 Chr.17: 43493670-43493670 on Build GRCh38
rs989501895 Chr.17: 43493398-43493398 on Build GRCh38
rs772793540 Chr.17: 43493824-43493824 on Build GRCh38
rs762908963 Chr.17: 43493438-43493438 on Build GRCh38
rs773212246 Chr.17: 43493420-43493420 on Build GRCh38
rs754400269 Chr.17: 43493488-43493488 on Build GRCh38
rs751733915 Chr.17: 43493751-43493751 on Build GRCh38
rs745755239 Chr.17: 43493649-43493649 on Build GRCh38
rs754808714 Chr.17: 43493619-43493620 on Build GRCh38
rs146528228 Chr.17: 43493451-43493451 on Build GRCh38
rs561424381 Chr.17: 43493842-43493842 on Build GRCh38
rs745833646 Chr.17: 43493529-43493529 on Build GRCh38
rs944969256 Chr.17: 43493585-43493585 on Build GRCh38
rs780990163 Chr.17: 43493629-43493629 on Build GRCh38
rs1005917773 Chr.17: 43493757-43493757 on Build GRCh38
rs368892637 Chr.17: 43493616-43493616 on Build GRCh38
rs757967298 Chr.17: 43493660-43493660 on Build GRCh38
rs765326952 Chr.17: 43493405-43493406 on Build GRCh38
rs887505846 Chr.17: 43493728-43493728 on Build GRCh38
rs767735314 Chr.17: 43493453-43493453 on Build GRCh38
rs776488618 Chr.17: 43493528-43493528 on Build GRCh38
rs751608922 Chr.17: 43493843-43493843 on Build GRCh38
rs564778404 Chr.17: 43493413-43493413 on Build GRCh38
rs751342267 Chr.17: 43493536-43493541 on Build GRCh38
rs373087636 Chr.17: 43493617-43493617 on Build GRCh38
rs377572676 Chr.17: 43493823-43493823 on Build GRCh38
rs770829567 Chr.17: 43493405-43493405 on Build GRCh38
rs34610973 Chr.17: 43493517-43493518 on Build GRCh38
rs770965497 Chr.17: 43493417-43493417 on Build GRCh38
rs766233524 Chr.17: 43493487-43493488 on Build GRCh38
rs777020341 Chr.17: 43493725-43493725 on Build GRCh38
rs746843678 Chr.17: 43493400-43493400 on Build GRCh38
rs778595067 Chr.17: 43493797-43493797 on Build GRCh38
rs750212053 Chr.17: 43493775-43493775 on Build GRCh38
rs774205130 Chr.17: 43493573-43493573 on Build GRCh38
rs765756648 Chr.17: 43493737-43493737 on Build GRCh38
rs750785920 Chr.17: 43493459-43493459 on Build GRCh38
rs762412448 Chr.17: 43493534-43493534 on Build GRCh38
rs769139935 Chr.17: 43493531-43493531 on Build GRCh38
rs774951011 Chr.17: 43493450-43493450 on Build GRCh38
rs150414727 Chr.17: 43493685-43493685 on Build GRCh38
rs966794123 Chr.17: 43493403-43493403 on Build GRCh38
rs775885889 Chr.17: 43493793-43493793 on Build GRCh38
rs766988080 Chr.17: 43493486-43493486 on Build GRCh38
rs762040259 Chr.17: 43493607-43493607 on Build GRCh38
rs764176217 Chr.17: 43493745-43493745 on Build GRCh38
rs774088630 Chr.17: 43493709-43493709 on Build GRCh38
rs759864291 Chr.17: 43493726-43493726 on Build GRCh38
rs761053649 Chr.17: 43493583-43493583 on Build GRCh38
rs756596077 Chr.17: 43493475-43493475 on Build GRCh38
rs368421084 Chr.17: 43493522-43493522 on Build GRCh38
rs763476779 Chr.17: 43493837-43493837 on Build GRCh38
rs764811072 Chr.17: 43493435-43493435 on Build GRCh38
rs746790573 Chr.17: 43493517-43493517 on Build GRCh38
rs375106079 Chr.17: 43493624-43493624 on Build GRCh38
rs763218908 Chr.17: 43493836-43493836 on Build GRCh38
rs755537171 Chr.17: 43493608-43493608 on Build GRCh38
rs768231533 Chr.17: 43493795-43493795 on Build GRCh38
rs780037875 Chr.17: 43493790-43493790 on Build GRCh38
rs776418373 Chr.17: 43493449-43493449 on Build GRCh38
rs377630686 Chr.17: 43493739-43493739 on Build GRCh38
rs370058622 Chr.17: 43493830-43493830 on Build GRCh38
rs755921581 Chr.17: 43493784-43493784 on Build GRCh38
rs140435622 Chr.17: 43493504-43493504 on Build GRCh38
rs747698603 Chr.17: 43493803-43493803 on Build GRCh38
rs769741522 Chr.17: 43493658-43493658 on Build GRCh38
rs1014589387 Chr.17: 43493772-43493772 on Build GRCh38
rs148880713 Chr.17: 43493754-43493754 on Build GRCh38
rs922206110 Chr.17: 43493622-43493622 on Build GRCh38
rs145746385 Chr.17: 43493553-43493553 on Build GRCh38
rs779070752 Chr.17: 43493500-43493500 on Build GRCh38
rs754952693 Chr.17: 43493489-43493489 on Build GRCh38
rs534370986 Chr.17: 43493613-43493613 on Build GRCh38
rs776611790 Chr.17: 43493409-43493409 on Build GRCh38
rs771381032 Chr.17: 43493718-43493718 on Build GRCh38
rs529814588 Chr.17: 43493712-43493712 on Build GRCh38
rs748275231 Chr.17: 43493503-43493503 on Build GRCh38
rs748819777 Chr.17: 43493664-43493664 on Build GRCh38
rs773018882 Chr.17: 43493396-43493396 on Build GRCh38
rs373563314 Chr.17: 43493786-43493786 on Build GRCh38
rs775097707 Chr.17: 43493532-43493532 on Build GRCh38
rs202226933 Chr.17: 43493740-43493740 on Build GRCh38
rs371554627 Chr.17: 43493599-43493599 on Build GRCh38

View Full Product Details