Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44171906-44172401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB16
Entrez Gene ID
92591
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
UniGene
Hs.534517
Gene Chromosome Location
Chr.17: 44170706-44179083 on Build GRCh38
Transcripts
NM_080863.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
472875 c.567G>A p.L189L Chr.17: 44172311-44172311 on Build GRCh38
3190711 c.441G>A p.L147L Chr.17: 44172185-44172185 on Build GRCh38
304363 c.559G>A p.E187K Chr.17: 44172303-44172303 on Build GRCh38
1183727 c.383G>A p.R128Q Chr.17: 44172127-44172127 on Build GRCh38
472874 c.443A>G p.H148R Chr.17: 44172187-44172187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779619372 Chr.17: 44172038-44172038 on Build GRCh38
rs924216340 Chr.17: 44172369-44172369 on Build GRCh38
rs370322072 Chr.17: 44172339-44172339 on Build GRCh38
rs151040840 Chr.17: 44172262-44172262 on Build GRCh38
rs752303230 Chr.17: 44172177-44172177 on Build GRCh38
rs200617617 Chr.17: 44172122-44172122 on Build GRCh38
rs866630358 Chr.17: 44172112-44172113 on Build GRCh38
rs371177984 Chr.17: 44172283-44172283 on Build GRCh38
rs755720972 Chr.17: 44172016-44172016 on Build GRCh38
rs773956658 Chr.17: 44172360-44172360 on Build GRCh38
rs766616021 Chr.17: 44172120-44172121 on Build GRCh38
rs907614410 Chr.17: 44171955-44171955 on Build GRCh38
rs386386144 Chr.17: 44172359-44172360 on Build GRCh38
rs544754486 Chr.17: 44172048-44172048 on Build GRCh38
rs531721947 Chr.17: 44172168-44172168 on Build GRCh38
rs928247495 Chr.17: 44171992-44171996 on Build GRCh38
rs562750398 Chr.17: 44172084-44172084 on Build GRCh38
rs972760460 Chr.17: 44172363-44172363 on Build GRCh38
rs139711178 Chr.17: 44172296-44172296 on Build GRCh38
rs554677112 Chr.17: 44171934-44171934 on Build GRCh38
rs780053792 Chr.17: 44172060-44172060 on Build GRCh38
rs749205673 Chr.17: 44172095-44172095 on Build GRCh38
rs561401977 Chr.17: 44172141-44172141 on Build GRCh38
rs144187817 Chr.17: 44172240-44172240 on Build GRCh38
rs772379935 Chr.17: 44172050-44172050 on Build GRCh38
rs770992986 Chr.17: 44172357-44172358 on Build GRCh38
rs34112234 Chr.17: 44172350-44172351 on Build GRCh38
rs146107168 Chr.17: 44172274-44172274 on Build GRCh38
rs927973298 Chr.17: 44172378-44172378 on Build GRCh38
rs373603799 Chr.17: 44172167-44172167 on Build GRCh38
rs144121405 Chr.17: 44172193-44172193 on Build GRCh38
rs928383301 Chr.17: 44172171-44172171 on Build GRCh38
rs771864972 Chr.17: 44172235-44172235 on Build GRCh38
rs376723178 Chr.17: 44172163-44172163 on Build GRCh38
rs747471011 Chr.17: 44172126-44172126 on Build GRCh38
rs747443541 Chr.17: 44172062-44172062 on Build GRCh38
rs536750671 Chr.17: 44172336-44172336 on Build GRCh38
rs759589978 Chr.17: 44172074-44172074 on Build GRCh38
rs370313213 Chr.17: 44172267-44172267 on Build GRCh38
rs367726107 Chr.17: 44172124-44172124 on Build GRCh38
rs185813372 Chr.17: 44172113-44172113 on Build GRCh38
rs201134747 Chr.17: 44172012-44172012 on Build GRCh38
rs796147740 Chr.17: 44172350-44172351 on Build GRCh38
rs755099721 Chr.17: 44172299-44172299 on Build GRCh38
rs750009699 Chr.17: 44172015-44172015 on Build GRCh38
rs545339146 Chr.17: 44172098-44172098 on Build GRCh38
rs1043941140 Chr.17: 44172080-44172080 on Build GRCh38
rs763298753 Chr.17: 44172083-44172083 on Build GRCh38
rs1035421746 Chr.17: 44172305-44172305 on Build GRCh38
rs149793071 Chr.17: 44172303-44172303 on Build GRCh38
rs370934626 Chr.17: 44172361-44172362 on Build GRCh38
rs377336077 Chr.17: 44172239-44172239 on Build GRCh38
rs758985812 Chr.17: 44172259-44172259 on Build GRCh38
rs144420642 Chr.17: 44172254-44172254 on Build GRCh38
rs759496969 Chr.17: 44172322-44172322 on Build GRCh38
rs749196106 Chr.17: 44172352-44172353 on Build GRCh38
rs369102293 Chr.17: 44172176-44172176 on Build GRCh38
rs865901269 Chr.17: 44172187-44172187 on Build GRCh38
rs397733526 Chr.17: 44172362-44172363 on Build GRCh38
rs368804726 Chr.17: 44172096-44172096 on Build GRCh38
rs763718741 Chr.17: 44172365-44172365 on Build GRCh38
rs746354714 Chr.17: 44172184-44172184 on Build GRCh38
rs766363799 Chr.17: 44172248-44172248 on Build GRCh38
rs1011077431 Chr.17: 44172104-44172104 on Build GRCh38
rs984664274 Chr.17: 44171948-44171948 on Build GRCh38
rs144512289 Chr.17: 44172152-44172152 on Build GRCh38
rs771202055 Chr.17: 44172127-44172127 on Build GRCh38
rs751418691 Chr.17: 44172089-44172089 on Build GRCh38
rs377473273 Chr.17: 44172269-44172269 on Build GRCh38
rs765847586 Chr.17: 44172173-44172173 on Build GRCh38
rs996161207 Chr.17: 44172121-44172121 on Build GRCh38
rs776008227 Chr.17: 44172351-44172352 on Build GRCh38
rs775541665 Chr.17: 44172079-44172079 on Build GRCh38
rs139281250 Chr.17: 44172174-44172174 on Build GRCh38
rs763998884 Chr.17: 44172008-44172008 on Build GRCh38
rs375065258 Chr.17: 44172321-44172321 on Build GRCh38
rs780266713 Chr.17: 44172201-44172201 on Build GRCh38
rs771519704 Chr.17: 44172065-44172065 on Build GRCh38
rs981606698 Chr.17: 44171990-44171990 on Build GRCh38
rs770004272 Chr.17: 44172075-44172075 on Build GRCh38
rs368394609 Chr.17: 44172331-44172331 on Build GRCh38
rs576045497 Chr.17: 44172302-44172302 on Build GRCh38
rs746876419 Chr.17: 44172316-44172316 on Build GRCh38
rs781441603 Chr.17: 44172180-44172180 on Build GRCh38
rs961433896 Chr.17: 44172353-44172353 on Build GRCh38
rs36084440 Chr.17: 44172222-44172222 on Build GRCh38
rs902256386 Chr.17: 44172223-44172223 on Build GRCh38
rs375638701 Chr.17: 44172175-44172175 on Build GRCh38
rs781679693 Chr.17: 44172131-44172131 on Build GRCh38
rs372078758 Chr.17: 44172140-44172140 on Build GRCh38
rs370024687 Chr.17: 44172166-44172166 on Build GRCh38
rs774013954 Chr.17: 44172162-44172162 on Build GRCh38
rs751959190 Chr.17: 44172212-44172215 on Build GRCh38
rs780543313 Chr.17: 44172014-44172014 on Build GRCh38
rs150509579 Chr.17: 44172090-44172090 on Build GRCh38
rs758929174 Chr.17: 44172030-44172031 on Build GRCh38
rs370302857 Chr.17: 44172106-44172106 on Build GRCh38
rs780643047 Chr.17: 44172275-44172275 on Build GRCh38
rs902204482 Chr.17: 44172115-44172115 on Build GRCh38
rs140204503 Chr.17: 44172282-44172282 on Build GRCh38
rs778890473 Chr.17: 44172295-44172295 on Build GRCh38
rs757589524 Chr.17: 44172179-44172179 on Build GRCh38
rs112609276 Chr.17: 44172139-44172139 on Build GRCh38
rs200305662 Chr.17: 44172105-44172105 on Build GRCh38
rs961855179 Chr.17: 44171960-44171960 on Build GRCh38
rs770723569 Chr.17: 44172319-44172319 on Build GRCh38
rs767900757 Chr.17: 44172272-44172272 on Build GRCh38
rs778143490 Chr.17: 44172061-44172061 on Build GRCh38
rs146027914 Chr.17: 44172202-44172202 on Build GRCh38
rs368821678 Chr.17: 44172004-44172004 on Build GRCh38
rs751499509 Chr.17: 44172011-44172011 on Build GRCh38
rs767559010 Chr.17: 44172092-44172092 on Build GRCh38
rs532057996 Chr.17: 44172301-44172301 on Build GRCh38
rs916865184 Chr.17: 44172037-44172037 on Build GRCh38
rs755037375 Chr.17: 44172289-44172289 on Build GRCh38
rs10628218 Chr.17: 44172349-44172350 on Build GRCh38
rs749497221 Chr.17: 44172148-44172148 on Build GRCh38
rs138426570 Chr.17: 44172138-44172138 on Build GRCh38
rs575093403 Chr.17: 44172172-44172172 on Build GRCh38

View Full Product Details