Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44192097-44192600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L3
Entrez Gene ID
56970
Gene Name
ataxin 7 like 3
Gene Aliases
-
UniGene
Hs.512651
Gene Chromosome Location
Chr.17: 44191805-44199976 on Build GRCh38
Transcripts
NM_001098833.1 NM_020218.1 XM_011525038.2 XM_011525044.2 XM_011525046.2 XM_017024880.1 XM_017024881.1 XM_017024882.1 XM_017024883.1 XM_017024884.1 XM_017024885.1 XM_017024886.1

SNP Detail

SNP ID Chromosome Location
rs1027603961 Chr.17: 44192165-44192165 on Build GRCh38
rs532272673 Chr.17: 44192311-44192311 on Build GRCh38
rs557974550 Chr.17: 44192567-44192567 on Build GRCh38
rs61416700 Chr.17: 44192204-44192205 on Build GRCh38
rs371512397 Chr.17: 44192517-44192517 on Build GRCh38
rs1039163417 Chr.17: 44192289-44192289 on Build GRCh38
rs900711641 Chr.17: 44192304-44192304 on Build GRCh38
rs989934750 Chr.17: 44192215-44192215 on Build GRCh38
rs912941437 Chr.17: 44192393-44192393 on Build GRCh38
rs1001481539 Chr.17: 44192351-44192351 on Build GRCh38
rs957096219 Chr.17: 44192213-44192213 on Build GRCh38
rs184335383 Chr.17: 44192392-44192392 on Build GRCh38
rs574066625 Chr.17: 44192157-44192157 on Build GRCh38
rs912105187 Chr.17: 44192286-44192286 on Build GRCh38
rs573188286 Chr.17: 44192569-44192569 on Build GRCh38
rs35675776 Chr.17: 44192130-44192131 on Build GRCh38
rs994908153 Chr.17: 44192162-44192162 on Build GRCh38
rs781645164 Chr.17: 44192160-44192160 on Build GRCh38
rs950815976 Chr.17: 44192176-44192176 on Build GRCh38
rs867262904 Chr.17: 44192497-44192497 on Build GRCh38
rs113669068 Chr.17: 44192212-44192212 on Build GRCh38
rs73316162 Chr.17: 44192319-44192319 on Build GRCh38
rs532950980 Chr.17: 44192255-44192255 on Build GRCh38
rs986852756 Chr.17: 44192419-44192420 on Build GRCh38
rs1024607765 Chr.17: 44192185-44192185 on Build GRCh38
rs1009449684 Chr.17: 44192125-44192125 on Build GRCh38
rs912255913 Chr.17: 44192423-44192423 on Build GRCh38
rs912897354 Chr.17: 44192229-44192229 on Build GRCh38
rs752647180 Chr.17: 44192502-44192502 on Build GRCh38
rs527632136 Chr.17: 44192383-44192383 on Build GRCh38
rs971703287 Chr.17: 44192342-44192342 on Build GRCh38
rs983472852 Chr.17: 44192181-44192182 on Build GRCh38
rs944950311 Chr.17: 44192458-44192458 on Build GRCh38
rs544410053 Chr.17: 44192161-44192161 on Build GRCh38
rs1024329041 Chr.17: 44192339-44192339 on Build GRCh38
rs75441414 Chr.17: 44192204-44192204 on Build GRCh38
rs561784163 Chr.17: 44192143-44192143 on Build GRCh38
rs967171207 Chr.17: 44192406-44192406 on Build GRCh38
rs188509705 Chr.17: 44192563-44192563 on Build GRCh38
rs1005264767 Chr.17: 44192330-44192330 on Build GRCh38
rs930777374 Chr.17: 44192507-44192507 on Build GRCh38
rs886500586 Chr.17: 44192556-44192556 on Build GRCh38
rs148149055 Chr.17: 44192247-44192247 on Build GRCh38
rs146073477 Chr.17: 44192249-44192249 on Build GRCh38
rs537868466 Chr.17: 44192499-44192499 on Build GRCh38
rs571326815 Chr.17: 44192525-44192525 on Build GRCh38
rs551611835 Chr.17: 44192279-44192279 on Build GRCh38
rs753363672 Chr.17: 44192318-44192318 on Build GRCh38
rs397737399 Chr.17: 44192211-44192212 on Build GRCh38
rs549575897 Chr.17: 44192522-44192522 on Build GRCh38
rs979909034 Chr.17: 44192205-44192206 on Build GRCh38
rs765954052 Chr.17: 44192313-44192313 on Build GRCh38
rs1034179367 Chr.17: 44192386-44192386 on Build GRCh38
rs538700690 Chr.17: 44192157-44192160 on Build GRCh38
rs1046132750 Chr.17: 44192570-44192570 on Build GRCh38

View Full Product Details