Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44801398-44801943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJC1
Entrez Gene ID
10052
Gene Name
gap junction protein gamma 1
Gene Aliases
CX45, GJA7
UniGene
Hs.712052
Gene Chromosome Location
Chr.17: 44794988-44830811 on Build GRCh38
Transcripts
NM_001080383.1 NM_005497.3 XM_005256920.1 XM_005256921.1

SNP Detail

SNP ID Chromosome Location
rs930150023 Chr.17: 44801461-44801461 on Build GRCh38
rs865898131 Chr.17: 44801749-44801749 on Build GRCh38
rs1010052487 Chr.17: 44801600-44801603 on Build GRCh38
rs531253492 Chr.17: 44801729-44801729 on Build GRCh38
rs1018304954 Chr.17: 44801902-44801902 on Build GRCh38
rs1139342 Chr.17: 44801878-44801878 on Build GRCh38
rs1042298519 Chr.17: 44801887-44801887 on Build GRCh38
rs1029903593 Chr.17: 44801608-44801620 on Build GRCh38
rs1055954965 Chr.17: 44801577-44801577 on Build GRCh38
rs183374593 Chr.17: 44801891-44801891 on Build GRCh38
rs568319314 Chr.17: 44801626-44801627 on Build GRCh38
rs1018188979 Chr.17: 44801608-44801612 on Build GRCh38
rs1026766880 Chr.17: 44801697-44801697 on Build GRCh38
rs893388961 Chr.17: 44801865-44801865 on Build GRCh38
rs570431953 Chr.17: 44801848-44801848 on Build GRCh38
rs1037344608 Chr.17: 44801491-44801492 on Build GRCh38
rs116155682 Chr.17: 44801657-44801657 on Build GRCh38
rs767828592 Chr.17: 44801474-44801474 on Build GRCh38
rs146159476 Chr.17: 44801667-44801667 on Build GRCh38
rs140163246 Chr.17: 44801917-44801917 on Build GRCh38
rs903270715 Chr.17: 44801630-44801630 on Build GRCh38
rs963340667 Chr.17: 44801654-44801654 on Build GRCh38
rs1056095985 Chr.17: 44801852-44801852 on Build GRCh38
rs893345725 Chr.17: 44801604-44801604 on Build GRCh38
rs907426557 Chr.17: 44801528-44801528 on Build GRCh38
rs977031231 Chr.17: 44801429-44801429 on Build GRCh38
rs1062354 Chr.17: 44801900-44801900 on Build GRCh38
rs1012068661 Chr.17: 44801606-44801606 on Build GRCh38
rs947542532 Chr.17: 44801875-44801875 on Build GRCh38
rs1020335411 Chr.17: 44801614-44801614 on Build GRCh38
rs1019568746 Chr.17: 44801895-44801895 on Build GRCh38
rs941726791 Chr.17: 44801648-44801648 on Build GRCh38
rs558582760 Chr.17: 44801897-44801897 on Build GRCh38
rs895708817 Chr.17: 44801756-44801756 on Build GRCh38
rs756617347 Chr.17: 44801770-44801770 on Build GRCh38
rs1057047270 Chr.17: 44801598-44801598 on Build GRCh38
rs566348485 Chr.17: 44801664-44801664 on Build GRCh38
rs910066136 Chr.17: 44801641-44801641 on Build GRCh38
rs937574455 Chr.17: 44801851-44801851 on Build GRCh38
rs777291440 Chr.17: 44801600-44801600 on Build GRCh38
rs926300508 Chr.17: 44801845-44801845 on Build GRCh38
rs949040970 Chr.17: 44801444-44801444 on Build GRCh38
rs747641131 Chr.17: 44801591-44801594 on Build GRCh38
rs775357523 Chr.17: 44801608-44801608 on Build GRCh38
rs539188622 Chr.17: 44801633-44801633 on Build GRCh38
rs905535794 Chr.17: 44801543-44801544 on Build GRCh38
rs780639885 Chr.17: 44801894-44801894 on Build GRCh38
rs1047709376 Chr.17: 44801471-44801471 on Build GRCh38
rs908262096 Chr.17: 44801736-44801736 on Build GRCh38
rs571865684 Chr.17: 44801442-44801442 on Build GRCh38
rs903253484 Chr.17: 44801889-44801889 on Build GRCh38
rs577866530 Chr.17: 44801608-44801615 on Build GRCh38
rs750917836 Chr.17: 44801717-44801717 on Build GRCh38
rs996810769 Chr.17: 44801899-44801899 on Build GRCh38
rs964074015 Chr.17: 44801608-44801614 on Build GRCh38
rs554933789 Chr.17: 44801722-44801722 on Build GRCh38
rs1000244306 Chr.17: 44801608-44801611 on Build GRCh38
rs770522166 Chr.17: 44801438-44801439 on Build GRCh38
rs771222530 Chr.17: 44801828-44801828 on Build GRCh38
rs937607039 Chr.17: 44801536-44801536 on Build GRCh38
rs945966914 Chr.17: 44801810-44801810 on Build GRCh38
rs982586850 Chr.17: 44801430-44801430 on Build GRCh38

View Full Product Details