Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44802606-44803135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJC1
Entrez Gene ID
10052
Gene Name
gap junction protein gamma 1
Gene Aliases
CX45, GJA7
UniGene
Hs.712052
Gene Chromosome Location
Chr.17: 44794988-44830811 on Build GRCh38
Transcripts
NM_001080383.1 NM_005497.3 XM_005256920.1 XM_005256921.1

SNP Detail

SNP ID Chromosome Location
rs775899426 Chr.17: 44802693-44802696 on Build GRCh38
rs896761840 Chr.17: 44803035-44803035 on Build GRCh38
rs530186424 Chr.17: 44802916-44802916 on Build GRCh38
rs925636961 Chr.17: 44802827-44802827 on Build GRCh38
rs867346457 Chr.17: 44802812-44802812 on Build GRCh38
rs76725776 Chr.17: 44802659-44802659 on Build GRCh38
rs575205985 Chr.17: 44802660-44802660 on Build GRCh38
rs933810993 Chr.17: 44802864-44802864 on Build GRCh38
rs57058959 Chr.17: 44803011-44803011 on Build GRCh38
rs1051613099 Chr.17: 44802635-44802635 on Build GRCh38
rs551337662 Chr.17: 44803050-44803066 on Build GRCh38
rs192740113 Chr.17: 44802792-44802792 on Build GRCh38
rs938032693 Chr.17: 44803025-44803025 on Build GRCh38
rs967122926 Chr.17: 44802804-44802804 on Build GRCh38
rs767890657 Chr.17: 44803053-44803053 on Build GRCh38
rs984647331 Chr.17: 44802849-44802849 on Build GRCh38
rs1050973231 Chr.17: 44802874-44802874 on Build GRCh38
rs976874556 Chr.17: 44802831-44802831 on Build GRCh38
rs965580888 Chr.17: 44803098-44803098 on Build GRCh38
rs921457098 Chr.17: 44802840-44802840 on Build GRCh38
rs902913138 Chr.17: 44803064-44803064 on Build GRCh38
rs906420314 Chr.17: 44802919-44802919 on Build GRCh38
rs768159301 Chr.17: 44802896-44802896 on Build GRCh38
rs939280684 Chr.17: 44802967-44802967 on Build GRCh38
rs111754039 Chr.17: 44802765-44802765 on Build GRCh38
rs921230499 Chr.17: 44802864-44802865 on Build GRCh38
rs1031861444 Chr.17: 44802793-44802793 on Build GRCh38
rs541684730 Chr.17: 44802825-44802825 on Build GRCh38
rs1021951863 Chr.17: 44802766-44802766 on Build GRCh38
rs542105237 Chr.17: 44802634-44802634 on Build GRCh38
rs1003356605 Chr.17: 44802661-44802661 on Build GRCh38
rs140830998 Chr.17: 44803031-44803031 on Build GRCh38
rs570216351 Chr.17: 44803039-44803039 on Build GRCh38
rs73307237 Chr.17: 44803084-44803084 on Build GRCh38
rs966742418 Chr.17: 44802769-44802769 on Build GRCh38
rs895013121 Chr.17: 44803011-44803011 on Build GRCh38
rs909977116 Chr.17: 44802860-44802860 on Build GRCh38
rs940099560 Chr.17: 44802946-44802946 on Build GRCh38
rs769579986 Chr.17: 44802673-44802673 on Build GRCh38
rs773979920 Chr.17: 44802931-44802931 on Build GRCh38
rs1038344077 Chr.17: 44803009-44803009 on Build GRCh38
rs199743580 Chr.17: 44802650-44802651 on Build GRCh38
rs112806816 Chr.17: 44802733-44802733 on Build GRCh38
rs939979049 Chr.17: 44802875-44802875 on Build GRCh38
rs549357137 Chr.17: 44802738-44802738 on Build GRCh38
rs761501910 Chr.17: 44803013-44803013 on Build GRCh38
rs572866362 Chr.17: 44802877-44802877 on Build GRCh38
rs1054960986 Chr.17: 44802990-44802990 on Build GRCh38
rs1037323008 Chr.17: 44802651-44802651 on Build GRCh38
rs540393901 Chr.17: 44802980-44802980 on Build GRCh38
rs184353266 Chr.17: 44802844-44802844 on Build GRCh38

View Full Product Details