Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32209004-32209508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC2B
Entrez Gene ID
149069
Gene Name
doublecortin domain containing 2B
Gene Aliases
-
UniGene
Hs.424588
Gene Chromosome Location
Chr.1: 32208813-32216196 on Build GRCh38
Transcripts
NM_001099434.1 XM_017000413.1 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Gene Symbol
TMEM234
Entrez Gene ID
56063
Gene Name
transmembrane protein 234
Gene Aliases
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
UniGene
Hs.272299
Gene Chromosome Location
Chr.1: 32204807-32222371 on Build GRCh38
Transcripts
NM_001099434.1 XM_017000413.1 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1341797 c.41A>G p.Y14C Chr.1: 32209134-32209134 on Build GRCh38
1341798 c.142C>T p.Q48* Chr.1: 32209235-32209235 on Build GRCh38
681115 c.11G>A p.G4D Chr.1: 32209104-32209104 on Build GRCh38
5889496 c.64C>T p.P22S Chr.1: 32209157-32209157 on Build GRCh38
5969356 c.44G>A p.R15Q Chr.1: 32209137-32209137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370188556 Chr.1: 32209216-32209216 on Build GRCh38
rs374034989 Chr.1: 32209210-32209210 on Build GRCh38
rs370197875 Chr.1: 32209085-32209085 on Build GRCh38
rs752226727 Chr.1: 32209329-32209329 on Build GRCh38
rs182469374 Chr.1: 32209463-32209463 on Build GRCh38
rs757890291 Chr.1: 32209178-32209178 on Build GRCh38
rs760762297 Chr.1: 32209141-32209141 on Build GRCh38
rs775909721 Chr.1: 32209317-32209317 on Build GRCh38
rs541608920 Chr.1: 32209169-32209169 on Build GRCh38
rs201054559 Chr.1: 32209404-32209404 on Build GRCh38
rs751030874 Chr.1: 32209241-32209241 on Build GRCh38
rs770072822 Chr.1: 32209386-32209386 on Build GRCh38
rs771010041 Chr.1: 32209093-32209094 on Build GRCh38
rs758437536 Chr.1: 32209365-32209365 on Build GRCh38
rs535222020 Chr.1: 32209426-32209426 on Build GRCh38
rs748337153 Chr.1: 32209296-32209296 on Build GRCh38
rs761069388 Chr.1: 32209084-32209084 on Build GRCh38
rs755707694 Chr.1: 32209330-32209330 on Build GRCh38
rs765825628 Chr.1: 32209168-32209168 on Build GRCh38
rs111521118 Chr.1: 32209250-32209250 on Build GRCh38
rs749596756 Chr.1: 32209400-32209400 on Build GRCh38
rs565412152 Chr.1: 32209437-32209437 on Build GRCh38
rs558265251 Chr.1: 32209091-32209091 on Build GRCh38
rs952808854 Chr.1: 32209413-32209413 on Build GRCh38
rs755795758 Chr.1: 32209253-32209253 on Build GRCh38
rs988115084 Chr.1: 32209430-32209430 on Build GRCh38
rs770924908 Chr.1: 32209198-32209198 on Build GRCh38
rs200321560 Chr.1: 32209414-32209414 on Build GRCh38
rs773196452 Chr.1: 32209124-32209124 on Build GRCh38
rs60252907 Chr.1: 32209026-32209026 on Build GRCh38
rs367815378 Chr.1: 32209257-32209257 on Build GRCh38
rs754279333 Chr.1: 32209095-32209095 on Build GRCh38
rs762245396 Chr.1: 32209223-32209223 on Build GRCh38
rs757131404 Chr.1: 32209348-32209348 on Build GRCh38
rs546032813 Chr.1: 32209187-32209187 on Build GRCh38
rs200429239 Chr.1: 32209314-32209314 on Build GRCh38
rs757076900 Chr.1: 32209274-32209274 on Build GRCh38
rs903637223 Chr.1: 32209299-32209299 on Build GRCh38
rs754187448 Chr.1: 32209143-32209143 on Build GRCh38
rs185834533 Chr.1: 32209142-32209142 on Build GRCh38
rs1008861244 Chr.1: 32209334-32209334 on Build GRCh38
rs969163049 Chr.1: 32209130-32209130 on Build GRCh38
rs376703406 Chr.1: 32209188-32209188 on Build GRCh38
rs139959094 Chr.1: 32209123-32209123 on Build GRCh38
rs1013540136 Chr.1: 32209372-32209372 on Build GRCh38
rs746676743 Chr.1: 32209108-32209108 on Build GRCh38
rs925171375 Chr.1: 32209170-32209170 on Build GRCh38
rs747928383 Chr.1: 32209186-32209186 on Build GRCh38
rs144804850 Chr.1: 32209326-32209326 on Build GRCh38
rs376580354 Chr.1: 32209362-32209362 on Build GRCh38
rs200086869 Chr.1: 32209141-32209142 on Build GRCh38
rs773544057 Chr.1: 32209399-32209399 on Build GRCh38
rs778693860 Chr.1: 32209349-32209349 on Build GRCh38
rs372189623 Chr.1: 32209217-32209217 on Build GRCh38
rs747388452 Chr.1: 32209145-32209146 on Build GRCh38
rs567075358 Chr.1: 32209381-32209381 on Build GRCh38
rs552584926 Chr.1: 32209422-32209422 on Build GRCh38
rs770158832 Chr.1: 32209298-32209298 on Build GRCh38
rs201668101 Chr.1: 32209153-32209153 on Build GRCh38
rs747943790 Chr.1: 32209116-32209116 on Build GRCh38
rs562138510 Chr.1: 32209275-32209275 on Build GRCh38
rs545194543 Chr.1: 32209429-32209429 on Build GRCh38
rs753826825 Chr.1: 32209343-32209343 on Build GRCh38
rs749183467 Chr.1: 32209192-32209192 on Build GRCh38
rs779389396 Chr.1: 32209182-32209182 on Build GRCh38
rs766920886 Chr.1: 32209240-32209240 on Build GRCh38
rs763317133 Chr.1: 32209237-32209237 on Build GRCh38
rs917860954 Chr.1: 32209467-32209467 on Build GRCh38
rs139670831 Chr.1: 32209406-32209406 on Build GRCh38
rs552010076 Chr.1: 32209344-32209344 on Build GRCh38
rs947873722 Chr.1: 32209476-32209476 on Build GRCh38
rs149829527 Chr.1: 32209185-32209185 on Build GRCh38
rs556643639 Chr.1: 32209462-32209462 on Build GRCh38
rs556685749 Chr.1: 32209383-32209384 on Build GRCh38
rs193258165 Chr.1: 32209482-32209482 on Build GRCh38
rs529439787 Chr.1: 32209286-32209286 on Build GRCh38
rs528451377 Chr.1: 32209245-32209245 on Build GRCh38
rs749947975 Chr.1: 32209105-32209105 on Build GRCh38
rs766785566 Chr.1: 32209316-32209316 on Build GRCh38
rs758040202 Chr.1: 32209106-32209106 on Build GRCh38
rs748726107 Chr.1: 32209306-32209308 on Build GRCh38
rs754576599 Chr.1: 32209244-32209244 on Build GRCh38
rs546697848 Chr.1: 32209254-32209254 on Build GRCh38
rs551185618 Chr.1: 32209309-32209309 on Build GRCh38
rs747044777 Chr.1: 32209280-32209280 on Build GRCh38
rs373482876 Chr.1: 32209405-32209405 on Build GRCh38
rs780798619 Chr.1: 32209110-32209110 on Build GRCh38
rs779565006 Chr.1: 32209104-32209104 on Build GRCh38
rs1044909295 Chr.1: 32209477-32209477 on Build GRCh38
rs775670535 Chr.1: 32209137-32209137 on Build GRCh38
rs758108692 Chr.1: 32209453-32209453 on Build GRCh38
rs375238305 Chr.1: 32209246-32209246 on Build GRCh38
rs370943891 Chr.1: 32209135-32209135 on Build GRCh38
rs781687933 Chr.1: 32209042-32209042 on Build GRCh38
rs752390515 Chr.1: 32209252-32209252 on Build GRCh38
rs781771823 Chr.1: 32209301-32209317 on Build GRCh38
rs187561475 Chr.1: 32209481-32209481 on Build GRCh38
rs543475738 Chr.1: 32209331-32209331 on Build GRCh38
rs768905507 Chr.1: 32209285-32209285 on Build GRCh38
rs765733487 Chr.1: 32209232-32209232 on Build GRCh38
rs371689391 Chr.1: 32209269-32209269 on Build GRCh38
rs1027056724 Chr.1: 32209410-32209410 on Build GRCh38
rs574186982 Chr.1: 32209425-32209425 on Build GRCh38
rs773296459 Chr.1: 32209184-32209184 on Build GRCh38
rs201134740 Chr.1: 32209324-32209324 on Build GRCh38
rs760812516 Chr.1: 32209214-32209214 on Build GRCh38
rs200333134 Chr.1: 32209102-32209102 on Build GRCh38
rs759593031 Chr.1: 32209125-32209125 on Build GRCh38
rs769232405 Chr.1: 32209296-32209298 on Build GRCh38
rs190147459 Chr.1: 32209183-32209183 on Build GRCh38
rs374348561 Chr.1: 32209327-32209327 on Build GRCh38

View Full Product Details