Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32864571-32865056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
UniGene
Hs.524234
Gene Chromosome Location
Chr.1: 32862268-32872492 on Build GRCh38
Transcripts
NM_001171940.1 NM_001171941.2 NM_153756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4031546 c.347C>A p.T116N Chr.1: 32864725-32864725 on Build GRCh38
6665915 c.387C>T p.G129G Chr.1: 32864685-32864685 on Build GRCh38
6339497 c.393G>A p.G131G Chr.1: 32864679-32864679 on Build GRCh38
6460388 c.275-1G>T p.? Chr.1: 32864798-32864798 on Build GRCh38
1639844 c.275-8C>T p.? Chr.1: 32864805-32864805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753778328 Chr.1: 32864830-32864830 on Build GRCh38
rs1002247007 Chr.1: 32865000-32865000 on Build GRCh38
rs202111610 Chr.1: 32864736-32864736 on Build GRCh38
rs781141783 Chr.1: 32864801-32864802 on Build GRCh38
rs781114267 Chr.1: 32864616-32864616 on Build GRCh38
rs1054024714 Chr.1: 32865002-32865002 on Build GRCh38
rs751422783 Chr.1: 32864613-32864613 on Build GRCh38
rs760590429 Chr.1: 32864821-32864821 on Build GRCh38
rs559654305 Chr.1: 32864915-32864915 on Build GRCh38
rs976802356 Chr.1: 32864641-32864641 on Build GRCh38
rs759139981 Chr.1: 32864806-32864806 on Build GRCh38
rs200302649 Chr.1: 32864817-32864817 on Build GRCh38
rs754767987 Chr.1: 32864612-32864612 on Build GRCh38
rs535663114 Chr.1: 32864854-32864856 on Build GRCh38
rs143697125 Chr.1: 32864743-32864743 on Build GRCh38
rs1015268824 Chr.1: 32864993-32864993 on Build GRCh38
rs144593537 Chr.1: 32864671-32864671 on Build GRCh38
rs768908833 Chr.1: 32864659-32864659 on Build GRCh38
rs752535569 Chr.1: 32864818-32864818 on Build GRCh38
rs767234279 Chr.1: 32864698-32864698 on Build GRCh38
rs766250108 Chr.1: 32864608-32864608 on Build GRCh38
rs775703433 Chr.1: 32864646-32864646 on Build GRCh38
rs557154418 Chr.1: 32864719-32864719 on Build GRCh38
rs759315666 Chr.1: 32864693-32864693 on Build GRCh38
rs770475492 Chr.1: 32864777-32864777 on Build GRCh38
rs748095477 Chr.1: 32864802-32864803 on Build GRCh38
rs565030050 Chr.1: 32864802-32864802 on Build GRCh38
rs1046395666 Chr.1: 32864856-32864856 on Build GRCh38
rs780257477 Chr.1: 32864734-32864734 on Build GRCh38
rs781443548 Chr.1: 32864770-32864770 on Build GRCh38
rs757275309 Chr.1: 32864622-32864622 on Build GRCh38
rs925889359 Chr.1: 32864707-32864707 on Build GRCh38
rs377098383 Chr.1: 32864684-32864684 on Build GRCh38
rs1003844709 Chr.1: 32864988-32864988 on Build GRCh38
rs1021111696 Chr.1: 32864596-32864596 on Build GRCh38
rs777138875 Chr.1: 32864661-32864661 on Build GRCh38
rs886699947 Chr.1: 32864971-32864971 on Build GRCh38
rs185141197 Chr.1: 32864701-32864701 on Build GRCh38
rs900653780 Chr.1: 32864851-32864851 on Build GRCh38
rs770357935 Chr.1: 32864801-32864801 on Build GRCh38
rs894088416 Chr.1: 32865007-32865007 on Build GRCh38
rs761991187 Chr.1: 32864696-32864696 on Build GRCh38
rs769391902 Chr.1: 32864669-32864669 on Build GRCh38
rs762433406 Chr.1: 32864662-32864662 on Build GRCh38
rs72882318 Chr.1: 32864860-32864860 on Build GRCh38
rs140995767 Chr.1: 32864776-32864776 on Build GRCh38
rs905188918 Chr.1: 32864907-32864907 on Build GRCh38
rs748046461 Chr.1: 32864618-32864618 on Build GRCh38
rs934633233 Chr.1: 32864739-32864739 on Build GRCh38
rs988352401 Chr.1: 32864592-32864592 on Build GRCh38
rs755006302 Chr.1: 32864653-32864653 on Build GRCh38
rs145770830 Chr.1: 32864685-32864685 on Build GRCh38
rs754861512 Chr.1: 32864614-32864614 on Build GRCh38
rs764085437 Chr.1: 32864825-32864825 on Build GRCh38
rs948590303 Chr.1: 32864697-32864697 on Build GRCh38
rs758613897 Chr.1: 32864725-32864725 on Build GRCh38
rs756083445 Chr.1: 32864714-32864714 on Build GRCh38
rs778991988 Chr.1: 32864625-32864625 on Build GRCh38
rs1284368 Chr.1: 32864831-32864831 on Build GRCh38
rs772839146 Chr.1: 32864791-32864791 on Build GRCh38
rs372666011 Chr.1: 32864642-32864642 on Build GRCh38
rs1010121239 Chr.1: 32865028-32865028 on Build GRCh38
rs769018609 Chr.1: 32864745-32864745 on Build GRCh38
rs750559821 Chr.1: 32864722-32864722 on Build GRCh38
rs1027881252 Chr.1: 32864908-32864908 on Build GRCh38
rs960009018 Chr.1: 32865008-32865008 on Build GRCh38

View Full Product Details