Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32867933-32868427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
UniGene
Hs.524234
Gene Chromosome Location
Chr.1: 32862268-32872492 on Build GRCh38
Transcripts
NM_001171940.1 NM_001171941.2 NM_153756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665914 c.71C>T p.T24M Chr.1: 32868303-32868303 on Build GRCh38
1197152 c.19G>A p.E7K Chr.1: 32868355-32868355 on Build GRCh38
426114 c.99C>T p.I33I Chr.1: 32868275-32868275 on Build GRCh38
3488237 c.127G>A p.E43K Chr.1: 32868247-32868247 on Build GRCh38
908365 c.72G>A p.T24T Chr.1: 32868302-32868302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775897042 Chr.1: 32868383-32868385 on Build GRCh38
rs530794646 Chr.1: 32868094-32868094 on Build GRCh38
rs201963189 Chr.1: 32868224-32868224 on Build GRCh38
rs371967696 Chr.1: 32868174-32868174 on Build GRCh38
rs765656287 Chr.1: 32868339-32868339 on Build GRCh38
rs1021764052 Chr.1: 32868030-32868030 on Build GRCh38
rs545085198 Chr.1: 32868007-32868007 on Build GRCh38
rs376090198 Chr.1: 32868352-32868352 on Build GRCh38
rs1031880380 Chr.1: 32868367-32868367 on Build GRCh38
rs769640098 Chr.1: 32868247-32868247 on Build GRCh38
rs760769158 Chr.1: 32868400-32868400 on Build GRCh38
rs1057382663 Chr.1: 32868289-32868289 on Build GRCh38
rs185406506 Chr.1: 32868401-32868401 on Build GRCh38
rs140092865 Chr.1: 32868366-32868366 on Build GRCh38
rs543369094 Chr.1: 32868087-32868087 on Build GRCh38
rs373917435 Chr.1: 32868375-32868375 on Build GRCh38
rs562887636 Chr.1: 32867992-32867992 on Build GRCh38
rs777659320 Chr.1: 32868378-32868378 on Build GRCh38
rs924309421 Chr.1: 32868068-32868068 on Build GRCh38
rs561449267 Chr.1: 32868095-32868095 on Build GRCh38
rs780955257 Chr.1: 32868223-32868223 on Build GRCh38
rs369474610 Chr.1: 32868222-32868222 on Build GRCh38
rs958842415 Chr.1: 32868013-32868013 on Build GRCh38
rs754092673 Chr.1: 32868303-32868303 on Build GRCh38
rs761431219 Chr.1: 32868153-32868153 on Build GRCh38
rs759471585 Chr.1: 32868152-32868152 on Build GRCh38
rs772243605 Chr.1: 32868287-32868287 on Build GRCh38
rs747854483 Chr.1: 32868370-32868370 on Build GRCh38
rs1021798225 Chr.1: 32868337-32868337 on Build GRCh38
rs746399642 Chr.1: 32868204-32868204 on Build GRCh38
rs757797906 Chr.1: 32868212-32868212 on Build GRCh38
rs531847432 Chr.1: 32868128-32868128 on Build GRCh38
rs779570641 Chr.1: 32868215-32868215 on Build GRCh38
rs779220112 Chr.1: 32868203-32868203 on Build GRCh38
rs374868584 Chr.1: 32868139-32868139 on Build GRCh38
rs148205766 Chr.1: 32868005-32868005 on Build GRCh38
rs746104871 Chr.1: 32868155-32868164 on Build GRCh38
rs943195586 Chr.1: 32868126-32868126 on Build GRCh38
rs754211615 Chr.1: 32868180-32868180 on Build GRCh38
rs149496621 Chr.1: 32868326-32868326 on Build GRCh38
rs771047914 Chr.1: 32868394-32868394 on Build GRCh38
rs775706100 Chr.1: 32868396-32868396 on Build GRCh38
rs757775826 Chr.1: 32868181-32868181 on Build GRCh38
rs945457899 Chr.1: 32868064-32868064 on Build GRCh38
rs1015933326 Chr.1: 32868182-32868182 on Build GRCh38
rs752924099 Chr.1: 32868156-32868156 on Build GRCh38
rs776667620 Chr.1: 32868403-32868403 on Build GRCh38
rs377691602 Chr.1: 32868248-32868248 on Build GRCh38
rs887322173 Chr.1: 32867998-32867998 on Build GRCh38
rs775451802 Chr.1: 32868026-32868026 on Build GRCh38
rs768347509 Chr.1: 32868219-32868219 on Build GRCh38
rs910495617 Chr.1: 32868110-32868110 on Build GRCh38
rs1047622425 Chr.1: 32868244-32868244 on Build GRCh38
rs374085063 Chr.1: 32868342-32868342 on Build GRCh38
rs370511876 Chr.1: 32868369-32868369 on Build GRCh38
rs746635434 Chr.1: 32868217-32868217 on Build GRCh38
rs767723903 Chr.1: 32868155-32868155 on Build GRCh38
rs193075950 Chr.1: 32868048-32868048 on Build GRCh38
rs375151828 Chr.1: 32868361-32868361 on Build GRCh38
rs758944873 Chr.1: 32868348-32868348 on Build GRCh38
rs368291677 Chr.1: 32868173-32868173 on Build GRCh38
rs772354615 Chr.1: 32868299-32868299 on Build GRCh38
rs371060777 Chr.1: 32868302-32868302 on Build GRCh38
rs937981464 Chr.1: 32868031-32868031 on Build GRCh38
rs774488587 Chr.1: 32868149-32868149 on Build GRCh38
rs368641791 Chr.1: 32868081-32868081 on Build GRCh38
rs532898911 Chr.1: 32868294-32868294 on Build GRCh38
rs141144340 Chr.1: 32868058-32868058 on Build GRCh38
rs761156654 Chr.1: 32868399-32868400 on Build GRCh38
rs1053720473 Chr.1: 32868045-32868045 on Build GRCh38
rs765148360 Chr.1: 32868146-32868146 on Build GRCh38
rs762976068 Chr.1: 32868286-32868286 on Build GRCh38
rs368098828 Chr.1: 32868385-32868385 on Build GRCh38

View Full Product Details