Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33514641-33515146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000190.1 XM_017000194.1

SNP Detail

SNP ID Chromosome Location
rs1773021 Chr.1: 33514847-33514847 on Build GRCh38
rs983904285 Chr.1: 33514885-33514887 on Build GRCh38
rs946617222 Chr.1: 33514998-33514998 on Build GRCh38
rs143108281 Chr.1: 33514720-33514720 on Build GRCh38
rs144378343 Chr.1: 33514770-33514770 on Build GRCh38
rs1026799684 Chr.1: 33514749-33514749 on Build GRCh38
rs564357510 Chr.1: 33515005-33515005 on Build GRCh38
rs1008276503 Chr.1: 33514719-33514719 on Build GRCh38
rs1051259621 Chr.1: 33515037-33515037 on Build GRCh38
rs80144753 Chr.1: 33514947-33514947 on Build GRCh38
rs561489321 Chr.1: 33515051-33515051 on Build GRCh38
rs1019273747 Chr.1: 33514952-33514952 on Build GRCh38
rs960555086 Chr.1: 33514940-33514940 on Build GRCh38
rs148779568 Chr.1: 33514855-33514855 on Build GRCh38
rs530042126 Chr.1: 33514753-33514753 on Build GRCh38
rs915903987 Chr.1: 33514831-33514831 on Build GRCh38
rs963969446 Chr.1: 33514733-33514733 on Build GRCh38
rs950838943 Chr.1: 33514883-33514883 on Build GRCh38
rs775801962 Chr.1: 33514737-33514737 on Build GRCh38
rs1004412735 Chr.1: 33515068-33515068 on Build GRCh38
rs556377549 Chr.1: 33514852-33514852 on Build GRCh38
rs761226602 Chr.1: 33514840-33514840 on Build GRCh38
rs142412581 Chr.1: 33514936-33514936 on Build GRCh38
rs1031145681 Chr.1: 33514675-33514675 on Build GRCh38
rs890867135 Chr.1: 33515041-33515041 on Build GRCh38
rs934797415 Chr.1: 33514862-33514862 on Build GRCh38
rs540242394 Chr.1: 33514951-33514951 on Build GRCh38
rs935113388 Chr.1: 33515028-33515028 on Build GRCh38
rs754167578 Chr.1: 33515030-33515030 on Build GRCh38
rs1026829315 Chr.1: 33515032-33515032 on Build GRCh38
rs904712838 Chr.1: 33514860-33514860 on Build GRCh38
rs952524142 Chr.1: 33514768-33514768 on Build GRCh38
rs1015423411 Chr.1: 33515082-33515082 on Build GRCh38
rs952583361 Chr.1: 33515054-33515054 on Build GRCh38
rs1043781192 Chr.1: 33514672-33514672 on Build GRCh38
rs982899163 Chr.1: 33514781-33514781 on Build GRCh38
rs1009167422 Chr.1: 33514774-33514774 on Build GRCh38
rs996737062 Chr.1: 33515010-33515010 on Build GRCh38
rs938696890 Chr.1: 33514792-33514792 on Build GRCh38
rs1043407140 Chr.1: 33515000-33515000 on Build GRCh38
rs570861936 Chr.1: 33514971-33514971 on Build GRCh38
rs545543425 Chr.1: 33514915-33514915 on Build GRCh38
rs908585395 Chr.1: 33514787-33514787 on Build GRCh38
rs892145850 Chr.1: 33514710-33514710 on Build GRCh38
rs538125211 Chr.1: 33514805-33514805 on Build GRCh38
rs960432914 Chr.1: 33515086-33515086 on Build GRCh38
rs1017899225 Chr.1: 33514783-33514783 on Build GRCh38
rs761357510 Chr.1: 33514817-33514818 on Build GRCh38

View Full Product Details