Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33519336-33519840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000190.1 XM_017000194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7136553 c.10792C>A p.R3598R Chr.1: 33519622-33519622 on Build GRCh38
4995443 c.10347G>T p.E3449D Chr.1: 33519635-33519635 on Build GRCh38
4031633 c.10388A>G p.D3463G Chr.1: 33519594-33519594 on Build GRCh38
4995444 c.10779G>T p.E3593D Chr.1: 33519635-33519635 on Build GRCh38
3488355 c.10855G>A p.E3619K Chr.1: 33519559-33519559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963448972 Chr.1: 33519627-33519627 on Build GRCh38
rs763420853 Chr.1: 33519663-33519663 on Build GRCh38
rs775462284 Chr.1: 33519555-33519555 on Build GRCh38
rs745595223 Chr.1: 33519792-33519792 on Build GRCh38
rs752763690 Chr.1: 33519472-33519472 on Build GRCh38
rs967286435 Chr.1: 33519367-33519368 on Build GRCh38
rs776841949 Chr.1: 33519649-33519649 on Build GRCh38
rs924064764 Chr.1: 33519808-33519808 on Build GRCh38
rs754279272 Chr.1: 33519487-33519487 on Build GRCh38
rs765758109 Chr.1: 33519578-33519578 on Build GRCh38
rs748871928 Chr.1: 33519700-33519700 on Build GRCh38
rs553425214 Chr.1: 33519723-33519723 on Build GRCh38
rs144539492 Chr.1: 33519626-33519626 on Build GRCh38
rs1025727891 Chr.1: 33519661-33519661 on Build GRCh38
rs547464815 Chr.1: 33519465-33519465 on Build GRCh38
rs377399157 Chr.1: 33519560-33519560 on Build GRCh38
rs780673057 Chr.1: 33519595-33519595 on Build GRCh38
rs758443617 Chr.1: 33519797-33519797 on Build GRCh38
rs375756871 Chr.1: 33519718-33519718 on Build GRCh38
rs1018849607 Chr.1: 33519466-33519466 on Build GRCh38
rs777433837 Chr.1: 33519692-33519692 on Build GRCh38
rs769640452 Chr.1: 33519544-33519544 on Build GRCh38
rs144356782 Chr.1: 33519622-33519622 on Build GRCh38
rs758836289 Chr.1: 33519589-33519589 on Build GRCh38
rs760212066 Chr.1: 33519720-33519721 on Build GRCh38
rs139494192 Chr.1: 33519773-33519773 on Build GRCh38
rs2131776 Chr.1: 33519370-33519370 on Build GRCh38
rs750357513 Chr.1: 33519683-33519683 on Build GRCh38
rs964519993 Chr.1: 33519479-33519479 on Build GRCh38
rs758828284 Chr.1: 33519789-33519789 on Build GRCh38
rs367889259 Chr.1: 33519493-33519493 on Build GRCh38
rs1007049745 Chr.1: 33519412-33519412 on Build GRCh38
rs778569152 Chr.1: 33519705-33519705 on Build GRCh38
rs376968534 Chr.1: 33519806-33519806 on Build GRCh38
rs747855239 Chr.1: 33519596-33519596 on Build GRCh38
rs201053172 Chr.1: 33519591-33519591 on Build GRCh38
rs369421698 Chr.1: 33519682-33519682 on Build GRCh38
rs368570386 Chr.1: 33519380-33519380 on Build GRCh38
rs750745887 Chr.1: 33519587-33519587 on Build GRCh38
rs115143061 Chr.1: 33519577-33519577 on Build GRCh38
rs768925616 Chr.1: 33519638-33519638 on Build GRCh38
rs1026204466 Chr.1: 33519515-33519515 on Build GRCh38
rs140074927 Chr.1: 33519488-33519488 on Build GRCh38
rs747765036 Chr.1: 33519512-33519512 on Build GRCh38
rs761947014 Chr.1: 33519717-33519717 on Build GRCh38
rs369816497 Chr.1: 33519688-33519688 on Build GRCh38
rs777547697 Chr.1: 33519606-33519606 on Build GRCh38
rs776310945 Chr.1: 33519511-33519511 on Build GRCh38
rs147373387 Chr.1: 33519746-33519746 on Build GRCh38
rs574800074 Chr.1: 33519729-33519729 on Build GRCh38
rs767551024 Chr.1: 33519471-33519471 on Build GRCh38
rs752224836 Chr.1: 33519768-33519768 on Build GRCh38
rs765491387 Chr.1: 33519656-33519656 on Build GRCh38
rs544262159 Chr.1: 33519527-33519527 on Build GRCh38
rs1043755134 Chr.1: 33519386-33519386 on Build GRCh38
rs766075912 Chr.1: 33519751-33519751 on Build GRCh38
rs151161296 Chr.1: 33519675-33519675 on Build GRCh38
rs190091468 Chr.1: 33519408-33519408 on Build GRCh38
rs372027465 Chr.1: 33519570-33519570 on Build GRCh38
rs772977995 Chr.1: 33519551-33519551 on Build GRCh38
rs2641959 Chr.1: 33519753-33519753 on Build GRCh38
rs758934954 Chr.1: 33519494-33519494 on Build GRCh38
rs4653183 Chr.1: 33519387-33519387 on Build GRCh38
rs939135392 Chr.1: 33519362-33519362 on Build GRCh38
rs769379800 Chr.1: 33519597-33519597 on Build GRCh38
rs1011401456 Chr.1: 33519378-33519378 on Build GRCh38
rs762143412 Chr.1: 33519651-33519651 on Build GRCh38
rs745876607 Chr.1: 33519711-33519711 on Build GRCh38
rs999745624 Chr.1: 33519388-33519388 on Build GRCh38
rs145499643 Chr.1: 33519504-33519507 on Build GRCh38
rs974219459 Chr.1: 33519633-33519633 on Build GRCh38
rs916387848 Chr.1: 33519526-33519526 on Build GRCh38
rs770802057 Chr.1: 33519704-33519704 on Build GRCh38
rs144791588 Chr.1: 33519407-33519407 on Build GRCh38
rs370039558 Chr.1: 33519722-33519722 on Build GRCh38
rs1055913545 Chr.1: 33519531-33519531 on Build GRCh38
rs930193402 Chr.1: 33519497-33519497 on Build GRCh38
rs773521570 Chr.1: 33519721-33519721 on Build GRCh38
rs779447156 Chr.1: 33519502-33519502 on Build GRCh38
rs755671126 Chr.1: 33519771-33519771 on Build GRCh38
rs755793348 Chr.1: 33519691-33519691 on Build GRCh38
rs1029971221 Chr.1: 33519375-33519375 on Build GRCh38
rs760761552 Chr.1: 33519559-33519559 on Build GRCh38
rs752245115 Chr.1: 33519683-33519683 on Build GRCh38
rs375118791 Chr.1: 33519817-33519817 on Build GRCh38
rs868675974 Chr.1: 33519557-33519557 on Build GRCh38
rs766618327 Chr.1: 33519672-33519672 on Build GRCh38
rs373497277 Chr.1: 33519787-33519787 on Build GRCh38
rs914697607 Chr.1: 33519737-33519737 on Build GRCh38
rs766993483 Chr.1: 33519675-33519677 on Build GRCh38
rs983155423 Chr.1: 33519752-33519752 on Build GRCh38
rs546495050 Chr.1: 33519786-33519786 on Build GRCh38
rs529700677 Chr.1: 33519503-33519503 on Build GRCh38
rs752793462 Chr.1: 33519552-33519552 on Build GRCh38
rs750787323 Chr.1: 33519662-33519662 on Build GRCh38
rs201139787 Chr.1: 33519504-33519504 on Build GRCh38
rs764136636 Chr.1: 33519507-33519508 on Build GRCh38
rs754553648 Chr.1: 33519593-33519593 on Build GRCh38
rs376862875 Chr.1: 33519592-33519592 on Build GRCh38
rs77991929 Chr.1: 33519635-33519635 on Build GRCh38
rs149418436 Chr.1: 33519359-33519359 on Build GRCh38
rs370646948 Chr.1: 33519807-33519807 on Build GRCh38
rs774397249 Chr.1: 33519630-33519630 on Build GRCh38
rs767914404 Chr.1: 33519478-33519478 on Build GRCh38
rs145294754 Chr.1: 33519654-33519654 on Build GRCh38
rs144802253 Chr.1: 33519499-33519500 on Build GRCh38
rs766271230 Chr.1: 33519554-33519554 on Build GRCh38
rs372464364 Chr.1: 33519714-33519714 on Build GRCh38
rs898637308 Chr.1: 33519703-33519703 on Build GRCh38
rs1055218802 Chr.1: 33519365-33519365 on Build GRCh38

View Full Product Details