Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33523201-33523711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000189.1 XM_017000190.1 XM_017000194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6405912 c.10457T>G p.V3486G Chr.1: 33523359-33523359 on Build GRCh38
6579597 c.10443G>C p.Q3481H Chr.1: 33523373-33523373 on Build GRCh38
4031638 c.10497T>G p.A3499A Chr.1: 33523319-33523319 on Build GRCh38
3488366 c.10431C>T p.L3477L Chr.1: 33523385-33523385 on Build GRCh38
6579596 c.10011G>C p.Q3337H Chr.1: 33523373-33523373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898010985 Chr.1: 33523446-33523446 on Build GRCh38
rs909389271 Chr.1: 33523600-33523600 on Build GRCh38
rs932359975 Chr.1: 33523313-33523313 on Build GRCh38
rs560050966 Chr.1: 33523496-33523496 on Build GRCh38
rs538183222 Chr.1: 33523562-33523562 on Build GRCh38
rs563041307 Chr.1: 33523533-33523533 on Build GRCh38
rs12038835 Chr.1: 33523561-33523561 on Build GRCh38
rs1021027351 Chr.1: 33523570-33523570 on Build GRCh38
rs781410958 Chr.1: 33523386-33523386 on Build GRCh38
rs555430648 Chr.1: 33523413-33523413 on Build GRCh38
rs988656248 Chr.1: 33523567-33523567 on Build GRCh38
rs771970521 Chr.1: 33523534-33523534 on Build GRCh38
rs767275022 Chr.1: 33523291-33523291 on Build GRCh38
rs939669821 Chr.1: 33523657-33523657 on Build GRCh38
rs766419014 Chr.1: 33523377-33523377 on Build GRCh38
rs750527237 Chr.1: 33523348-33523348 on Build GRCh38
rs760612736 Chr.1: 33523339-33523339 on Build GRCh38
rs543154254 Chr.1: 33523237-33523237 on Build GRCh38
rs183674702 Chr.1: 33523635-33523635 on Build GRCh38
rs773899854 Chr.1: 33523432-33523432 on Build GRCh38
rs909527774 Chr.1: 33523650-33523650 on Build GRCh38
rs967426270 Chr.1: 33523457-33523457 on Build GRCh38
rs200252481 Chr.1: 33523320-33523320 on Build GRCh38
rs757048148 Chr.1: 33523653-33523657 on Build GRCh38
rs770541007 Chr.1: 33523273-33523273 on Build GRCh38
rs773915507 Chr.1: 33523277-33523277 on Build GRCh38
rs923279131 Chr.1: 33523523-33523523 on Build GRCh38
rs201889409 Chr.1: 33523396-33523396 on Build GRCh38
rs769971240 Chr.1: 33523332-33523332 on Build GRCh38
rs553898625 Chr.1: 33523675-33523675 on Build GRCh38
rs755166197 Chr.1: 33523385-33523385 on Build GRCh38
rs770355203 Chr.1: 33523258-33523258 on Build GRCh38
rs932244716 Chr.1: 33523525-33523525 on Build GRCh38
rs756710152 Chr.1: 33523393-33523393 on Build GRCh38
rs577822488 Chr.1: 33523640-33523640 on Build GRCh38
rs772877640 Chr.1: 33523261-33523261 on Build GRCh38
rs759221735 Chr.1: 33523285-33523285 on Build GRCh38
rs921184003 Chr.1: 33523626-33523626 on Build GRCh38
rs976611718 Chr.1: 33523601-33523601 on Build GRCh38
rs748502931 Chr.1: 33523388-33523388 on Build GRCh38
rs138224779 Chr.1: 33523363-33523363 on Build GRCh38
rs533868406 Chr.1: 33523238-33523238 on Build GRCh38
rs1002204780 Chr.1: 33523521-33523521 on Build GRCh38
rs267598561 Chr.1: 33523310-33523310 on Build GRCh38
rs1036532251 Chr.1: 33523658-33523658 on Build GRCh38
rs372999987 Chr.1: 33523400-33523400 on Build GRCh38
rs751799901 Chr.1: 33523379-33523379 on Build GRCh38
rs749781963 Chr.1: 33523411-33523411 on Build GRCh38
rs775424760 Chr.1: 33523683-33523683 on Build GRCh38
rs777822018 Chr.1: 33523387-33523387 on Build GRCh38
rs186106486 Chr.1: 33523651-33523651 on Build GRCh38
rs35685821 Chr.1: 33523258-33523259 on Build GRCh38
rs763981166 Chr.1: 33523340-33523340 on Build GRCh38

View Full Product Details