Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33526962-33527371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000189.1 XM_017000190.1 XM_017000194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5557172 c.10197C>T p.I3399I Chr.1: 33527233-33527233 on Build GRCh38
1582241 c.10177C>T p.R3393W Chr.1: 33527253-33527253 on Build GRCh38
4596290 c.9776G>A p.R3259Q Chr.1: 33527222-33527222 on Build GRCh38
4675022 c.10192C>G p.P3398A Chr.1: 33527238-33527238 on Build GRCh38
5557171 c.9765C>T p.I3255I Chr.1: 33527233-33527233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779428319 Chr.1: 33527185-33527185 on Build GRCh38
rs759399395 Chr.1: 33527125-33527125 on Build GRCh38
rs920061483 Chr.1: 33527178-33527178 on Build GRCh38
rs770321719 Chr.1: 33527287-33527287 on Build GRCh38
rs541384120 Chr.1: 33527052-33527052 on Build GRCh38
rs149656829 Chr.1: 33527223-33527223 on Build GRCh38
rs929040350 Chr.1: 33526991-33526991 on Build GRCh38
rs563257236 Chr.1: 33527065-33527065 on Build GRCh38
rs756937873 Chr.1: 33527268-33527268 on Build GRCh38
rs762288960 Chr.1: 33527272-33527272 on Build GRCh38
rs773240938 Chr.1: 33527235-33527235 on Build GRCh38
rs760982366 Chr.1: 33527253-33527253 on Build GRCh38
rs1012061292 Chr.1: 33526989-33526989 on Build GRCh38
rs370729424 Chr.1: 33527222-33527222 on Build GRCh38
rs979456684 Chr.1: 33527050-33527050 on Build GRCh38
rs769450644 Chr.1: 33527233-33527233 on Build GRCh38
rs757969708 Chr.1: 33527209-33527209 on Build GRCh38
rs762794065 Chr.1: 33527237-33527237 on Build GRCh38
rs746725403 Chr.1: 33527214-33527214 on Build GRCh38
rs1000913899 Chr.1: 33527307-33527307 on Build GRCh38
rs1031120378 Chr.1: 33527331-33527331 on Build GRCh38
rs757738247 Chr.1: 33527162-33527162 on Build GRCh38
rs1023444456 Chr.1: 33527295-33527295 on Build GRCh38
rs1023405063 Chr.1: 33527006-33527006 on Build GRCh38
rs956699010 Chr.1: 33527069-33527069 on Build GRCh38
rs968120385 Chr.1: 33527043-33527043 on Build GRCh38
rs754279246 Chr.1: 33527259-33527259 on Build GRCh38
rs1012923748 Chr.1: 33527335-33527335 on Build GRCh38
rs975606309 Chr.1: 33527134-33527134 on Build GRCh38
rs111577457 Chr.1: 33527212-33527212 on Build GRCh38
rs1031426899 Chr.1: 33527062-33527062 on Build GRCh38
rs765490689 Chr.1: 33527301-33527301 on Build GRCh38
rs148437366 Chr.1: 33527252-33527252 on Build GRCh38
rs942911238 Chr.1: 33527128-33527128 on Build GRCh38
rs372797467 Chr.1: 33527324-33527324 on Build GRCh38
rs912853138 Chr.1: 33527126-33527126 on Build GRCh38
rs754189365 Chr.1: 33527161-33527161 on Build GRCh38
rs530687401 Chr.1: 33527192-33527192 on Build GRCh38
rs764372678 Chr.1: 33527257-33527257 on Build GRCh38

View Full Product Details