Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34761574-34762070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB4
Entrez Gene ID
127534
Gene Name
gap junction protein beta 4
Gene Aliases
CX30.3, EKV
UniGene
Hs.351203
Gene Chromosome Location
Chr.1: 34759741-34763724 on Build GRCh38
Transcripts
NM_153212.2 XM_011540679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488818 c.544G>A p.E182K Chr.1: 34761798-34761798 on Build GRCh38
4687023 c.452G>A p.R151H Chr.1: 34761706-34761706 on Build GRCh38
1342069 c.668G>A p.R223Q Chr.1: 34761922-34761922 on Build GRCh38
6210627 c.536G>A p.R179Q Chr.1: 34761790-34761790 on Build GRCh38
3488816 c.405C>T p.L135L Chr.1: 34761659-34761659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561459140 Chr.1: 34761678-34761678 on Build GRCh38
rs757433582 Chr.1: 34761656-34761656 on Build GRCh38
rs749875009 Chr.1: 34761923-34761923 on Build GRCh38
rs373126632 Chr.1: 34761624-34761624 on Build GRCh38
rs745548746 Chr.1: 34761887-34761887 on Build GRCh38
rs1032959733 Chr.1: 34761618-34761618 on Build GRCh38
rs992335161 Chr.1: 34761781-34761781 on Build GRCh38
rs375489958 Chr.1: 34761928-34761928 on Build GRCh38
rs779150355 Chr.1: 34761605-34761605 on Build GRCh38
rs781088813 Chr.1: 34761941-34761941 on Build GRCh38
rs771162179 Chr.1: 34761906-34761906 on Build GRCh38
rs758125233 Chr.1: 34761927-34761927 on Build GRCh38
rs759845411 Chr.1: 34761908-34761908 on Build GRCh38
rs763878424 Chr.1: 34761838-34761838 on Build GRCh38
rs201339671 Chr.1: 34761783-34761783 on Build GRCh38
rs199901284 Chr.1: 34761695-34761695 on Build GRCh38
rs750680293 Chr.1: 34761723-34761723 on Build GRCh38
rs760944349 Chr.1: 34761913-34761913 on Build GRCh38
rs78499418 Chr.1: 34761705-34761705 on Build GRCh38
rs77021296 Chr.1: 34761703-34761703 on Build GRCh38
rs905889823 Chr.1: 34761608-34761608 on Build GRCh38
rs762708249 Chr.1: 34761689-34761689 on Build GRCh38
rs199685128 Chr.1: 34761762-34761762 on Build GRCh38
rs780567526 Chr.1: 34761729-34761729 on Build GRCh38
rs765716688 Chr.1: 34761988-34761988 on Build GRCh38
rs80358206 Chr.1: 34761665-34761665 on Build GRCh38
rs756019816 Chr.1: 34762025-34762025 on Build GRCh38
rs527959955 Chr.1: 34761646-34761646 on Build GRCh38
rs185327282 Chr.1: 34761933-34761933 on Build GRCh38
rs180965135 Chr.1: 34761918-34761918 on Build GRCh38
rs768318044 Chr.1: 34761934-34761934 on Build GRCh38
rs561519962 Chr.1: 34761851-34761851 on Build GRCh38
rs370157756 Chr.1: 34761674-34761674 on Build GRCh38
rs778129905 Chr.1: 34761868-34761868 on Build GRCh38
rs200936960 Chr.1: 34761921-34761921 on Build GRCh38
rs199688130 Chr.1: 34761595-34761595 on Build GRCh38
rs572049955 Chr.1: 34761801-34761801 on Build GRCh38
rs748688362 Chr.1: 34761800-34761800 on Build GRCh38
rs756737829 Chr.1: 34761880-34761880 on Build GRCh38
rs534122483 Chr.1: 34761606-34761606 on Build GRCh38
rs749997834 Chr.1: 34761989-34761989 on Build GRCh38
rs766751826 Chr.1: 34761846-34761846 on Build GRCh38
rs1034367247 Chr.1: 34761772-34761772 on Build GRCh38
rs111693060 Chr.1: 34761770-34761770 on Build GRCh38
rs754364013 Chr.1: 34761919-34761919 on Build GRCh38
rs140996335 Chr.1: 34761625-34761625 on Build GRCh38
rs764398093 Chr.1: 34761965-34761965 on Build GRCh38
rs201369860 Chr.1: 34761740-34761740 on Build GRCh38
rs142410428 Chr.1: 34761796-34761796 on Build GRCh38
rs565868320 Chr.1: 34761947-34761947 on Build GRCh38
rs146979528 Chr.1: 34761630-34761630 on Build GRCh38
rs548184524 Chr.1: 34761962-34761962 on Build GRCh38
rs774534707 Chr.1: 34761907-34761907 on Build GRCh38
rs753414684 Chr.1: 34761879-34761879 on Build GRCh38
rs757329422 Chr.1: 34761711-34761711 on Build GRCh38
rs80358207 Chr.1: 34761663-34761663 on Build GRCh38
rs142843509 Chr.1: 34761623-34761623 on Build GRCh38
rs765425278 Chr.1: 34761712-34761712 on Build GRCh38
rs776109822 Chr.1: 34761653-34761653 on Build GRCh38
rs757298436 Chr.1: 34761799-34761801 on Build GRCh38
rs760662564 Chr.1: 34761831-34761831 on Build GRCh38
rs776912649 Chr.1: 34761975-34761975 on Build GRCh38
rs145404340 Chr.1: 34761953-34761953 on Build GRCh38
rs980447753 Chr.1: 34761982-34761982 on Build GRCh38
rs758507242 Chr.1: 34761727-34761727 on Build GRCh38
rs753694964 Chr.1: 34761796-34761796 on Build GRCh38
rs751196580 Chr.1: 34762013-34762013 on Build GRCh38
rs79193415 Chr.1: 34761761-34761761 on Build GRCh38
rs141637346 Chr.1: 34761750-34761750 on Build GRCh38
rs775812981 Chr.1: 34761931-34761931 on Build GRCh38
rs750400283 Chr.1: 34761765-34761765 on Build GRCh38
rs1010697749 Chr.1: 34762022-34762022 on Build GRCh38
rs148710003 Chr.1: 34761732-34761732 on Build GRCh38
rs901054727 Chr.1: 34761759-34761760 on Build GRCh38
rs766101103 Chr.1: 34762012-34762012 on Build GRCh38
rs190460237 Chr.1: 34761610-34761610 on Build GRCh38
rs777849647 Chr.1: 34762027-34762027 on Build GRCh38
rs190194468 Chr.1: 34761963-34761963 on Build GRCh38
rs1019540630 Chr.1: 34761628-34761628 on Build GRCh38
rs537606599 Chr.1: 34761766-34761766 on Build GRCh38
rs539139170 Chr.1: 34761733-34761733 on Build GRCh38
rs1024273124 Chr.1: 34761939-34761939 on Build GRCh38
rs976388855 Chr.1: 34762034-34762034 on Build GRCh38
rs146404415 Chr.1: 34761596-34761596 on Build GRCh38
rs267598569 Chr.1: 34761798-34761798 on Build GRCh38
rs769403446 Chr.1: 34761683-34761683 on Build GRCh38
rs923722828 Chr.1: 34761708-34761708 on Build GRCh38
rs755856129 Chr.1: 34761761-34761762 on Build GRCh38
rs543341299 Chr.1: 34761822-34761822 on Build GRCh38
rs147912521 Chr.1: 34761749-34761749 on Build GRCh38
rs772459717 Chr.1: 34761667-34761667 on Build GRCh38
rs776398059 Chr.1: 34761901-34761901 on Build GRCh38
rs538347700 Chr.1: 34762039-34762039 on Build GRCh38
rs770506626 Chr.1: 34761756-34761756 on Build GRCh38
rs370286559 Chr.1: 34761706-34761706 on Build GRCh38
rs142189692 Chr.1: 34761675-34761675 on Build GRCh38
rs74320948 Chr.1: 34761700-34761700 on Build GRCh38
rs747548027 Chr.1: 34761672-34761672 on Build GRCh38
rs201000959 Chr.1: 34761984-34761984 on Build GRCh38
rs778397300 Chr.1: 34761922-34761922 on Build GRCh38
rs773395030 Chr.1: 34761843-34761843 on Build GRCh38
rs779909279 Chr.1: 34761895-34761895 on Build GRCh38
rs148656277 Chr.1: 34762024-34762024 on Build GRCh38
rs547453110 Chr.1: 34761734-34761734 on Build GRCh38
rs3738346 Chr.1: 34761865-34761865 on Build GRCh38
rs779192023 Chr.1: 34761659-34761659 on Build GRCh38
rs777430011 Chr.1: 34761845-34761845 on Build GRCh38
rs758544896 Chr.1: 34761878-34761878 on Build GRCh38
rs148182439 Chr.1: 34761640-34761640 on Build GRCh38
rs769791668 Chr.1: 34761949-34761949 on Build GRCh38
rs370780147 Chr.1: 34761823-34761823 on Build GRCh38
rs147456169 Chr.1: 34761899-34761899 on Build GRCh38
rs376895405 Chr.1: 34761644-34761644 on Build GRCh38
rs753166307 Chr.1: 34761910-34761910 on Build GRCh38
rs568452902 Chr.1: 34761952-34761952 on Build GRCh38
rs80358213 Chr.1: 34761820-34761820 on Build GRCh38
rs759786284 Chr.1: 34761857-34761857 on Build GRCh38
rs765388097 Chr.1: 34761764-34761764 on Build GRCh38
rs773961527 Chr.1: 34761819-34761819 on Build GRCh38
rs775597117 Chr.1: 34762046-34762046 on Build GRCh38
rs549139696 Chr.1: 34761829-34761829 on Build GRCh38
rs748713797 Chr.1: 34761790-34761790 on Build GRCh38
rs753849633 Chr.1: 34761651-34761651 on Build GRCh38
rs199996022 Chr.1: 34762017-34762017 on Build GRCh38
rs147830069 Chr.1: 34761629-34761629 on Build GRCh38
rs80142792 Chr.1: 34761881-34761881 on Build GRCh38
rs566099103 Chr.1: 34761735-34761735 on Build GRCh38
rs752939052 Chr.1: 34761862-34761862 on Build GRCh38
rs145234213 Chr.1: 34761789-34761789 on Build GRCh38
rs199674742 Chr.1: 34761643-34761643 on Build GRCh38
rs760477640 Chr.1: 34761763-34761763 on Build GRCh38
rs531991614 Chr.1: 34761685-34761685 on Build GRCh38
rs1033572727 Chr.1: 34761832-34761832 on Build GRCh38
rs770576472 Chr.1: 34761635-34761635 on Build GRCh38
rs1033213319 Chr.1: 34762001-34762001 on Build GRCh38
rs150737005 Chr.1: 34761671-34761671 on Build GRCh38
rs556066067 Chr.1: 34762018-34762018 on Build GRCh38
rs766186018 Chr.1: 34761696-34761696 on Build GRCh38
rs199544688 Chr.1: 34761797-34761797 on Build GRCh38
rs149110828 Chr.1: 34761638-34761638 on Build GRCh38
rs777679951 Chr.1: 34761762-34761763 on Build GRCh38
rs565502726 Chr.1: 34761905-34761905 on Build GRCh38
rs772066664 Chr.1: 34762045-34762045 on Build GRCh38
rs771645189 Chr.1: 34761892-34761892 on Build GRCh38

View Full Product Details