Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59272593-59273089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD1
Entrez Gene ID
284161
Gene Name
glycerophosphodiester phosphodiesterase domain containing 1
Gene Aliases
GDE4
UniGene
Hs.631744
Gene Chromosome Location
Chr.17: 59220467-59275970 on Build GRCh38
Transcripts
NM_001165993.1 NM_001165994.1 NM_182569.3 XM_017024521.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670351 c.799C>T p.H267Y Chr.17: 59272813-59272813 on Build GRCh38
4408336 c.771-6T>G p.? Chr.17: 59272779-59272779 on Build GRCh38
6670356 c.860G>T p.S287I Chr.17: 59272874-59272874 on Build GRCh38
6281719 c.791+10C>G p.? Chr.17: 59272815-59272815 on Build GRCh38
6864843 c.771-9C>G p.? Chr.17: 59272776-59272776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772718486 Chr.17: 59272846-59272846 on Build GRCh38
rs747394498 Chr.17: 59272869-59272869 on Build GRCh38
rs944080163 Chr.17: 59272963-59272963 on Build GRCh38
rs1000943995 Chr.17: 59273026-59273026 on Build GRCh38
rs971066470 Chr.17: 59272694-59272694 on Build GRCh38
rs754070029 Chr.17: 59272796-59272796 on Build GRCh38
rs762559243 Chr.17: 59272881-59272881 on Build GRCh38
rs113976338 Chr.17: 59272680-59272680 on Build GRCh38
rs760440423 Chr.17: 59272902-59272902 on Build GRCh38
rs531664396 Chr.17: 59272875-59272875 on Build GRCh38
rs904385847 Chr.17: 59272803-59272806 on Build GRCh38
rs779163162 Chr.17: 59272825-59272825 on Build GRCh38
rs1004973026 Chr.17: 59272903-59272903 on Build GRCh38
rs752671670 Chr.17: 59272901-59272901 on Build GRCh38
rs780543732 Chr.17: 59272861-59272861 on Build GRCh38
rs763920757 Chr.17: 59272907-59272907 on Build GRCh38
rs773867622 Chr.17: 59272894-59272894 on Build GRCh38
rs911562218 Chr.17: 59273063-59273063 on Build GRCh38
rs1015076070 Chr.17: 59272956-59272956 on Build GRCh38
rs745682734 Chr.17: 59272886-59272886 on Build GRCh38
rs751535149 Chr.17: 59272749-59272749 on Build GRCh38
rs963592003 Chr.17: 59272967-59272967 on Build GRCh38
rs867705113 Chr.17: 59272808-59272808 on Build GRCh38
rs1032519864 Chr.17: 59273033-59273033 on Build GRCh38
rs867056648 Chr.17: 59272656-59272656 on Build GRCh38
rs931693413 Chr.17: 59272648-59272648 on Build GRCh38
rs767434226 Chr.17: 59272776-59272776 on Build GRCh38
rs1042764065 Chr.17: 59272984-59272984 on Build GRCh38
rs1012523836 Chr.17: 59273062-59273062 on Build GRCh38
rs377264346 Chr.17: 59272754-59272754 on Build GRCh38
rs139432483 Chr.17: 59273005-59273005 on Build GRCh38
rs542196155 Chr.17: 59272886-59272886 on Build GRCh38
rs959647017 Chr.17: 59272672-59272672 on Build GRCh38
rs370216059 Chr.17: 59272757-59272757 on Build GRCh38
rs765849764 Chr.17: 59272740-59272740 on Build GRCh38
rs1041493731 Chr.17: 59272719-59272719 on Build GRCh38
rs77154698 Chr.17: 59272684-59272684 on Build GRCh38
rs375761996 Chr.17: 59272678-59272678 on Build GRCh38
rs755937632 Chr.17: 59272781-59272781 on Build GRCh38
rs74472114 Chr.17: 59272896-59272896 on Build GRCh38
rs778901623 Chr.17: 59272857-59272858 on Build GRCh38
rs572365736 Chr.17: 59272627-59272627 on Build GRCh38
rs921564570 Chr.17: 59272632-59272632 on Build GRCh38
rs945809983 Chr.17: 59272714-59272714 on Build GRCh38
rs753648441 Chr.17: 59272909-59272909 on Build GRCh38
rs895279782 Chr.17: 59273041-59273041 on Build GRCh38
rs777915020 Chr.17: 59272784-59272784 on Build GRCh38
rs745893247 Chr.17: 59272826-59272826 on Build GRCh38
rs757572596 Chr.17: 59272931-59272931 on Build GRCh38
rs973909313 Chr.17: 59272852-59272852 on Build GRCh38
rs561829130 Chr.17: 59272944-59272944 on Build GRCh38
rs572456157 Chr.17: 59272815-59272815 on Build GRCh38

View Full Product Details