Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60868590-60869094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1384964 c.583C>T p.R195W Chr.17: 60868682-60868682 on Build GRCh38
5005718 c.559A>G p.I187V Chr.17: 60868658-60868658 on Build GRCh38
5386946 c.524G>A p.G175E Chr.17: 60868623-60868623 on Build GRCh38
7262723 c.538T>C p.S180P Chr.17: 60868637-60868637 on Build GRCh38
706592 c.528G>C p.E176D Chr.17: 60868627-60868627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185132789 Chr.17: 60868689-60868689 on Build GRCh38
rs568030994 Chr.17: 60869059-60869059 on Build GRCh38
rs949430011 Chr.17: 60868620-60868620 on Build GRCh38
rs966510965 Chr.17: 60868836-60868836 on Build GRCh38
rs745740251 Chr.17: 60868868-60868868 on Build GRCh38
rs182399122 Chr.17: 60868772-60868772 on Build GRCh38
rs929593151 Chr.17: 60868788-60868788 on Build GRCh38
rs542393076 Chr.17: 60868724-60868724 on Build GRCh38
rs758431349 Chr.17: 60868963-60868963 on Build GRCh38
rs114162805 Chr.17: 60868850-60868850 on Build GRCh38
rs751781813 Chr.17: 60868725-60868725 on Build GRCh38
rs187003226 Chr.17: 60868959-60868959 on Build GRCh38
rs901919017 Chr.17: 60868914-60868914 on Build GRCh38
rs770388498 Chr.17: 60868630-60868630 on Build GRCh38
rs1032150988 Chr.17: 60868865-60868865 on Build GRCh38
rs759133042 Chr.17: 60868640-60868640 on Build GRCh38
rs867427251 Chr.17: 60868729-60868729 on Build GRCh38
rs8066880 Chr.17: 60868831-60868831 on Build GRCh38
rs949404818 Chr.17: 60868903-60868903 on Build GRCh38
rs775069588 Chr.17: 60868645-60868645 on Build GRCh38
rs8067208 Chr.17: 60869064-60869064 on Build GRCh38
rs956207765 Chr.17: 60869024-60869024 on Build GRCh38
rs749209369 Chr.17: 60868617-60868617 on Build GRCh38
rs957928593 Chr.17: 60868880-60868880 on Build GRCh38
rs537317548 Chr.17: 60868970-60868971 on Build GRCh38
rs771698366 Chr.17: 60868643-60868643 on Build GRCh38
rs773855608 Chr.17: 60868633-60868633 on Build GRCh38
rs762437454 Chr.17: 60868683-60868683 on Build GRCh38
rs377752373 Chr.17: 60868682-60868682 on Build GRCh38
rs1017740505 Chr.17: 60868897-60868897 on Build GRCh38
rs568757901 Chr.17: 60868851-60868851 on Build GRCh38
rs1029617080 Chr.17: 60868666-60868666 on Build GRCh38
rs916545345 Chr.17: 60868889-60868889 on Build GRCh38
rs559870934 Chr.17: 60868783-60868783 on Build GRCh38
rs932187242 Chr.17: 60868979-60868979 on Build GRCh38
rs778005110 Chr.17: 60868849-60868849 on Build GRCh38
rs982173313 Chr.17: 60868912-60868912 on Build GRCh38
rs1037897093 Chr.17: 60868698-60868698 on Build GRCh38
rs761069193 Chr.17: 60868675-60868675 on Build GRCh38
rs777460667 Chr.17: 60868727-60868727 on Build GRCh38
rs1007929333 Chr.17: 60868785-60868785 on Build GRCh38
rs899349404 Chr.17: 60868974-60868974 on Build GRCh38
rs527267658 Chr.17: 60868884-60868884 on Build GRCh38
rs773183785 Chr.17: 60868616-60868616 on Build GRCh38
rs114586742 Chr.17: 60868770-60868770 on Build GRCh38
rs939055435 Chr.17: 60868764-60868764 on Build GRCh38
rs997564643 Chr.17: 60868940-60868940 on Build GRCh38
rs151035589 Chr.17: 60868730-60868730 on Build GRCh38
rs758620599 Chr.17: 60868716-60868716 on Build GRCh38
rs188031327 Chr.17: 60868690-60868690 on Build GRCh38
rs764414602 Chr.17: 60868679-60868679 on Build GRCh38

View Full Product Details