Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61681960-61682466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRIP1
Entrez Gene ID
83990
Gene Name
BRCA1 interacting protein C-terminal helicase 1
Gene Aliases
BACH1, FANCJ, OF
UniGene
Hs.128903
Gene Chromosome Location
Chr.17: 61679186-61864120 on Build GRCh38
Transcripts
NM_032043.2 XM_011525332.2 XM_011525333.2 XM_011525334.2 XM_011525335.2 XM_011525336.2 XM_011525337.2 XM_011525338.2 XM_017025200.1 XM_017025201.1 XM_017025202.1 XM_017025203.1

SNP Detail

SNP ID Chromosome Location
rs984786105 Chr.17: 61682361-61682361 on Build GRCh38
rs1011164855 Chr.17: 61682050-61682050 on Build GRCh38
rs560959350 Chr.17: 61682380-61682380 on Build GRCh38
rs193165407 Chr.17: 61682421-61682421 on Build GRCh38
rs905772379 Chr.17: 61682076-61682076 on Build GRCh38
rs372430235 Chr.17: 61682063-61682063 on Build GRCh38
rs956280447 Chr.17: 61682373-61682373 on Build GRCh38
rs545980168 Chr.17: 61682006-61682006 on Build GRCh38
rs989473300 Chr.17: 61682396-61682396 on Build GRCh38
rs965315430 Chr.17: 61682419-61682419 on Build GRCh38
rs1026366869 Chr.17: 61682220-61682220 on Build GRCh38
rs577034037 Chr.17: 61681983-61681983 on Build GRCh38
rs964479619 Chr.17: 61682296-61682296 on Build GRCh38
rs886053210 Chr.17: 61682396-61682396 on Build GRCh38
rs543935297 Chr.17: 61682273-61682273 on Build GRCh38
rs993835069 Chr.17: 61682163-61682163 on Build GRCh38
rs747741322 Chr.17: 61682399-61682399 on Build GRCh38
rs886053209 Chr.17: 61682038-61682039 on Build GRCh38
rs1017351247 Chr.17: 61682278-61682278 on Build GRCh38
rs1042958451 Chr.17: 61682057-61682057 on Build GRCh38
rs575280579 Chr.17: 61682196-61682196 on Build GRCh38
rs959876498 Chr.17: 61682128-61682128 on Build GRCh38
rs896384839 Chr.17: 61682144-61682144 on Build GRCh38
rs374389912 Chr.17: 61682079-61682079 on Build GRCh38
rs564290244 Chr.17: 61682246-61682248 on Build GRCh38
rs909495802 Chr.17: 61682405-61682405 on Build GRCh38
rs1047353293 Chr.17: 61682173-61682173 on Build GRCh38
rs891476575 Chr.17: 61682041-61682041 on Build GRCh38
rs937834208 Chr.17: 61682114-61682114 on Build GRCh38
rs374318992 Chr.17: 61682045-61682045 on Build GRCh38
rs1035913949 Chr.17: 61682119-61682119 on Build GRCh38
rs367962443 Chr.17: 61682318-61682318 on Build GRCh38
rs571471772 Chr.17: 61682394-61682394 on Build GRCh38
rs746812029 Chr.17: 61681998-61681998 on Build GRCh38
rs73991940 Chr.17: 61682033-61682033 on Build GRCh38
rs113940104 Chr.17: 61682094-61682094 on Build GRCh38
rs560881577 Chr.17: 61682108-61682111 on Build GRCh38

View Full Product Details